Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 190629216-190629723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RAP
Entrez Gene ID
3556
Gene Name
interleukin 1 receptor accessory protein
Gene Aliases
C3orf13, IL-1RAcP, IL1R3
UniGene
Hs.478673
Gene Chromosome Location
Chr.3: 190514051-190657197 on Build GRCh38
Transcripts
NM_001167928.1 NM_001167929.1 NM_001167930.1 NM_001167931.1 NM_002182.3 NM_134470.3 XM_017006347.1 XM_017006348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4692251 c.943A>C p.I315L Chr.3: 190629390-190629390 on Build GRCh38
6468764 c.979delA p.K327fs*21 Chr.3: 190629426-190629426 on Build GRCh38
5953869 c.968C>T p.S323F Chr.3: 190629415-190629415 on Build GRCh38
4692250 c.943A>C p.I315L Chr.3: 190629390-190629390 on Build GRCh38
5953868 c.968C>T p.S323F Chr.3: 190629415-190629415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757378653 Chr.3: 190629547-190629547 on Build GRCh38
rs143764038 Chr.3: 190629605-190629605 on Build GRCh38
rs765289775 Chr.3: 190629427-190629427 on Build GRCh38
rs764474487 Chr.3: 190629667-190629667 on Build GRCh38
rs749023924 Chr.3: 190629255-190629255 on Build GRCh38
rs779917083 Chr.3: 190629240-190629240 on Build GRCh38
rs756444808 Chr.3: 190629437-190629437 on Build GRCh38
rs35274315 Chr.3: 190629549-190629550 on Build GRCh38
rs150751940 Chr.3: 190629451-190629451 on Build GRCh38
rs977726289 Chr.3: 190629241-190629241 on Build GRCh38
rs758948979 Chr.3: 190629564-190629564 on Build GRCh38
rs750120679 Chr.3: 190629429-190629429 on Build GRCh38
rs866815005 Chr.3: 190629428-190629428 on Build GRCh38
rs148996090 Chr.3: 190629458-190629458 on Build GRCh38
rs1038391920 Chr.3: 190629572-190629572 on Build GRCh38
rs148674484 Chr.3: 190629388-190629388 on Build GRCh38
rs768409834 Chr.3: 190629509-190629509 on Build GRCh38
rs978415111 Chr.3: 190629338-190629338 on Build GRCh38
rs556992311 Chr.3: 190629689-190629689 on Build GRCh38
rs780432173 Chr.3: 190629486-190629486 on Build GRCh38
rs779485745 Chr.3: 190629463-190629463 on Build GRCh38
rs573615292 Chr.3: 190629335-190629338 on Build GRCh38
rs751448745 Chr.3: 190629535-190629535 on Build GRCh38
rs530481089 Chr.3: 190629403-190629403 on Build GRCh38
rs769563293 Chr.3: 190629514-190629514 on Build GRCh38
rs35968058 Chr.3: 190629389-190629390 on Build GRCh38
rs145987111 Chr.3: 190629363-190629363 on Build GRCh38
rs187380478 Chr.3: 190629300-190629300 on Build GRCh38
rs768015847 Chr.3: 190629405-190629405 on Build GRCh38
rs746695778 Chr.3: 190629256-190629256 on Build GRCh38
rs547198945 Chr.3: 190629569-190629569 on Build GRCh38
rs746637562 Chr.3: 190629505-190629505 on Build GRCh38
rs756287441 Chr.3: 190629305-190629305 on Build GRCh38
rs532680154 Chr.3: 190629657-190629657 on Build GRCh38
rs368452306 Chr.3: 190629697-190629697 on Build GRCh38
rs1032660836 Chr.3: 190629687-190629687 on Build GRCh38
rs761639965 Chr.3: 190629418-190629418 on Build GRCh38
rs766246214 Chr.3: 190629529-190629529 on Build GRCh38
rs754008838 Chr.3: 190629453-190629453 on Build GRCh38
rs139016934 Chr.3: 190629454-190629454 on Build GRCh38
rs763259082 Chr.3: 190629660-190629660 on Build GRCh38
rs555401241 Chr.3: 190629647-190629647 on Build GRCh38
rs752999984 Chr.3: 190629434-190629434 on Build GRCh38
rs1056035494 Chr.3: 190629609-190629609 on Build GRCh38
rs749727569 Chr.3: 190629318-190629318 on Build GRCh38
rs778304059 Chr.3: 190629307-190629307 on Build GRCh38
rs775997818 Chr.3: 190629510-190629510 on Build GRCh38
rs184033247 Chr.3: 190629281-190629281 on Build GRCh38
rs780380929 Chr.3: 190629568-190629568 on Build GRCh38
rs746181729 Chr.3: 190629475-190629475 on Build GRCh38
rs561709068 Chr.3: 190629349-190629349 on Build GRCh38
rs750894731 Chr.3: 190629562-190629562 on Build GRCh38
rs776746299 Chr.3: 190629512-190629512 on Build GRCh38
rs924929877 Chr.3: 190629272-190629272 on Build GRCh38
rs545948454 Chr.3: 190629248-190629248 on Build GRCh38
rs571100036 Chr.3: 190629537-190629537 on Build GRCh38
rs142111398 Chr.3: 190629430-190629430 on Build GRCh38
rs774566906 Chr.3: 190629350-190629350 on Build GRCh38
rs1001485213 Chr.3: 190629603-190629603 on Build GRCh38
rs762800736 Chr.3: 190629528-190629528 on Build GRCh38
rs772456241 Chr.3: 190629485-190629485 on Build GRCh38
rs895686195 Chr.3: 190629616-190629616 on Build GRCh38
rs779246678 Chr.3: 190629560-190629560 on Build GRCh38
rs190520539 Chr.3: 190629543-190629543 on Build GRCh38
rs7611887 Chr.3: 190629602-190629602 on Build GRCh38
rs138390252 Chr.3: 190629672-190629672 on Build GRCh38
rs776251137 Chr.3: 190629407-190629407 on Build GRCh38
rs528224035 Chr.3: 190629296-190629296 on Build GRCh38
rs752310735 Chr.3: 190629254-190629270 on Build GRCh38
rs964827481 Chr.3: 190629470-190629470 on Build GRCh38
rs376236710 Chr.3: 190629432-190629432 on Build GRCh38
rs748143568 Chr.3: 190629398-190629398 on Build GRCh38

View Full Product Details