Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 190853025-190853523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMNC
Entrez Gene ID
647309
Gene Name
geminin coiled-coil domain containing
Gene Aliases
GEMC1
UniGene
Hs.518490
Gene Chromosome Location
Chr.3: 190843271-190864200 on Build GRCh38
Transcripts
NM_001146686.2

SNP Detail

SNP ID Chromosome Location
rs929823502 Chr.3: 190853304-190853304 on Build GRCh38
rs1021225124 Chr.3: 190853333-190853333 on Build GRCh38
rs964425708 Chr.3: 190853406-190853406 on Build GRCh38
rs967091712 Chr.3: 190853457-190853457 on Build GRCh38
rs1047666632 Chr.3: 190853161-190853161 on Build GRCh38
rs776810482 Chr.3: 190853352-190853352 on Build GRCh38
rs975837887 Chr.3: 190853475-190853475 on Build GRCh38
rs925541020 Chr.3: 190853143-190853143 on Build GRCh38
rs191321298 Chr.3: 190853066-190853066 on Build GRCh38
rs918636670 Chr.3: 190853285-190853285 on Build GRCh38
rs760925293 Chr.3: 190853123-190853123 on Build GRCh38
rs544464506 Chr.3: 190853180-190853180 on Build GRCh38
rs979568085 Chr.3: 190853069-190853069 on Build GRCh38
rs916618012 Chr.3: 190853261-190853261 on Build GRCh38
rs564152998 Chr.3: 190853191-190853191 on Build GRCh38
rs2173911 Chr.3: 190853353-190853353 on Build GRCh38
rs772193329 Chr.3: 190853085-190853085 on Build GRCh38
rs997814429 Chr.3: 190853430-190853430 on Build GRCh38
rs568803687 Chr.3: 190853476-190853476 on Build GRCh38
rs765139301 Chr.3: 190853374-190853374 on Build GRCh38
rs1038429938 Chr.3: 190853264-190853264 on Build GRCh38
rs1002329078 Chr.3: 190853274-190853274 on Build GRCh38
rs542534462 Chr.3: 190853241-190853244 on Build GRCh38
rs1040018884 Chr.3: 190853420-190853420 on Build GRCh38
rs1020056574 Chr.3: 190853441-190853441 on Build GRCh38
rs56660472 Chr.3: 190853095-190853095 on Build GRCh38
rs371479483 Chr.3: 190853130-190853130 on Build GRCh38
rs9813896 Chr.3: 190853311-190853311 on Build GRCh38
rs929204583 Chr.3: 190853141-190853141 on Build GRCh38
rs900876180 Chr.3: 190853428-190853428 on Build GRCh38
rs941385981 Chr.3: 190853262-190853262 on Build GRCh38
rs552774939 Chr.3: 190853093-190853093 on Build GRCh38
rs760716870 Chr.3: 190853182-190853182 on Build GRCh38
rs991465804 Chr.3: 190853205-190853205 on Build GRCh38
rs535937039 Chr.3: 190853109-190853130 on Build GRCh38
rs1000493906 Chr.3: 190853376-190853376 on Build GRCh38
rs1010216402 Chr.3: 190853327-190853327 on Build GRCh38
rs548579947 Chr.3: 190853433-190853433 on Build GRCh38
rs145656937 Chr.3: 190853272-190853272 on Build GRCh38
rs896681625 Chr.3: 190853315-190853315 on Build GRCh38
rs530728743 Chr.3: 190853105-190853105 on Build GRCh38
rs1017267861 Chr.3: 190853383-190853383 on Build GRCh38
rs543046003 Chr.3: 190853086-190853086 on Build GRCh38
rs528911681 Chr.3: 190853359-190853359 on Build GRCh38

View Full Product Details