Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 195757063-195757556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC4
Entrez Gene ID
4585
Gene Name
mucin 4, cell surface associated
Gene Aliases
ASGP, HSA276359, MUC-4
UniGene
Hs.369646
Gene Chromosome Location
Chr.3: 195746765-195811973 on Build GRCh38
Transcripts
NM_004532.5 NM_138297.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6369727 c.15142A>G p.S5048G Chr.3: 195757173-195757173 on Build GRCh38
1736047 c.15053G>C p.S5018T Chr.3: 195757262-195757262 on Build GRCh38
7160848 c.2407G>C p.E803Q Chr.3: 195757200-195757200 on Build GRCh38
6369728 c.2434A>G p.S812G Chr.3: 195757173-195757173 on Build GRCh38
4188801 c.15020T>C p.L5007P Chr.3: 195757295-195757295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897941346 Chr.3: 195757297-195757297 on Build GRCh38
rs748432606 Chr.3: 195757433-195757433 on Build GRCh38
rs767413135 Chr.3: 195757351-195757351 on Build GRCh38
rs771638420 Chr.3: 195757190-195757191 on Build GRCh38
rs745806425 Chr.3: 195757125-195757126 on Build GRCh38
rs925855249 Chr.3: 195757359-195757359 on Build GRCh38
rs779711379 Chr.3: 195757096-195757096 on Build GRCh38
rs1026373572 Chr.3: 195757493-195757493 on Build GRCh38
rs758206299 Chr.3: 195757181-195757181 on Build GRCh38
rs772409519 Chr.3: 195757273-195757273 on Build GRCh38
rs1017584007 Chr.3: 195757140-195757140 on Build GRCh38
rs779306993 Chr.3: 195757397-195757397 on Build GRCh38
rs761452349 Chr.3: 195757306-195757306 on Build GRCh38
rs747197338 Chr.3: 195757527-195757527 on Build GRCh38
rs909439301 Chr.3: 195757219-195757219 on Build GRCh38
rs543770287 Chr.3: 195757276-195757276 on Build GRCh38
rs777881591 Chr.3: 195757522-195757522 on Build GRCh38
rs761362941 Chr.3: 195757166-195757166 on Build GRCh38
rs574035882 Chr.3: 195757336-195757336 on Build GRCh38
rs764809381 Chr.3: 195757314-195757314 on Build GRCh38
rs199791027 Chr.3: 195757179-195757179 on Build GRCh38
rs373899045 Chr.3: 195757298-195757298 on Build GRCh38
rs564434211 Chr.3: 195757394-195757394 on Build GRCh38
rs903749046 Chr.3: 195757451-195757451 on Build GRCh38
rs531530612 Chr.3: 195757412-195757412 on Build GRCh38
rs112358974 Chr.3: 195757518-195757518 on Build GRCh38
rs370459734 Chr.3: 195757187-195757187 on Build GRCh38
rs368191941 Chr.3: 195757379-195757379 on Build GRCh38
rs956516688 Chr.3: 195757499-195757499 on Build GRCh38
rs966380772 Chr.3: 195757246-195757246 on Build GRCh38
rs879430537 Chr.3: 195757410-195757410 on Build GRCh38
rs111724668 Chr.3: 195757308-195757308 on Build GRCh38
rs747688465 Chr.3: 195757121-195757121 on Build GRCh38
rs763696983 Chr.3: 195757139-195757139 on Build GRCh38
rs150404508 Chr.3: 195757103-195757103 on Build GRCh38
rs772487998 Chr.3: 195757130-195757130 on Build GRCh38
rs201734576 Chr.3: 195757101-195757101 on Build GRCh38
rs753457899 Chr.3: 195757146-195757146 on Build GRCh38
rs762973968 Chr.3: 195757333-195757333 on Build GRCh38
rs367749099 Chr.3: 195757319-195757319 on Build GRCh38
rs979064813 Chr.3: 195757094-195757094 on Build GRCh38
rs758473241 Chr.3: 195757378-195757378 on Build GRCh38
rs376737348 Chr.3: 195757183-195757183 on Build GRCh38
rs182917285 Chr.3: 195757384-195757384 on Build GRCh38
rs754874334 Chr.3: 195757337-195757337 on Build GRCh38
rs147269033 Chr.3: 195757524-195757524 on Build GRCh38
rs2550270 Chr.3: 195757373-195757373 on Build GRCh38
rs775168285 Chr.3: 195757192-195757193 on Build GRCh38
rs1030659937 Chr.3: 195757482-195757482 on Build GRCh38
rs1000558466 Chr.3: 195757466-195757466 on Build GRCh38
rs760352152 Chr.3: 195757196-195757198 on Build GRCh38
rs976306104 Chr.3: 195757156-195757156 on Build GRCh38
rs762713854 Chr.3: 195757196-195757196 on Build GRCh38
rs576334017 Chr.3: 195757236-195757236 on Build GRCh38
rs760341199 Chr.3: 195757132-195757132 on Build GRCh38
rs778026546 Chr.3: 195757110-195757110 on Build GRCh38
rs879945190 Chr.3: 195757414-195757414 on Build GRCh38
rs73892860 Chr.3: 195757372-195757372 on Build GRCh38
rs757475796 Chr.3: 195757242-195757242 on Build GRCh38
rs747413793 Chr.3: 195757291-195757291 on Build GRCh38
rs796636788 Chr.3: 195757398-195757398 on Build GRCh38
rs138139346 Chr.3: 195757205-195757205 on Build GRCh38
rs373628892 Chr.3: 195757335-195757335 on Build GRCh38
rs768987413 Chr.3: 195757295-195757295 on Build GRCh38
rs748959469 Chr.3: 195757336-195757336 on Build GRCh38
rs1005718885 Chr.3: 195757406-195757406 on Build GRCh38
rs778789896 Chr.3: 195757243-195757243 on Build GRCh38
rs552494023 Chr.3: 195757165-195757165 on Build GRCh38
rs954441423 Chr.3: 195757244-195757244 on Build GRCh38
rs964548976 Chr.3: 195757534-195757534 on Build GRCh38
rs763620428 Chr.3: 195757320-195757320 on Build GRCh38
rs775658590 Chr.3: 195757131-195757131 on Build GRCh38
rs749415350 Chr.3: 195757111-195757111 on Build GRCh38
rs187255664 Chr.3: 195757436-195757436 on Build GRCh38
rs368926047 Chr.3: 195757361-195757361 on Build GRCh38
rs112337020 Chr.3: 195757218-195757218 on Build GRCh38
rs201691250 Chr.3: 195757284-195757284 on Build GRCh38
rs759954038 Chr.3: 195757128-195757128 on Build GRCh38
rs954366774 Chr.3: 195757500-195757500 on Build GRCh38
rs754114153 Chr.3: 195757356-195757356 on Build GRCh38
rs746397929 Chr.3: 195757262-195757262 on Build GRCh38

View Full Product Details