Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196233245-196233755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC51A
Entrez Gene ID
200931
Gene Name
solute carrier family 51 alpha subunit
Gene Aliases
OSTA, OSTalpha
UniGene
Hs.630585
Gene Chromosome Location
Chr.3: 196216512-196233430 on Build GRCh38
Transcripts
NM_152672.5

SNP Detail

SNP ID Chromosome Location
rs943378493 Chr.3: 196233406-196233406 on Build GRCh38
rs919231354 Chr.3: 196233305-196233305 on Build GRCh38
rs978964186 Chr.3: 196233721-196233721 on Build GRCh38
rs1032753399 Chr.3: 196233555-196233555 on Build GRCh38
rs1048367139 Chr.3: 196233472-196233473 on Build GRCh38
rs772467537 Chr.3: 196233717-196233717 on Build GRCh38
rs943588884 Chr.3: 196233348-196233348 on Build GRCh38
rs910567324 Chr.3: 196233387-196233388 on Build GRCh38
rs993604502 Chr.3: 196233560-196233560 on Build GRCh38
rs892157126 Chr.3: 196233457-196233457 on Build GRCh38
rs920712320 Chr.3: 196233432-196233433 on Build GRCh38
rs987466659 Chr.3: 196233614-196233614 on Build GRCh38
rs752209789 Chr.3: 196233302-196233302 on Build GRCh38
rs534856838 Chr.3: 196233315-196233315 on Build GRCh38
rs955988896 Chr.3: 196233557-196233557 on Build GRCh38
rs556128910 Chr.3: 196233726-196233726 on Build GRCh38
rs1029520385 Chr.3: 196233574-196233574 on Build GRCh38
rs1013193928 Chr.3: 196233469-196233469 on Build GRCh38
rs925061663 Chr.3: 196233385-196233385 on Build GRCh38
rs538649315 Chr.3: 196233624-196233624 on Build GRCh38
rs767050966 Chr.3: 196233564-196233564 on Build GRCh38
rs111367094 Chr.3: 196233608-196233608 on Build GRCh38
rs987749901 Chr.3: 196233319-196233319 on Build GRCh38
rs911163494 Chr.3: 196233616-196233616 on Build GRCh38
rs771136069 Chr.3: 196233496-196233496 on Build GRCh38
rs1014473687 Chr.3: 196233493-196233493 on Build GRCh38
rs1002046423 Chr.3: 196233497-196233497 on Build GRCh38
rs1026387887 Chr.3: 196233569-196233569 on Build GRCh38
rs371533940 Chr.3: 196233655-196233655 on Build GRCh38
rs963508259 Chr.3: 196233540-196233540 on Build GRCh38
rs896048362 Chr.3: 196233487-196233487 on Build GRCh38
rs1046334393 Chr.3: 196233494-196233494 on Build GRCh38
rs34711276 Chr.3: 196233518-196233519 on Build GRCh38
rs377656557 Chr.3: 196233435-196233435 on Build GRCh38
rs929459137 Chr.3: 196233467-196233467 on Build GRCh38
rs910805181 Chr.3: 196233682-196233682 on Build GRCh38
rs779214427 Chr.3: 196233539-196233539 on Build GRCh38
rs1000307330 Chr.3: 196233508-196233508 on Build GRCh38
rs746087609 Chr.3: 196233586-196233586 on Build GRCh38
rs936394837 Chr.3: 196233422-196233422 on Build GRCh38
rs964734559 Chr.3: 196233648-196233648 on Build GRCh38
rs9343 Chr.3: 196233354-196233354 on Build GRCh38
rs910787704 Chr.3: 196233332-196233332 on Build GRCh38
rs955245976 Chr.3: 196233606-196233606 on Build GRCh38

View Full Product Details