Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2257902-2258404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MXD4
Entrez Gene ID
10608
Gene Name
MAX dimerization protein 4
Gene Aliases
MAD4, MST149, MSTP149, bHLHc12
UniGene
Hs.655020
Gene Chromosome Location
Chr.4: 2247432-2262143 on Build GRCh38
Transcripts
NM_006454.2 XM_011513388.2 XM_017007655.1 XM_017007656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317026 c.184G>A p.E62K Chr.4: 2257992-2257992 on Build GRCh38
3428436 c.167C>A p.S56Y Chr.4: 2258009-2258009 on Build GRCh38
3946283 c.165-5A>G p.? Chr.4: 2258016-2258016 on Build GRCh38
6718384 c.178G>A p.E60K Chr.4: 2257998-2257998 on Build GRCh38
3825691 c.170C>T p.S57L Chr.4: 2258006-2258006 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs939536990 Chr.4: 2258231-2258231 on Build GRCh38
rs1031605681 Chr.4: 2258061-2258061 on Build GRCh38
rs546966527 Chr.4: 2258353-2258353 on Build GRCh38
rs371930251 Chr.4: 2258251-2258251 on Build GRCh38
rs774766563 Chr.4: 2257960-2257960 on Build GRCh38
rs146792190 Chr.4: 2258120-2258120 on Build GRCh38
rs746203844 Chr.4: 2258044-2258044 on Build GRCh38
rs987377956 Chr.4: 2258301-2258302 on Build GRCh38
rs915223089 Chr.4: 2258148-2258148 on Build GRCh38
rs2233037 Chr.4: 2257923-2257923 on Build GRCh38
rs571551358 Chr.4: 2258000-2258000 on Build GRCh38
rs148346640 Chr.4: 2258166-2258166 on Build GRCh38
rs780675048 Chr.4: 2258031-2258031 on Build GRCh38
rs73797001 Chr.4: 2257927-2257927 on Build GRCh38
rs994451560 Chr.4: 2258185-2258185 on Build GRCh38
rs940541081 Chr.4: 2258118-2258118 on Build GRCh38
rs201616610 Chr.4: 2257970-2257970 on Build GRCh38
rs891749171 Chr.4: 2258308-2258308 on Build GRCh38
rs773650975 Chr.4: 2257940-2257940 on Build GRCh38
rs1025658837 Chr.4: 2258233-2258233 on Build GRCh38
rs753466039 Chr.4: 2258250-2258250 on Build GRCh38
rs759726955 Chr.4: 2257969-2257969 on Build GRCh38
rs73083193 Chr.4: 2258266-2258266 on Build GRCh38
rs879698516 Chr.4: 2258261-2258261 on Build GRCh38
rs778897633 Chr.4: 2258005-2258005 on Build GRCh38
rs956136453 Chr.4: 2258088-2258088 on Build GRCh38
rs760763648 Chr.4: 2257973-2257973 on Build GRCh38
rs752219549 Chr.4: 2258026-2258027 on Build GRCh38
rs760844904 Chr.4: 2258063-2258063 on Build GRCh38
rs2233036 Chr.4: 2258035-2258035 on Build GRCh38
rs73797002 Chr.4: 2258090-2258090 on Build GRCh38
rs375171530 Chr.4: 2258260-2258260 on Build GRCh38
rs919535072 Chr.4: 2258241-2258241 on Build GRCh38
rs182798422 Chr.4: 2258378-2258378 on Build GRCh38
rs754417107 Chr.4: 2258019-2258019 on Build GRCh38
rs141483722 Chr.4: 2258198-2258198 on Build GRCh38
rs540608295 Chr.4: 2258169-2258169 on Build GRCh38
rs1036048096 Chr.4: 2258176-2258176 on Build GRCh38
rs868816331 Chr.4: 2258253-2258253 on Build GRCh38
rs544807372 Chr.4: 2258109-2258109 on Build GRCh38
rs566728250 Chr.4: 2258363-2258363 on Build GRCh38
rs1012463896 Chr.4: 2258204-2258204 on Build GRCh38
rs953510831 Chr.4: 2258281-2258281 on Build GRCh38
rs559099440 Chr.4: 2258072-2258072 on Build GRCh38
rs763311685 Chr.4: 2257945-2257945 on Build GRCh38
rs989477435 Chr.4: 2258264-2258264 on Build GRCh38
rs943011281 Chr.4: 2258257-2258257 on Build GRCh38
rs764143961 Chr.4: 2257976-2257976 on Build GRCh38
rs1052117929 Chr.4: 2258059-2258059 on Build GRCh38
rs767427982 Chr.4: 2257949-2257949 on Build GRCh38
rs752751316 Chr.4: 2257972-2257972 on Build GRCh38
rs760347451 Chr.4: 2258030-2258033 on Build GRCh38
rs968453233 Chr.4: 2258319-2258319 on Build GRCh38
rs866238576 Chr.4: 2257922-2257922 on Build GRCh38
rs1056547205 Chr.4: 2258237-2258237 on Build GRCh38
rs1011898524 Chr.4: 2258316-2258316 on Build GRCh38
rs190673921 Chr.4: 2258181-2258181 on Build GRCh38
rs753764925 Chr.4: 2257998-2257998 on Build GRCh38
rs771407992 Chr.4: 2258040-2258040 on Build GRCh38
rs757259427 Chr.4: 2257999-2257999 on Build GRCh38
rs775727690 Chr.4: 2258058-2258058 on Build GRCh38
rs980827180 Chr.4: 2258197-2258197 on Build GRCh38
rs1021506473 Chr.4: 2258317-2258317 on Build GRCh38
rs765581487 Chr.4: 2258212-2258212 on Build GRCh38
rs544563337 Chr.4: 2258242-2258242 on Build GRCh38
rs971309090 Chr.4: 2257930-2257930 on Build GRCh38
rs530496105 Chr.4: 2258259-2258259 on Build GRCh38
rs752205168 Chr.4: 2258076-2258076 on Build GRCh38
rs529889664 Chr.4: 2258346-2258346 on Build GRCh38
rs992327939 Chr.4: 2258370-2258370 on Build GRCh38
rs779704621 Chr.4: 2258016-2258016 on Build GRCh38
rs758014445 Chr.4: 2258014-2258014 on Build GRCh38
rs770304208 Chr.4: 2257938-2257938 on Build GRCh38
rs774619906 Chr.4: 2258042-2258042 on Build GRCh38
rs752932233 Chr.4: 2258265-2258265 on Build GRCh38
rs940511480 Chr.4: 2258334-2258334 on Build GRCh38
rs200832798 Chr.4: 2258011-2258011 on Build GRCh38
rs772205963 Chr.4: 2258053-2258053 on Build GRCh38
rs567575145 Chr.4: 2257928-2257928 on Build GRCh38
rs1033756549 Chr.4: 2258360-2258360 on Build GRCh38
rs114820443 Chr.4: 2258017-2258017 on Build GRCh38
rs567384574 Chr.4: 2258297-2258297 on Build GRCh38

View Full Product Details