Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 4457064-4457570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STX18
Entrez Gene ID
53407
Gene Name
syntaxin 18
Gene Aliases
Ufe1
UniGene
Hs.584913
Gene Chromosome Location
Chr.4: 4418968-4542338 on Build GRCh38
Transcripts
NM_016930.2 XM_011513480.1 XM_011513483.1 XM_011513485.2 XM_017008296.1 XM_017008297.1 XM_017008298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7392366 c.439A>G p.K147E Chr.4: 4457249-4457249 on Build GRCh38
5807736 c.379G>T p.V127L Chr.4: 4457474-4457474 on Build GRCh38
5941920 c.466C>T p.R156* Chr.4: 4457222-4457222 on Build GRCh38
1429835 c.460G>A p.A154T Chr.4: 4457228-4457228 on Build GRCh38
6792587 c.450A>G p.S150S Chr.4: 4457238-4457238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017369906 Chr.4: 4457094-4457094 on Build GRCh38
rs574044851 Chr.4: 4457133-4457133 on Build GRCh38
rs754995865 Chr.4: 4457524-4457524 on Build GRCh38
rs73086249 Chr.4: 4457095-4457095 on Build GRCh38
rs781186059 Chr.4: 4457151-4457151 on Build GRCh38
rs765304584 Chr.4: 4457528-4457528 on Build GRCh38
rs1006699133 Chr.4: 4457312-4457312 on Build GRCh38
rs564846043 Chr.4: 4457336-4457336 on Build GRCh38
rs780087802 Chr.4: 4457269-4457269 on Build GRCh38
rs768109962 Chr.4: 4457492-4457492 on Build GRCh38
rs373971847 Chr.4: 4457399-4457399 on Build GRCh38
rs1039545295 Chr.4: 4457315-4457315 on Build GRCh38
rs369817269 Chr.4: 4457264-4457264 on Build GRCh38
rs894691201 Chr.4: 4457343-4457343 on Build GRCh38
rs200389671 Chr.4: 4457497-4457497 on Build GRCh38
rs777715238 Chr.4: 4457180-4457188 on Build GRCh38
rs766290903 Chr.4: 4457516-4457516 on Build GRCh38
rs755992642 Chr.4: 4457440-4457440 on Build GRCh38
rs761106265 Chr.4: 4457193-4457193 on Build GRCh38
rs375609558 Chr.4: 4457444-4457444 on Build GRCh38
rs749364308 Chr.4: 4457284-4457284 on Build GRCh38
rs761432951 Chr.4: 4457495-4457495 on Build GRCh38
rs190759970 Chr.4: 4457529-4457529 on Build GRCh38
rs772429275 Chr.4: 4457380-4457380 on Build GRCh38
rs758565155 Chr.4: 4457532-4457532 on Build GRCh38
rs941705297 Chr.4: 4457485-4457485 on Build GRCh38
rs773738573 Chr.4: 4457537-4457537 on Build GRCh38
rs1021337216 Chr.4: 4457328-4457328 on Build GRCh38
rs750328289 Chr.4: 4457259-4457259 on Build GRCh38
rs745798543 Chr.4: 4457152-4457152 on Build GRCh38
rs746392099 Chr.4: 4457490-4457490 on Build GRCh38
rs1049195112 Chr.4: 4457301-4457301 on Build GRCh38
rs780154919 Chr.4: 4457096-4457096 on Build GRCh38
rs1000877913 Chr.4: 4457371-4457371 on Build GRCh38
rs114891587 Chr.4: 4457533-4457533 on Build GRCh38
rs889222893 Chr.4: 4457311-4457311 on Build GRCh38
rs777055880 Chr.4: 4457489-4457489 on Build GRCh38
rs763726059 Chr.4: 4457246-4457246 on Build GRCh38
rs771322961 Chr.4: 4457397-4457397 on Build GRCh38
rs919230478 Chr.4: 4457538-4457538 on Build GRCh38
rs148358484 Chr.4: 4457318-4457318 on Build GRCh38
rs1036767581 Chr.4: 4457226-4457226 on Build GRCh38
rs571862656 Chr.4: 4457457-4457457 on Build GRCh38
rs763599104 Chr.4: 4457401-4457401 on Build GRCh38
rs141506981 Chr.4: 4457396-4457396 on Build GRCh38
rs768923669 Chr.4: 4457306-4457306 on Build GRCh38
rs773885817 Chr.4: 4457493-4457493 on Build GRCh38
rs1030706720 Chr.4: 4457091-4457091 on Build GRCh38
rs370164301 Chr.4: 4457453-4457453 on Build GRCh38
rs750030946 Chr.4: 4457156-4457159 on Build GRCh38
rs773478191 Chr.4: 4457391-4457391 on Build GRCh38
rs751336701 Chr.4: 4457145-4457145 on Build GRCh38
rs181888966 Chr.4: 4457103-4457103 on Build GRCh38
rs781092166 Chr.4: 4457172-4457172 on Build GRCh38
rs772981481 Chr.4: 4457505-4457505 on Build GRCh38
rs773936922 Chr.4: 4457407-4457407 on Build GRCh38
rs758515694 Chr.4: 4457468-4457468 on Build GRCh38
rs919297026 Chr.4: 4457279-4457279 on Build GRCh38
rs200846315 Chr.4: 4457441-4457441 on Build GRCh38
rs778107911 Chr.4: 4457478-4457478 on Build GRCh38
rs774771558 Chr.4: 4457180-4457180 on Build GRCh38
rs139040070 Chr.4: 4457447-4457447 on Build GRCh38
rs761396530 Chr.4: 4457422-4457422 on Build GRCh38
rs756980900 Chr.4: 4457250-4457250 on Build GRCh38
rs939623817 Chr.4: 4457225-4457225 on Build GRCh38
rs764762469 Chr.4: 4457214-4457214 on Build GRCh38
rs956397351 Chr.4: 4457365-4457365 on Build GRCh38
rs77097979 Chr.4: 4457327-4457327 on Build GRCh38
rs780022085 Chr.4: 4457171-4457171 on Build GRCh38
rs376671232 Chr.4: 4457174-4457174 on Build GRCh38
rs752175452 Chr.4: 4457219-4457219 on Build GRCh38
rs927068709 Chr.4: 4457518-4457518 on Build GRCh38
rs201174408 Chr.4: 4457168-4457168 on Build GRCh38
rs760316108 Chr.4: 4457515-4457515 on Build GRCh38
rs762502032 Chr.4: 4457238-4457238 on Build GRCh38
rs772324336 Chr.4: 4457188-4457188 on Build GRCh38
rs969507173 Chr.4: 4457118-4457118 on Build GRCh38
rs773707590 Chr.4: 4457191-4457191 on Build GRCh38
rs1031071964 Chr.4: 4457344-4457344 on Build GRCh38
rs376086196 Chr.4: 4457517-4457517 on Build GRCh38
rs779206014 Chr.4: 4457376-4457376 on Build GRCh38
rs748392035 Chr.4: 4457379-4457379 on Build GRCh38
rs767305451 Chr.4: 4457430-4457430 on Build GRCh38
rs757105833 Chr.4: 4457146-4457146 on Build GRCh38
rs930699919 Chr.4: 4457299-4457299 on Build GRCh38
rs750159538 Chr.4: 4457432-4457432 on Build GRCh38
rs745714497 Chr.4: 4457488-4457488 on Build GRCh38
rs187576032 Chr.4: 4457136-4457136 on Build GRCh38
rs201844923 Chr.4: 4457278-4457278 on Build GRCh38
rs556573540 Chr.4: 4457253-4457253 on Build GRCh38
rs768976831 Chr.4: 4457179-4457179 on Build GRCh38
rs747322825 Chr.4: 4457482-4457482 on Build GRCh38
rs755050963 Chr.4: 4457456-4457456 on Build GRCh38

View Full Product Details