Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 5836676-5837167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRMP1
Entrez Gene ID
1400
Gene Name
collapsin response mediator protein 1
Gene Aliases
CRMP-1, DPYSL1, DRP-1, DRP1, ULIP-3
UniGene
Hs.135270
Gene Chromosome Location
Chr.4: 5820764-5893083 on Build GRCh38
Transcripts
NM_001014809.2 NM_001288661.1 NM_001288662.1 NM_001313.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6207488 c.1026G>A p.A342A Chr.4: 5836849-5836849 on Build GRCh38
5023500 c.1332C>A p.G444G Chr.4: 5836885-5836885 on Build GRCh38
4125346 c.1367C>T p.A456V Chr.4: 5836850-5836850 on Build GRCh38
4125347 c.1347C>T p.P449P Chr.4: 5836870-5836870 on Build GRCh38
1242158 c.1331G>A p.G444D Chr.4: 5836886-5836886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984439783 Chr.4: 5837037-5837038 on Build GRCh38
rs548053386 Chr.4: 5836819-5836819 on Build GRCh38
rs999793883 Chr.4: 5837001-5837002 on Build GRCh38
rs764002640 Chr.4: 5836719-5836719 on Build GRCh38
rs370480340 Chr.4: 5836785-5836785 on Build GRCh38
rs963620822 Chr.4: 5836989-5836989 on Build GRCh38
rs761630158 Chr.4: 5836916-5836916 on Build GRCh38
rs564519423 Chr.4: 5836726-5836726 on Build GRCh38
rs566130382 Chr.4: 5837046-5837046 on Build GRCh38
rs573563373 Chr.4: 5837064-5837064 on Build GRCh38
rs947484971 Chr.4: 5836930-5836930 on Build GRCh38
rs777709266 Chr.4: 5836745-5836746 on Build GRCh38
rs371310830 Chr.4: 5836850-5836850 on Build GRCh38
rs146254380 Chr.4: 5836849-5836849 on Build GRCh38
rs768478598 Chr.4: 5836894-5836894 on Build GRCh38
rs756250733 Chr.4: 5837136-5837136 on Build GRCh38
rs774155847 Chr.4: 5836900-5836900 on Build GRCh38
rs374326359 Chr.4: 5836881-5836881 on Build GRCh38
rs969134569 Chr.4: 5836781-5836781 on Build GRCh38
rs146972312 Chr.4: 5836828-5836828 on Build GRCh38
rs937156298 Chr.4: 5837073-5837073 on Build GRCh38
rs748624496 Chr.4: 5836774-5836774 on Build GRCh38
rs969019972 Chr.4: 5837033-5837033 on Build GRCh38
rs745308619 Chr.4: 5836783-5836783 on Build GRCh38
rs756157482 Chr.4: 5836745-5836745 on Build GRCh38
rs1029213878 Chr.4: 5837011-5837011 on Build GRCh38
rs896397300 Chr.4: 5837020-5837020 on Build GRCh38
rs141437952 Chr.4: 5836808-5836808 on Build GRCh38
rs755289455 Chr.4: 5836758-5836758 on Build GRCh38
rs1016421343 Chr.4: 5836976-5836976 on Build GRCh38
rs571660375 Chr.4: 5837024-5837024 on Build GRCh38
rs762836705 Chr.4: 5837008-5837008 on Build GRCh38
rs769323304 Chr.4: 5836875-5836875 on Build GRCh38
rs1043098997 Chr.4: 5836980-5836980 on Build GRCh38
rs757298363 Chr.4: 5836728-5836728 on Build GRCh38
rs537203686 Chr.4: 5837051-5837051 on Build GRCh38
rs12331 Chr.4: 5836786-5836786 on Build GRCh38
rs767820653 Chr.4: 5837045-5837045 on Build GRCh38
rs183322981 Chr.4: 5836872-5836872 on Build GRCh38
rs960641811 Chr.4: 5836713-5836713 on Build GRCh38
rs749546776 Chr.4: 5836750-5836750 on Build GRCh38
rs1023319776 Chr.4: 5837032-5837032 on Build GRCh38
rs767487824 Chr.4: 5836734-5836734 on Build GRCh38
rs1056037201 Chr.4: 5837100-5837100 on Build GRCh38
rs949959919 Chr.4: 5837127-5837127 on Build GRCh38
rs923554051 Chr.4: 5836848-5836848 on Build GRCh38
rs1025331776 Chr.4: 5836910-5836910 on Build GRCh38
rs566109918 Chr.4: 5836896-5836896 on Build GRCh38
rs761824632 Chr.4: 5836807-5836807 on Build GRCh38
rs904384968 Chr.4: 5836827-5836827 on Build GRCh38
rs748941880 Chr.4: 5836882-5836882 on Build GRCh38
rs879888250 Chr.4: 5837058-5837058 on Build GRCh38
rs1005331640 Chr.4: 5836962-5836962 on Build GRCh38
rs755592264 Chr.4: 5836996-5836999 on Build GRCh38
rs7654446 Chr.4: 5837110-5837110 on Build GRCh38
rs779301264 Chr.4: 5836768-5836768 on Build GRCh38
rs760581438 Chr.4: 5836955-5836955 on Build GRCh38
rs551891845 Chr.4: 5837006-5837006 on Build GRCh38
rs773157126 Chr.4: 5836941-5836941 on Build GRCh38
rs779247738 Chr.4: 5836839-5836839 on Build GRCh38
rs774352363 Chr.4: 5836798-5836798 on Build GRCh38
rs750570699 Chr.4: 5836738-5836738 on Build GRCh38
rs139299333 Chr.4: 5837057-5837057 on Build GRCh38
rs199717161 Chr.4: 5836705-5836705 on Build GRCh38
rs369545308 Chr.4: 5836981-5836981 on Build GRCh38
rs992274674 Chr.4: 5836739-5836739 on Build GRCh38
rs533613606 Chr.4: 5836782-5836782 on Build GRCh38
rs771797216 Chr.4: 5836917-5836917 on Build GRCh38
rs780259951 Chr.4: 5836748-5836748 on Build GRCh38
rs778302109 Chr.4: 5836858-5836858 on Build GRCh38
rs750337007 Chr.4: 5836812-5836812 on Build GRCh38

View Full Product Details