Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117605846-117606385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM46C
Entrez Gene ID
54855
Gene Name
family with sequence similarity 46 member C
Gene Aliases
-
UniGene
Hs.356216
Gene Chromosome Location
Chr.1: 117605982-117628389 on Build GRCh38
Transcripts
NM_017709.3

SNP Detail

SNP ID Chromosome Location
rs978348399 Chr.1: 117606011-117606011 on Build GRCh38
rs907528982 Chr.1: 117606004-117606004 on Build GRCh38
rs974210297 Chr.1: 117605947-117605947 on Build GRCh38
rs945906351 Chr.1: 117606203-117606203 on Build GRCh38
rs956639679 Chr.1: 117605924-117605924 on Build GRCh38
rs1054333012 Chr.1: 117606166-117606166 on Build GRCh38
rs913263161 Chr.1: 117605956-117605956 on Build GRCh38
rs911187464 Chr.1: 117606204-117606204 on Build GRCh38
rs931916723 Chr.1: 117606019-117606019 on Build GRCh38
rs548124280 Chr.1: 117606080-117606080 on Build GRCh38
rs921584073 Chr.1: 117606295-117606295 on Build GRCh38
rs904943465 Chr.1: 117606087-117606087 on Build GRCh38
rs927559775 Chr.1: 117605938-117605938 on Build GRCh38
rs576977713 Chr.1: 117606152-117606152 on Build GRCh38
rs1034820245 Chr.1: 117605920-117605920 on Build GRCh38
rs892970015 Chr.1: 117606178-117606178 on Build GRCh38
rs951561143 Chr.1: 117606054-117606054 on Build GRCh38
rs1040276106 Chr.1: 117606017-117606017 on Build GRCh38
rs1023263829 Chr.1: 117606056-117606056 on Build GRCh38
rs944411836 Chr.1: 117606010-117606010 on Build GRCh38
rs879622443 Chr.1: 117606124-117606124 on Build GRCh38
rs557430573 Chr.1: 117606028-117606028 on Build GRCh38
rs958888880 Chr.1: 117605944-117605944 on Build GRCh38
rs947372507 Chr.1: 117605998-117605998 on Build GRCh38
rs983324247 Chr.1: 117606260-117606260 on Build GRCh38
rs1047324309 Chr.1: 117606041-117606041 on Build GRCh38
rs931864550 Chr.1: 117606165-117606165 on Build GRCh38
rs981713904 Chr.1: 117605935-117605935 on Build GRCh38
rs959190423 Chr.1: 117606040-117606040 on Build GRCh38
rs770029460 Chr.1: 117606278-117606287 on Build GRCh38
rs1003314779 Chr.1: 117606035-117606035 on Build GRCh38
rs562949644 Chr.1: 117606233-117606233 on Build GRCh38
rs1056123355 Chr.1: 117606351-117606351 on Build GRCh38
rs979179658 Chr.1: 117606173-117606173 on Build GRCh38
rs759747369 Chr.1: 117606237-117606252 on Build GRCh38
rs901320399 Chr.1: 117606032-117606032 on Build GRCh38
rs1020207852 Chr.1: 117605921-117605921 on Build GRCh38
rs79358822 Chr.1: 117606029-117606029 on Build GRCh38
rs772201118 Chr.1: 117606151-117606151 on Build GRCh38
rs573196012 Chr.1: 117606030-117606031 on Build GRCh38
rs575667393 Chr.1: 117606043-117606043 on Build GRCh38
rs866799989 Chr.1: 117605898-117605898 on Build GRCh38
rs541250717 Chr.1: 117606179-117606179 on Build GRCh38
rs537344518 Chr.1: 117606049-117606049 on Build GRCh38
rs1027123606 Chr.1: 117606052-117606052 on Build GRCh38
rs375020200 Chr.1: 117606356-117606356 on Build GRCh38
rs976128201 Chr.1: 117606131-117606131 on Build GRCh38
rs975871935 Chr.1: 117606286-117606286 on Build GRCh38
rs879544015 Chr.1: 117606039-117606039 on Build GRCh38
rs535949318 Chr.1: 117605919-117605919 on Build GRCh38
rs1041901898 Chr.1: 117606340-117606340 on Build GRCh38
rs990231148 Chr.1: 117606355-117606355 on Build GRCh38
rs914525154 Chr.1: 117606316-117606316 on Build GRCh38
rs968782967 Chr.1: 117606162-117606162 on Build GRCh38
rs910141768 Chr.1: 117606261-117606261 on Build GRCh38
rs1034829664 Chr.1: 117606095-117606095 on Build GRCh38
rs574518184 Chr.1: 117606216-117606216 on Build GRCh38
rs558861088 Chr.1: 117606102-117606102 on Build GRCh38
rs953074153 Chr.1: 117606304-117606304 on Build GRCh38
rs530397467 Chr.1: 117606243-117606243 on Build GRCh38
rs945974500 Chr.1: 117606336-117606336 on Build GRCh38
rs1054177102 Chr.1: 117606020-117606020 on Build GRCh38
rs987988789 Chr.1: 117605941-117605941 on Build GRCh38
rs1009463897 Chr.1: 117606142-117606142 on Build GRCh38
rs911289653 Chr.1: 117606018-117606018 on Build GRCh38
rs541486204 Chr.1: 117606223-117606223 on Build GRCh38
rs904826342 Chr.1: 117606294-117606294 on Build GRCh38
rs970912901 Chr.1: 117606006-117606006 on Build GRCh38
rs930124032 Chr.1: 117606229-117606229 on Build GRCh38
rs982360356 Chr.1: 117605879-117605879 on Build GRCh38
rs995945576 Chr.1: 117605979-117605979 on Build GRCh38
rs907247395 Chr.1: 117606207-117606207 on Build GRCh38
rs920110596 Chr.1: 117605976-117605976 on Build GRCh38
rs1775815 Chr.1: 117606208-117606208 on Build GRCh38
rs990024112 Chr.1: 117606306-117606306 on Build GRCh38
rs1003732962 Chr.1: 117606319-117606319 on Build GRCh38
rs1034739065 Chr.1: 117606209-117606209 on Build GRCh38
rs924228183 Chr.1: 117606120-117606120 on Build GRCh38
rs918102341 Chr.1: 117606000-117606000 on Build GRCh38
rs990963170 Chr.1: 117605999-117605999 on Build GRCh38
rs942767501 Chr.1: 117606024-117606024 on Build GRCh38
rs959444071 Chr.1: 117605994-117605994 on Build GRCh38
rs976836209 Chr.1: 117606221-117606221 on Build GRCh38
rs572038933 Chr.1: 117605870-117605870 on Build GRCh38
rs1010278256 Chr.1: 117605904-117605904 on Build GRCh38
rs1045147473 Chr.1: 117606268-117606268 on Build GRCh38
rs1027034667 Chr.1: 117605989-117605989 on Build GRCh38
rs1003269787 Chr.1: 117606090-117606090 on Build GRCh38
rs941581247 Chr.1: 117606265-117606265 on Build GRCh38

View Full Product Details