Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117625038-117625549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM46C
Entrez Gene ID
54855
Gene Name
family with sequence similarity 46 member C
Gene Aliases
-
UniGene
Hs.356216
Gene Chromosome Location
Chr.1: 117605982-117628389 on Build GRCh38
Transcripts
NM_017709.3

SNP Detail

SNP ID Chromosome Location
rs967631164 Chr.1: 117625463-117625463 on Build GRCh38
rs566629153 Chr.1: 117625402-117625402 on Build GRCh38
rs1039047473 Chr.1: 117625491-117625491 on Build GRCh38
rs761311224 Chr.1: 117625229-117625229 on Build GRCh38
rs1026361564 Chr.1: 117625159-117625159 on Build GRCh38
rs536102125 Chr.1: 117625389-117625389 on Build GRCh38
rs76074370 Chr.1: 117625284-117625284 on Build GRCh38
rs977542298 Chr.1: 117625464-117625464 on Build GRCh38
rs764207919 Chr.1: 117625459-117625459 on Build GRCh38
rs530987102 Chr.1: 117625147-117625147 on Build GRCh38
rs772630462 Chr.1: 117625176-117625176 on Build GRCh38
rs200998786 Chr.1: 117625118-117625118 on Build GRCh38
rs190035193 Chr.1: 117625177-117625177 on Build GRCh38
rs1012009581 Chr.1: 117625416-117625416 on Build GRCh38
rs569128175 Chr.1: 117625146-117625146 on Build GRCh38
rs942610549 Chr.1: 117625473-117625473 on Build GRCh38
rs776960087 Chr.1: 117625468-117625468 on Build GRCh38
rs397975061 Chr.1: 117625117-117625118 on Build GRCh38
rs988974635 Chr.1: 117625285-117625285 on Build GRCh38
rs970281223 Chr.1: 117625195-117625195 on Build GRCh38
rs370137032 Chr.1: 117625353-117625353 on Build GRCh38
rs142726505 Chr.1: 117625148-117625148 on Build GRCh38
rs889675726 Chr.1: 117625064-117625064 on Build GRCh38
rs370760304 Chr.1: 117625483-117625483 on Build GRCh38
rs1021848779 Chr.1: 117625417-117625417 on Build GRCh38
rs979794118 Chr.1: 117625228-117625228 on Build GRCh38
rs967547967 Chr.1: 117625119-117625119 on Build GRCh38
rs912950361 Chr.1: 117625289-117625289 on Build GRCh38
rs1007537476 Chr.1: 117625067-117625067 on Build GRCh38
rs563673641 Chr.1: 117625124-117625124 on Build GRCh38
rs775963518 Chr.1: 117625189-117625189 on Build GRCh38
rs555152244 Chr.1: 117625105-117625106 on Build GRCh38
rs901967646 Chr.1: 117625144-117625144 on Build GRCh38
rs1042850704 Chr.1: 117625386-117625386 on Build GRCh38
rs565945418 Chr.1: 117625376-117625376 on Build GRCh38
rs754223021 Chr.1: 117625445-117625445 on Build GRCh38
rs947225693 Chr.1: 117625344-117625344 on Build GRCh38
rs1017544969 Chr.1: 117625117-117625117 on Build GRCh38
rs538951887 Chr.1: 117625252-117625252 on Build GRCh38
rs760894148 Chr.1: 117625269-117625269 on Build GRCh38
rs889082791 Chr.1: 117625399-117625399 on Build GRCh38
rs997633179 Chr.1: 117625286-117625286 on Build GRCh38

View Full Product Details