Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 118924303-118924810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX15
Entrez Gene ID
6913
Gene Name
T-box 15
Gene Aliases
TBX14
UniGene
Hs.146196
Gene Chromosome Location
Chr.1: 118883043-118989556 on Build GRCh38
Transcripts
NM_152380.2 XM_005271161.3 XM_005271162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6057821 c.271T>A p.Y91N Chr.1: 118924750-118924750 on Build GRCh38
6797882 c.245C>T p.A82V Chr.1: 118924776-118924776 on Build GRCh38
3472157 c.312G>A p.M104I Chr.1: 118924709-118924709 on Build GRCh38
6057825 c.305C>A p.T102N Chr.1: 118924716-118924716 on Build GRCh38
6057823 c.302A>T p.D101V Chr.1: 118924719-118924719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202071853 Chr.1: 118924734-118924734 on Build GRCh38
rs891555608 Chr.1: 118924445-118924445 on Build GRCh38
rs145168263 Chr.1: 118924336-118924337 on Build GRCh38
rs987821098 Chr.1: 118924331-118924331 on Build GRCh38
rs996010408 Chr.1: 118924668-118924668 on Build GRCh38
rs770855716 Chr.1: 118924601-118924601 on Build GRCh38
rs1779437 Chr.1: 118924504-118924504 on Build GRCh38
rs529903513 Chr.1: 118924426-118924426 on Build GRCh38
rs532509100 Chr.1: 118924638-118924638 on Build GRCh38
rs918645647 Chr.1: 118924591-118924591 on Build GRCh38
rs772300361 Chr.1: 118924502-118924502 on Build GRCh38
rs764990663 Chr.1: 118924603-118924604 on Build GRCh38
rs774248393 Chr.1: 118924763-118924763 on Build GRCh38
rs1047458849 Chr.1: 118924565-118924565 on Build GRCh38
rs768602481 Chr.1: 118924695-118924695 on Build GRCh38
rs12134163 Chr.1: 118924370-118924370 on Build GRCh38
rs748042008 Chr.1: 118924722-118924722 on Build GRCh38
rs367625608 Chr.1: 118924442-118924442 on Build GRCh38
rs374742990 Chr.1: 118924430-118924430 on Build GRCh38
rs781421196 Chr.1: 118924609-118924609 on Build GRCh38
rs558608963 Chr.1: 118924345-118924345 on Build GRCh38
rs776404850 Chr.1: 118924778-118924778 on Build GRCh38
rs147993592 Chr.1: 118924395-118924395 on Build GRCh38
rs189451226 Chr.1: 118924558-118924558 on Build GRCh38
rs951480480 Chr.1: 118924715-118924715 on Build GRCh38
rs749418037 Chr.1: 118924689-118924689 on Build GRCh38
rs757712578 Chr.1: 118924597-118924597 on Build GRCh38
rs34356524 Chr.1: 118924382-118924382 on Build GRCh38
rs756356225 Chr.1: 118924624-118924624 on Build GRCh38
rs759177438 Chr.1: 118924785-118924785 on Build GRCh38
rs1006802396 Chr.1: 118924518-118924518 on Build GRCh38
rs958057853 Chr.1: 118924729-118924729 on Build GRCh38
rs774411551 Chr.1: 118924708-118924708 on Build GRCh38
rs777627943 Chr.1: 118924476-118924476 on Build GRCh38
rs913971444 Chr.1: 118924737-118924737 on Build GRCh38
rs541353660 Chr.1: 118924464-118924464 on Build GRCh38
rs928221601 Chr.1: 118924480-118924480 on Build GRCh38
rs181047240 Chr.1: 118924596-118924596 on Build GRCh38
rs760507467 Chr.1: 118924744-118924744 on Build GRCh38
rs965330908 Chr.1: 118924575-118924575 on Build GRCh38
rs75248344 Chr.1: 118924500-118924500 on Build GRCh38
rs746177703 Chr.1: 118924610-118924610 on Build GRCh38
rs948318520 Chr.1: 118924348-118924348 on Build GRCh38
rs138987096 Chr.1: 118924766-118924766 on Build GRCh38
rs749219938 Chr.1: 118924365-118924365 on Build GRCh38
rs552136332 Chr.1: 118924578-118924578 on Build GRCh38
rs771989768 Chr.1: 118924732-118924732 on Build GRCh38
rs964607192 Chr.1: 118924630-118924630 on Build GRCh38
rs1028320196 Chr.1: 118924704-118924704 on Build GRCh38

View Full Product Details