Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119799569-119800061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REG4
Entrez Gene ID
83998
Gene Name
regenerating family member 4
Gene Aliases
GISP, REG-IV, RELP
UniGene
Hs.660883
Gene Chromosome Location
Chr.1: 119794018-119811580 on Build GRCh38
Transcripts
NM_001159352.1 NM_032044.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
675014 c.210C>A p.I70I Chr.1: 119799818-119799818 on Build GRCh38
675013 c.169G>T p.E57* Chr.1: 119799859-119799859 on Build GRCh38
5377406 c.250A>G p.S84G Chr.1: 119799778-119799778 on Build GRCh38
116559 c.199C>G p.L67V Chr.1: 119799829-119799829 on Build GRCh38
3472284 c.189C>T p.N63N Chr.1: 119799839-119799839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368208050 Chr.1: 119799675-119799675 on Build GRCh38
rs754764217 Chr.1: 119799812-119799812 on Build GRCh38
rs368379408 Chr.1: 119799624-119799624 on Build GRCh38
rs775997617 Chr.1: 119799770-119799770 on Build GRCh38
rs760496345 Chr.1: 119799846-119799848 on Build GRCh38
rs775201168 Chr.1: 119799759-119799760 on Build GRCh38
rs918281820 Chr.1: 119799626-119799626 on Build GRCh38
rs764843070 Chr.1: 119799706-119799706 on Build GRCh38
rs945710266 Chr.1: 119799786-119799786 on Build GRCh38
rs35669595 Chr.1: 119799999-119800000 on Build GRCh38
rs368857883 Chr.1: 119799736-119799736 on Build GRCh38
rs1025633971 Chr.1: 119799594-119799594 on Build GRCh38
rs763170486 Chr.1: 119799681-119799681 on Build GRCh38
rs1992377 Chr.1: 119799592-119799592 on Build GRCh38
rs779918758 Chr.1: 119799726-119799726 on Build GRCh38
rs775703821 Chr.1: 119799871-119799871 on Build GRCh38
rs587651480 Chr.1: 119800021-119800021 on Build GRCh38
rs587730010 Chr.1: 119800026-119800026 on Build GRCh38
rs373182467 Chr.1: 119799752-119799752 on Build GRCh38
rs587751115 Chr.1: 119799882-119799882 on Build GRCh38
rs771774182 Chr.1: 119799847-119799847 on Build GRCh38
rs749947655 Chr.1: 119799795-119799795 on Build GRCh38
rs112242267 Chr.1: 119799703-119799703 on Build GRCh38
rs202110610 Chr.1: 119799891-119799891 on Build GRCh38
rs587612717 Chr.1: 119799956-119799956 on Build GRCh38
rs190853679 Chr.1: 119799929-119799929 on Build GRCh38
rs1016996562 Chr.1: 119799965-119799965 on Build GRCh38
rs756035160 Chr.1: 119799839-119799839 on Build GRCh38
rs761353787 Chr.1: 119799688-119799688 on Build GRCh38
rs369912783 Chr.1: 119799821-119799821 on Build GRCh38
rs762594247 Chr.1: 119799797-119799797 on Build GRCh38
rs149425305 Chr.1: 119799996-119799996 on Build GRCh38
rs377287370 Chr.1: 119799810-119799810 on Build GRCh38
rs143784058 Chr.1: 119800003-119800003 on Build GRCh38
rs780857927 Chr.1: 119799818-119799818 on Build GRCh38
rs587773650 Chr.1: 119799755-119799755 on Build GRCh38
rs766674786 Chr.1: 119799686-119799686 on Build GRCh38
rs587771695 Chr.1: 119799605-119799605 on Build GRCh38
rs186245838 Chr.1: 119799918-119799918 on Build GRCh38
rs56123124 Chr.1: 119799654-119799654 on Build GRCh38
rs772350964 Chr.1: 119799767-119799767 on Build GRCh38
rs146476726 Chr.1: 119799845-119799845 on Build GRCh38
rs149733140 Chr.1: 119799859-119799859 on Build GRCh38
rs753162163 Chr.1: 119799701-119799701 on Build GRCh38
rs112419394 Chr.1: 119799827-119799827 on Build GRCh38
rs1001314148 Chr.1: 119799831-119799831 on Build GRCh38
rs951019836 Chr.1: 119799631-119799631 on Build GRCh38
rs769254624 Chr.1: 119799877-119799877 on Build GRCh38
rs887083848 Chr.1: 119799933-119799933 on Build GRCh38
rs371715163 Chr.1: 119799700-119799700 on Build GRCh38
rs766109983 Chr.1: 119799798-119799798 on Build GRCh38
rs111854244 Chr.1: 119799938-119799938 on Build GRCh38
rs587714351 Chr.1: 119799934-119799934 on Build GRCh38
rs777758243 Chr.1: 119799840-119799840 on Build GRCh38
rs987741571 Chr.1: 119799634-119799634 on Build GRCh38
rs963180830 Chr.1: 119799680-119799680 on Build GRCh38
rs587707085 Chr.1: 119799750-119799750 on Build GRCh38
rs587676154 Chr.1: 119800019-119800019 on Build GRCh38
rs972807044 Chr.1: 119799622-119799622 on Build GRCh38
rs984184495 Chr.1: 119799650-119799650 on Build GRCh38
rs774688143 Chr.1: 119799689-119799689 on Build GRCh38
rs759855943 Chr.1: 119799693-119799693 on Build GRCh38
rs761100121 Chr.1: 119799782-119799782 on Build GRCh38
rs772423011 Chr.1: 119799866-119799866 on Build GRCh38
rs200030201 Chr.1: 119799874-119799874 on Build GRCh38
rs769801763 Chr.1: 119799753-119799753 on Build GRCh38
rs587636387 Chr.1: 119799860-119799860 on Build GRCh38
rs772920823 Chr.1: 119799982-119799982 on Build GRCh38
rs587669560 Chr.1: 119799958-119799958 on Build GRCh38
rs951503252 Chr.1: 119799649-119799649 on Build GRCh38
rs766018303 Chr.1: 119799887-119799887 on Build GRCh38
rs764753106 Chr.1: 119799785-119799785 on Build GRCh38
rs759891812 Chr.1: 119799762-119799762 on Build GRCh38
rs370933200 Chr.1: 119799863-119799863 on Build GRCh38
rs1005981516 Chr.1: 119799961-119799961 on Build GRCh38
rs749344394 Chr.1: 119799844-119799844 on Build GRCh38
rs139174797 Chr.1: 119799838-119799838 on Build GRCh38
rs771107744 Chr.1: 119799756-119799756 on Build GRCh38
rs761186703 Chr.1: 119799875-119799875 on Build GRCh38
rs587694568 Chr.1: 119799595-119799595 on Build GRCh38
rs750037891 Chr.1: 119799717-119799717 on Build GRCh38
rs762505241 Chr.1: 119799884-119799884 on Build GRCh38
rs969444015 Chr.1: 119799977-119799977 on Build GRCh38
rs758108074 Chr.1: 119799721-119799721 on Build GRCh38
rs998447786 Chr.1: 119799928-119799928 on Build GRCh38
rs142055185 Chr.1: 119799737-119799737 on Build GRCh38
rs1045625285 Chr.1: 119799814-119799814 on Build GRCh38

View Full Product Details