Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7054364-7054863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TADA2B
Entrez Gene ID
93624
Gene Name
transcriptional adaptor 2B
Gene Aliases
ADA2(beta), ADA2B
UniGene
Hs.518614
Gene Chromosome Location
Chr.4: 7043429-7057952 on Build GRCh38
Transcripts
NM_152293.2 XM_011513595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4125708 c.655C>T p.R219W Chr.4: 7054446-7054446 on Build GRCh38
1057005 c.763C>T p.L255L Chr.4: 7054554-7054554 on Build GRCh38
1057004 c.667G>A p.A223T Chr.4: 7054458-7054458 on Build GRCh38
4125710 c.1072C>A p.L358I Chr.4: 7054863-7054863 on Build GRCh38
4125709 c.767G>A p.R256H Chr.4: 7054558-7054558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752071626 Chr.4: 7054522-7054522 on Build GRCh38
rs367868603 Chr.4: 7054639-7054639 on Build GRCh38
rs768057126 Chr.4: 7054625-7054625 on Build GRCh38
rs772692077 Chr.4: 7054673-7054673 on Build GRCh38
rs372684985 Chr.4: 7054790-7054790 on Build GRCh38
rs773540665 Chr.4: 7054742-7054744 on Build GRCh38
rs779568198 Chr.4: 7054539-7054539 on Build GRCh38
rs773346952 Chr.4: 7054454-7054454 on Build GRCh38
rs746850955 Chr.4: 7054669-7054669 on Build GRCh38
rs750943990 Chr.4: 7054736-7054736 on Build GRCh38
rs199685832 Chr.4: 7054751-7054751 on Build GRCh38
rs903727001 Chr.4: 7054597-7054597 on Build GRCh38
rs1004859156 Chr.4: 7054637-7054637 on Build GRCh38
rs368691486 Chr.4: 7054574-7054574 on Build GRCh38
rs768705454 Chr.4: 7054811-7054811 on Build GRCh38
rs763030316 Chr.4: 7054589-7054589 on Build GRCh38
rs768313668 Chr.4: 7054440-7054440 on Build GRCh38
rs761955767 Chr.4: 7054815-7054815 on Build GRCh38
rs768029764 Chr.4: 7054549-7054549 on Build GRCh38
rs775464029 Chr.4: 7054802-7054802 on Build GRCh38
rs75397160 Chr.4: 7054469-7054469 on Build GRCh38
rs374613250 Chr.4: 7054493-7054493 on Build GRCh38
rs557910360 Chr.4: 7054753-7054753 on Build GRCh38
rs200382873 Chr.4: 7054519-7054519 on Build GRCh38
rs777627029 Chr.4: 7054766-7054766 on Build GRCh38
rs1012465138 Chr.4: 7054540-7054540 on Build GRCh38
rs771747907 Chr.4: 7054449-7054449 on Build GRCh38
rs753932914 Chr.4: 7054466-7054466 on Build GRCh38
rs753150104 Chr.4: 7054401-7054401 on Build GRCh38
rs779180780 Chr.4: 7054399-7054399 on Build GRCh38
rs550257653 Chr.4: 7054459-7054459 on Build GRCh38
rs370055655 Chr.4: 7054609-7054609 on Build GRCh38
rs754128322 Chr.4: 7054388-7054388 on Build GRCh38
rs764337259 Chr.4: 7054723-7054723 on Build GRCh38
rs1041449605 Chr.4: 7054407-7054407 on Build GRCh38
rs963844224 Chr.4: 7054795-7054795 on Build GRCh38
rs762223816 Chr.4: 7054726-7054726 on Build GRCh38
rs763401693 Chr.4: 7054443-7054443 on Build GRCh38
rs780794883 Chr.4: 7054775-7054775 on Build GRCh38
rs1055804392 Chr.4: 7054446-7054446 on Build GRCh38
rs567853124 Chr.4: 7054403-7054403 on Build GRCh38
rs755182242 Chr.4: 7054391-7054391 on Build GRCh38
rs775380828 Chr.4: 7054709-7054709 on Build GRCh38
rs747512193 Chr.4: 7054418-7054418 on Build GRCh38
rs766846018 Chr.4: 7054839-7054839 on Build GRCh38
rs766858303 Chr.4: 7054590-7054590 on Build GRCh38
rs773844145 Chr.4: 7054442-7054442 on Build GRCh38
rs760726462 Chr.4: 7054572-7054572 on Build GRCh38
rs757583905 Chr.4: 7054620-7054620 on Build GRCh38
rs760620156 Chr.4: 7054457-7054457 on Build GRCh38
rs778684378 Chr.4: 7054551-7054551 on Build GRCh38
rs369931391 Chr.4: 7054814-7054814 on Build GRCh38
rs779480252 Chr.4: 7054432-7054432 on Build GRCh38
rs373694530 Chr.4: 7054638-7054638 on Build GRCh38
rs754582445 Chr.4: 7054634-7054634 on Build GRCh38
rs866492528 Chr.4: 7054623-7054623 on Build GRCh38
rs752972123 Chr.4: 7054583-7054583 on Build GRCh38
rs757709054 Chr.4: 7054536-7054536 on Build GRCh38
rs377183651 Chr.4: 7054808-7054808 on Build GRCh38
rs765549514 Chr.4: 7054483-7054483 on Build GRCh38
rs767937296 Chr.4: 7054816-7054816 on Build GRCh38
rs371816393 Chr.4: 7054693-7054693 on Build GRCh38
rs767862945 Chr.4: 7054734-7054734 on Build GRCh38
rs906964326 Chr.4: 7054505-7054505 on Build GRCh38
rs773398504 Chr.4: 7054827-7054827 on Build GRCh38
rs777537320 Chr.4: 7054656-7054656 on Build GRCh38
rs971508352 Chr.4: 7054614-7054614 on Build GRCh38
rs748914763 Chr.4: 7054436-7054436 on Build GRCh38
rs538574410 Chr.4: 7054430-7054430 on Build GRCh38
rs757960745 Chr.4: 7054761-7054761 on Build GRCh38
rs774530772 Chr.4: 7054724-7054724 on Build GRCh38
rs376472841 Chr.4: 7054767-7054767 on Build GRCh38
rs370972038 Chr.4: 7054462-7054462 on Build GRCh38
rs539508764 Chr.4: 7054682-7054682 on Build GRCh38
rs748866730 Chr.4: 7054545-7054545 on Build GRCh38
rs1022146320 Chr.4: 7054585-7054585 on Build GRCh38
rs771996154 Chr.4: 7054558-7054558 on Build GRCh38
rs761071358 Chr.4: 7054831-7054831 on Build GRCh38
rs758866010 Chr.4: 7054516-7054516 on Build GRCh38
rs765232007 Chr.4: 7054580-7054580 on Build GRCh38
rs758668459 Chr.4: 7054402-7054402 on Build GRCh38
rs752190983 Chr.4: 7054754-7054754 on Build GRCh38
rs771033560 Chr.4: 7054573-7054573 on Build GRCh38
rs773094997 Chr.4: 7054562-7054562 on Build GRCh38
rs764462211 Chr.4: 7054607-7054607 on Build GRCh38
rs758085565 Chr.4: 7054644-7054644 on Build GRCh38
rs914496603 Chr.4: 7054836-7054836 on Build GRCh38
rs368347599 Chr.4: 7054776-7054776 on Build GRCh38
rs745728932 Chr.4: 7054685-7054685 on Build GRCh38
rs747749615 Chr.4: 7054553-7054553 on Build GRCh38
rs750881291 Chr.4: 7054626-7054626 on Build GRCh38

View Full Product Details