Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8389026-8389518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOX3
Entrez Gene ID
8310
Gene Name
acyl-CoA oxidase 3, pristanoyl
Gene Aliases
-
UniGene
Hs.479122
Gene Chromosome Location
Chr.4: 8366263-8440725 on Build GRCh38
Transcripts
NM_001101667.1 NM_003501.2 XM_005248011.4 XM_005248013.3 XM_011513565.2 XM_011513566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1058232 c.1481G>A p.G494D Chr.4: 8389229-8389229 on Build GRCh38
5898230 c.1537+7A>G p.? Chr.4: 8389166-8389166 on Build GRCh38
308824 c.1520A>G p.D507G Chr.4: 8389190-8389190 on Build GRCh38
4467216 c.1477C>T p.P493S Chr.4: 8389233-8389233 on Build GRCh38
6101295 c.1468G>T p.D490Y Chr.4: 8389242-8389242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146891905 Chr.4: 8389176-8389176 on Build GRCh38
rs531417132 Chr.4: 8389051-8389051 on Build GRCh38
rs115146093 Chr.4: 8389264-8389264 on Build GRCh38
rs1048052837 Chr.4: 8389086-8389086 on Build GRCh38
rs768089067 Chr.4: 8389222-8389222 on Build GRCh38
rs892707239 Chr.4: 8389389-8389389 on Build GRCh38
rs1045263721 Chr.4: 8389343-8389343 on Build GRCh38
rs895552345 Chr.4: 8389099-8389099 on Build GRCh38
rs975261453 Chr.4: 8389048-8389048 on Build GRCh38
rs113402127 Chr.4: 8389231-8389231 on Build GRCh38
rs943894463 Chr.4: 8389044-8389044 on Build GRCh38
rs143888158 Chr.4: 8389131-8389131 on Build GRCh38
rs933754649 Chr.4: 8389059-8389059 on Build GRCh38
rs750858650 Chr.4: 8389145-8389145 on Build GRCh38
rs142001570 Chr.4: 8389238-8389238 on Build GRCh38
rs139709179 Chr.4: 8389098-8389098 on Build GRCh38
rs777424017 Chr.4: 8389313-8389313 on Build GRCh38
rs368490039 Chr.4: 8389461-8389461 on Build GRCh38
rs748737411 Chr.4: 8389177-8389178 on Build GRCh38
rs1022592513 Chr.4: 8389105-8389105 on Build GRCh38
rs548170046 Chr.4: 8389191-8389191 on Build GRCh38
rs370874561 Chr.4: 8389202-8389202 on Build GRCh38
rs755036511 Chr.4: 8389154-8389154 on Build GRCh38
rs113317365 Chr.4: 8389153-8389153 on Build GRCh38
rs13136987 Chr.4: 8389132-8389132 on Build GRCh38
rs866122970 Chr.4: 8389360-8389360 on Build GRCh38
rs778343652 Chr.4: 8389185-8389185 on Build GRCh38
rs902758298 Chr.4: 8389106-8389106 on Build GRCh38
rs756736494 Chr.4: 8389315-8389315 on Build GRCh38
rs985134972 Chr.4: 8389438-8389438 on Build GRCh38
rs765322454 Chr.4: 8389163-8389163 on Build GRCh38
rs778401352 Chr.4: 8389336-8389336 on Build GRCh38
rs955254924 Chr.4: 8389425-8389425 on Build GRCh38
rs572242062 Chr.4: 8389316-8389316 on Build GRCh38
rs773885685 Chr.4: 8389227-8389227 on Build GRCh38
rs905444308 Chr.4: 8389165-8389165 on Build GRCh38
rs996906980 Chr.4: 8389420-8389420 on Build GRCh38
rs532690394 Chr.4: 8389473-8389473 on Build GRCh38
rs202154683 Chr.4: 8389240-8389240 on Build GRCh38
rs767345331 Chr.4: 8389129-8389129 on Build GRCh38
rs999743548 Chr.4: 8389110-8389110 on Build GRCh38
rs752904139 Chr.4: 8389177-8389177 on Build GRCh38
rs1056776904 Chr.4: 8389484-8389484 on Build GRCh38
rs554082211 Chr.4: 8389312-8389312 on Build GRCh38
rs935470341 Chr.4: 8389477-8389477 on Build GRCh38
rs759487661 Chr.4: 8389242-8389242 on Build GRCh38
rs758671602 Chr.4: 8389183-8389183 on Build GRCh38
rs16842165 Chr.4: 8389392-8389392 on Build GRCh38
rs753749133 Chr.4: 8389148-8389148 on Build GRCh38
rs1028130468 Chr.4: 8389455-8389455 on Build GRCh38
rs543507124 Chr.4: 8389393-8389393 on Build GRCh38
rs555256838 Chr.4: 8389410-8389410 on Build GRCh38
rs145751243 Chr.4: 8389245-8389245 on Build GRCh38
rs761655620 Chr.4: 8389411-8389411 on Build GRCh38
rs202006899 Chr.4: 8389336-8389336 on Build GRCh38
rs571839254 Chr.4: 8389075-8389075 on Build GRCh38
rs144357499 Chr.4: 8389474-8389474 on Build GRCh38
rs1023703877 Chr.4: 8389192-8389192 on Build GRCh38
rs747851224 Chr.4: 8389229-8389229 on Build GRCh38
rs770674680 Chr.4: 8389321-8389321 on Build GRCh38
rs530752406 Chr.4: 8389071-8389071 on Build GRCh38
rs939805064 Chr.4: 8389092-8389092 on Build GRCh38
rs1054065133 Chr.4: 8389376-8389376 on Build GRCh38
rs905329895 Chr.4: 8389355-8389355 on Build GRCh38
rs747661619 Chr.4: 8389308-8389308 on Build GRCh38
rs550424228 Chr.4: 8389070-8389070 on Build GRCh38
rs138564245 Chr.4: 8389252-8389252 on Build GRCh38
rs771708462 Chr.4: 8389230-8389230 on Build GRCh38
rs766147750 Chr.4: 8389239-8389239 on Build GRCh38
rs999795396 Chr.4: 8389311-8389311 on Build GRCh38
rs36055234 Chr.4: 8389438-8389439 on Build GRCh38
rs760724003 Chr.4: 8389335-8389335 on Build GRCh38
rs1049463170 Chr.4: 8389066-8389066 on Build GRCh38
rs778350485 Chr.4: 8389276-8389276 on Build GRCh38
rs961166143 Chr.4: 8389402-8389402 on Build GRCh38
rs201425518 Chr.4: 8389265-8389265 on Build GRCh38
rs114126152 Chr.4: 8389271-8389271 on Build GRCh38
rs138860196 Chr.4: 8389272-8389272 on Build GRCh38
rs886746353 Chr.4: 8389087-8389087 on Build GRCh38
rs781566252 Chr.4: 8389215-8389215 on Build GRCh38
rs766306525 Chr.4: 8389142-8389142 on Build GRCh38
rs956670655 Chr.4: 8389194-8389194 on Build GRCh38
rs1012249601 Chr.4: 8389349-8389349 on Build GRCh38
rs2242483 Chr.4: 8389322-8389322 on Build GRCh38
rs13434465 Chr.4: 8389221-8389221 on Build GRCh38
rs900808266 Chr.4: 8389052-8389052 on Build GRCh38

View Full Product Details