Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8582439-8582936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR78
Entrez Gene ID
27201
Gene Name
G protein-coupled receptor 78
Gene Aliases
-
UniGene
Hs.740355
Gene Chromosome Location
Chr.4: 8580237-8590023 on Build GRCh38
Transcripts
NM_080819.4 NR_045511.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
351083 c.731G>T p.G244V Chr.4: 8582593-8582593 on Build GRCh38
7287400 c.686T>G p.L229R Chr.4: 8582548-8582548 on Build GRCh38
4957793 c.781A>C p.R261R Chr.4: 8582643-8582643 on Build GRCh38
734411 c.715G>C p.A239P Chr.4: 8582577-8582577 on Build GRCh38
3606717 c.693G>A p.Q231Q Chr.4: 8582555-8582555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896739457 Chr.4: 8582770-8582770 on Build GRCh38
rs199935344 Chr.4: 8582566-8582566 on Build GRCh38
rs945366931 Chr.4: 8582794-8582794 on Build GRCh38
rs140868152 Chr.4: 8582606-8582606 on Build GRCh38
rs1001459938 Chr.4: 8582810-8582810 on Build GRCh38
rs988143588 Chr.4: 8582718-8582718 on Build GRCh38
rs760202454 Chr.4: 8582488-8582488 on Build GRCh38
rs753669582 Chr.4: 8582629-8582629 on Build GRCh38
rs35460303 Chr.4: 8582576-8582576 on Build GRCh38
rs776195249 Chr.4: 8582541-8582541 on Build GRCh38
rs370573791 Chr.4: 8582514-8582514 on Build GRCh38
rs767824059 Chr.4: 8582613-8582613 on Build GRCh38
rs956669045 Chr.4: 8582713-8582713 on Build GRCh38
rs778237308 Chr.4: 8582535-8582535 on Build GRCh38
rs879705557 Chr.4: 8582486-8582486 on Build GRCh38
rs1019667868 Chr.4: 8582848-8582848 on Build GRCh38
rs753730950 Chr.4: 8582494-8582494 on Build GRCh38
rs9992205 Chr.4: 8582745-8582745 on Build GRCh38
rs540679842 Chr.4: 8582760-8582760 on Build GRCh38
rs202089891 Chr.4: 8582575-8582575 on Build GRCh38
rs774867098 Chr.4: 8582641-8582641 on Build GRCh38
rs759012493 Chr.4: 8582601-8582601 on Build GRCh38
rs931119830 Chr.4: 8582882-8582882 on Build GRCh38
rs552546058 Chr.4: 8582661-8582661 on Build GRCh38
rs373204707 Chr.4: 8582649-8582649 on Build GRCh38
rs749746307 Chr.4: 8582621-8582621 on Build GRCh38
rs780067541 Chr.4: 8582644-8582644 on Build GRCh38
rs984365884 Chr.4: 8582891-8582891 on Build GRCh38
rs767177899 Chr.4: 8582462-8582462 on Build GRCh38
rs775445723 Chr.4: 8582604-8582604 on Build GRCh38
rs531270618 Chr.4: 8582496-8582496 on Build GRCh38
rs757556219 Chr.4: 8582573-8582573 on Build GRCh38
rs747482675 Chr.4: 8582800-8582800 on Build GRCh38
rs777448014 Chr.4: 8582570-8582570 on Build GRCh38
rs6857501 Chr.4: 8582766-8582766 on Build GRCh38
rs564777172 Chr.4: 8582751-8582751 on Build GRCh38
rs939652949 Chr.4: 8582753-8582753 on Build GRCh38
rs755529039 Chr.4: 8582526-8582527 on Build GRCh38
rs1041610354 Chr.4: 8582620-8582620 on Build GRCh38
rs749349279 Chr.4: 8582685-8582685 on Build GRCh38
rs781227331 Chr.4: 8582543-8582544 on Build GRCh38
rs765821153 Chr.4: 8582562-8582562 on Build GRCh38
rs562973164 Chr.4: 8582788-8582788 on Build GRCh38
rs868472426 Chr.4: 8582666-8582666 on Build GRCh38
rs771227274 Chr.4: 8582619-8582619 on Build GRCh38
rs140825114 Chr.4: 8582557-8582557 on Build GRCh38
rs113233011 Chr.4: 8582725-8582725 on Build GRCh38
rs368181816 Chr.4: 8582543-8582543 on Build GRCh38
rs774393155 Chr.4: 8582653-8582653 on Build GRCh38
rs775007611 Chr.4: 8582851-8582851 on Build GRCh38
rs141741405 Chr.4: 8582646-8582646 on Build GRCh38
rs578141698 Chr.4: 8582493-8582493 on Build GRCh38
rs752813458 Chr.4: 8582564-8582574 on Build GRCh38
rs374263163 Chr.4: 8582524-8582524 on Build GRCh38
rs938575271 Chr.4: 8582841-8582841 on Build GRCh38
rs749861236 Chr.4: 8582574-8582574 on Build GRCh38
rs151017686 Chr.4: 8582599-8582599 on Build GRCh38
rs6824455 Chr.4: 8582721-8582721 on Build GRCh38
rs781276686 Chr.4: 8582793-8582793 on Build GRCh38
rs147129513 Chr.4: 8582815-8582815 on Build GRCh38
rs372697554 Chr.4: 8582523-8582523 on Build GRCh38
rs369729333 Chr.4: 8582875-8582875 on Build GRCh38
rs534327250 Chr.4: 8582634-8582634 on Build GRCh38
rs755147480 Chr.4: 8582669-8582669 on Build GRCh38
rs556983708 Chr.4: 8582731-8582731 on Build GRCh38
rs756418159 Chr.4: 8582663-8582663 on Build GRCh38
rs199619277 Chr.4: 8582608-8582608 on Build GRCh38
rs1003850565 Chr.4: 8582618-8582618 on Build GRCh38
rs943972554 Chr.4: 8582723-8582723 on Build GRCh38
rs770418487 Chr.4: 8582536-8582536 on Build GRCh38
rs116438375 Chr.4: 8582779-8582779 on Build GRCh38
rs750624516 Chr.4: 8582662-8582662 on Build GRCh38
rs201754764 Chr.4: 8582593-8582593 on Build GRCh38
rs143649237 Chr.4: 8582579-8582579 on Build GRCh38
rs925373998 Chr.4: 8582711-8582711 on Build GRCh38
rs147709824 Chr.4: 8582542-8582542 on Build GRCh38
rs1012661253 Chr.4: 8582720-8582720 on Build GRCh38
rs144674484 Chr.4: 8582565-8582565 on Build GRCh38
rs761671141 Chr.4: 8582660-8582660 on Build GRCh38
rs755166281 Chr.4: 8582694-8582694 on Build GRCh38
rs147903053 Chr.4: 8582568-8582568 on Build GRCh38
rs780580777 Chr.4: 8582577-8582577 on Build GRCh38
rs573566609 Chr.4: 8582754-8582754 on Build GRCh38
rs868285652 Chr.4: 8582778-8582778 on Build GRCh38
rs762621976 Chr.4: 8582487-8582487 on Build GRCh38
rs746600928 Chr.4: 8582598-8582598 on Build GRCh38
rs753862812 Chr.4: 8582667-8582667 on Build GRCh38
rs376391641 Chr.4: 8582664-8582664 on Build GRCh38
rs570455602 Chr.4: 8582846-8582846 on Build GRCh38
rs542455475 Chr.4: 8582469-8582469 on Build GRCh38
rs558447540 Chr.4: 8582628-8582628 on Build GRCh38
rs371542714 Chr.4: 8582569-8582569 on Build GRCh38
rs138639661 Chr.4: 8582630-8582630 on Build GRCh38
rs375928457 Chr.4: 8582504-8582504 on Build GRCh38
rs767187077 Chr.4: 8582484-8582484 on Build GRCh38
rs189070828 Chr.4: 8582605-8582605 on Build GRCh38
rs547044437 Chr.4: 8582905-8582905 on Build GRCh38
rs200777923 Chr.4: 8582549-8582549 on Build GRCh38
rs144794260 Chr.4: 8582625-8582625 on Build GRCh38
rs763923576 Chr.4: 8582648-8582648 on Build GRCh38
rs190840752 Chr.4: 8582838-8582838 on Build GRCh38
rs921197146 Chr.4: 8582866-8582866 on Build GRCh38
rs748683356 Chr.4: 8582696-8582696 on Build GRCh38
rs6857510 Chr.4: 8582784-8582784 on Build GRCh38
rs779268746 Chr.4: 8582682-8582682 on Build GRCh38
rs975386844 Chr.4: 8582862-8582862 on Build GRCh38
rs764754180 Chr.4: 8582603-8582603 on Build GRCh38
rs563708354 Chr.4: 8582479-8582479 on Build GRCh38
rs756223947 Chr.4: 8582617-8582617 on Build GRCh38
rs771080876 Chr.4: 8582531-8582531 on Build GRCh38
rs376662328 Chr.4: 8582572-8582572 on Build GRCh38
rs186447375 Chr.4: 8582744-8582744 on Build GRCh38
rs750716357 Chr.4: 8582615-8582615 on Build GRCh38

View Full Product Details