Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17633608-17634057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM184B
Entrez Gene ID
27146
Gene Name
family with sequence similarity 184 member B
Gene Aliases
-
UniGene
Hs.744951
Gene Chromosome Location
Chr.4: 17632073-17781512 on Build GRCh38
Transcripts
NM_015688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6267785 c.3060C>A p.N1020K Chr.4: 17633718-17633718 on Build GRCh38
6409898 c.2927G>T p.S976I Chr.4: 17633851-17633851 on Build GRCh38
1733592 c.2978G>A p.S993N Chr.4: 17633800-17633800 on Build GRCh38
3993555 c.2915G>T p.S972I Chr.4: 17633863-17633863 on Build GRCh38
6275135 c.3013C>A p.P1005T Chr.4: 17633765-17633765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553782107 Chr.4: 17633963-17633963 on Build GRCh38
rs139341568 Chr.4: 17633803-17633803 on Build GRCh38
rs1016629489 Chr.4: 17633652-17633652 on Build GRCh38
rs765417435 Chr.4: 17633867-17633867 on Build GRCh38
rs569401398 Chr.4: 17633849-17633849 on Build GRCh38
rs1037962319 Chr.4: 17633878-17633878 on Build GRCh38
rs1029910127 Chr.4: 17633827-17633827 on Build GRCh38
rs6449318 Chr.4: 17634028-17634028 on Build GRCh38
rs368942670 Chr.4: 17633860-17633860 on Build GRCh38
rs975520151 Chr.4: 17633814-17633814 on Build GRCh38
rs752679659 Chr.4: 17633675-17633675 on Build GRCh38
rs188210452 Chr.4: 17633861-17633861 on Build GRCh38
rs202198134 Chr.4: 17633777-17633777 on Build GRCh38
rs190016398 Chr.4: 17633676-17633676 on Build GRCh38
rs892301224 Chr.4: 17634026-17634026 on Build GRCh38
rs764268487 Chr.4: 17633931-17633931 on Build GRCh38
rs562811677 Chr.4: 17634001-17634001 on Build GRCh38
rs534006353 Chr.4: 17633939-17633939 on Build GRCh38
rs565032163 Chr.4: 17633646-17633646 on Build GRCh38
rs201997472 Chr.4: 17633937-17633937 on Build GRCh38
rs972445765 Chr.4: 17633886-17633886 on Build GRCh38
rs772467729 Chr.4: 17634033-17634033 on Build GRCh38
rs374123291 Chr.4: 17633988-17633988 on Build GRCh38
rs183471400 Chr.4: 17633858-17633858 on Build GRCh38
rs140729140 Chr.4: 17633934-17633938 on Build GRCh38
rs766612325 Chr.4: 17633808-17633808 on Build GRCh38
rs981799386 Chr.4: 17633940-17633940 on Build GRCh38
rs370827552 Chr.4: 17633850-17633850 on Build GRCh38
rs374104756 Chr.4: 17633793-17633793 on Build GRCh38
rs552757786 Chr.4: 17634019-17634020 on Build GRCh38
rs763185151 Chr.4: 17633899-17633899 on Build GRCh38
rs927159216 Chr.4: 17633690-17633690 on Build GRCh38
rs950039229 Chr.4: 17633678-17633678 on Build GRCh38
rs958833482 Chr.4: 17633988-17633990 on Build GRCh38
rs769141159 Chr.4: 17633881-17633881 on Build GRCh38
rs547445434 Chr.4: 17633699-17633699 on Build GRCh38
rs778474478 Chr.4: 17633702-17633702 on Build GRCh38
rs1010785297 Chr.4: 17633785-17633785 on Build GRCh38
rs893898305 Chr.4: 17633732-17633732 on Build GRCh38
rs948315383 Chr.4: 17633981-17633981 on Build GRCh38
rs61741415 Chr.4: 17633765-17633765 on Build GRCh38
rs368263511 Chr.4: 17633807-17633807 on Build GRCh38
rs191729460 Chr.4: 17633851-17633851 on Build GRCh38
rs752100556 Chr.4: 17633704-17633704 on Build GRCh38
rs1025289958 Chr.4: 17633921-17633921 on Build GRCh38
rs61741403 Chr.4: 17633713-17633713 on Build GRCh38
rs74627932 Chr.4: 17633938-17633938 on Build GRCh38
rs760600397 Chr.4: 17633822-17633822 on Build GRCh38
rs752946960 Chr.4: 17633871-17633871 on Build GRCh38
rs939451197 Chr.4: 17633995-17633995 on Build GRCh38
rs946759250 Chr.4: 17633736-17633736 on Build GRCh38
rs1003562624 Chr.4: 17633758-17633758 on Build GRCh38
rs530767348 Chr.4: 17633740-17633740 on Build GRCh38
rs1017017383 Chr.4: 17633859-17633859 on Build GRCh38
rs16343 Chr.4: 17633937-17633941 on Build GRCh38
rs576248417 Chr.4: 17634014-17634014 on Build GRCh38
rs914378209 Chr.4: 17633990-17633992 on Build GRCh38
rs16895358 Chr.4: 17633987-17633987 on Build GRCh38
rs765315987 Chr.4: 17633933-17633934 on Build GRCh38
rs981365069 Chr.4: 17633681-17633681 on Build GRCh38
rs757766404 Chr.4: 17633712-17633712 on Build GRCh38
rs1052436372 Chr.4: 17634022-17634022 on Build GRCh38
rs906637177 Chr.4: 17633737-17633737 on Build GRCh38
rs770765197 Chr.4: 17633844-17633844 on Build GRCh38
rs759610545 Chr.4: 17633665-17633665 on Build GRCh38
rs374041125 Chr.4: 17633730-17633730 on Build GRCh38
rs765371700 Chr.4: 17633673-17633673 on Build GRCh38
rs954219469 Chr.4: 17633642-17633642 on Build GRCh38
rs796818449 Chr.4: 17633973-17633973 on Build GRCh38

View Full Product Details