Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 20727285-20727782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PACRGL
Entrez Gene ID
133015
Gene Name
PARK2 coregulated like
Gene Aliases
C4orf28
UniGene
Hs.479298
Gene Chromosome Location
Chr.4: 20696282-20757766 on Build GRCh38
Transcripts
NM_001130727.1 NM_001258345.1 NM_001258346.1 NM_001317849.1 NM_145048.3 NR_047661.1 NR_133933.1 XM_011513780.1 XM_011513781.1 XM_011513784.1 XM_011513785.1 XM_011513786.2 XM_011513787.1 XM_011513788.1 XM_011513791.1 XM_011513792.1 XM_011513794.1 XM_011513795.1 XM_011513796.1 XM_011513797.2 XM_011513798.2 XM_011513804.1 XM_011513805.2 XM_017007747.1 XM_017007748.1 XM_017007749.1 XM_017007750.1 XM_017007751.1 XM_017007752.1 XM_017007753.1 XM_017007754.1 XM_017007755.1 XM_017007756.1 XM_017007757.1 XM_017007758.1 XM_017007759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6167317 c.706A>T p.I236F Chr.4: 20727300-20727300 on Build GRCh38
1663308 c.626T>A p.I209N Chr.4: 20727301-20727301 on Build GRCh38
6167318 c.625A>T p.I209F Chr.4: 20727300-20727300 on Build GRCh38
3602902 c.734C>T p.S245F Chr.4: 20727328-20727328 on Build GRCh38
3602905 c.654C>T p.S218S Chr.4: 20727329-20727329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906732971 Chr.4: 20727651-20727651 on Build GRCh38
rs989438410 Chr.4: 20727328-20727328 on Build GRCh38
rs1026447794 Chr.4: 20727424-20727424 on Build GRCh38
rs1004303843 Chr.4: 20727392-20727392 on Build GRCh38
rs997088343 Chr.4: 20727440-20727440 on Build GRCh38
rs757463354 Chr.4: 20727363-20727363 on Build GRCh38
rs529968084 Chr.4: 20727558-20727558 on Build GRCh38
rs977971375 Chr.4: 20727545-20727545 on Build GRCh38
rs933790333 Chr.4: 20727556-20727556 on Build GRCh38
rs369197815 Chr.4: 20727381-20727381 on Build GRCh38
rs142306794 Chr.4: 20727653-20727655 on Build GRCh38
rs966339522 Chr.4: 20727700-20727700 on Build GRCh38
rs200328704 Chr.4: 20727653-20727653 on Build GRCh38
rs906680751 Chr.4: 20727439-20727439 on Build GRCh38
rs769031129 Chr.4: 20727382-20727382 on Build GRCh38
rs989540165 Chr.4: 20727481-20727481 on Build GRCh38
rs922444599 Chr.4: 20727549-20727549 on Build GRCh38
rs13145309 Chr.4: 20727571-20727571 on Build GRCh38
rs956101959 Chr.4: 20727488-20727488 on Build GRCh38
rs752450725 Chr.4: 20727347-20727347 on Build GRCh38
rs778504014 Chr.4: 20727363-20727364 on Build GRCh38
rs113484601 Chr.4: 20727681-20727681 on Build GRCh38
rs546316258 Chr.4: 20727464-20727464 on Build GRCh38
rs781507318 Chr.4: 20727441-20727441 on Build GRCh38
rs901325716 Chr.4: 20727671-20727671 on Build GRCh38
rs765159207 Chr.4: 20727346-20727346 on Build GRCh38
rs966622492 Chr.4: 20727543-20727544 on Build GRCh38
rs772785267 Chr.4: 20727386-20727386 on Build GRCh38
rs565452972 Chr.4: 20727372-20727372 on Build GRCh38
rs182839459 Chr.4: 20727667-20727667 on Build GRCh38
rs929573136 Chr.4: 20727623-20727623 on Build GRCh38
rs1037987053 Chr.4: 20727401-20727401 on Build GRCh38
rs114693292 Chr.4: 20727607-20727607 on Build GRCh38
rs145894825 Chr.4: 20727480-20727480 on Build GRCh38
rs765849337 Chr.4: 20727333-20727333 on Build GRCh38
rs760216144 Chr.4: 20727321-20727321 on Build GRCh38
rs568100194 Chr.4: 20727513-20727513 on Build GRCh38
rs747224102 Chr.4: 20727362-20727362 on Build GRCh38
rs943347146 Chr.4: 20727393-20727393 on Build GRCh38
rs527876816 Chr.4: 20727423-20727423 on Build GRCh38
rs760338949 Chr.4: 20727388-20727388 on Build GRCh38
rs901460154 Chr.4: 20727421-20727421 on Build GRCh38
rs17552929 Chr.4: 20727343-20727343 on Build GRCh38
rs549734644 Chr.4: 20727512-20727512 on Build GRCh38
rs944456126 Chr.4: 20727648-20727648 on Build GRCh38
rs548238136 Chr.4: 20727489-20727491 on Build GRCh38
rs201585500 Chr.4: 20727655-20727655 on Build GRCh38
rs1041187034 Chr.4: 20727409-20727409 on Build GRCh38
rs59189281 Chr.4: 20727657-20727659 on Build GRCh38
rs143461358 Chr.4: 20727683-20727683 on Build GRCh38
rs776050680 Chr.4: 20727373-20727373 on Build GRCh38
rs534870880 Chr.4: 20727402-20727402 on Build GRCh38
rs753291944 Chr.4: 20727334-20727334 on Build GRCh38
rs1033826606 Chr.4: 20727477-20727480 on Build GRCh38
rs753131745 Chr.4: 20727540-20727540 on Build GRCh38
rs13145492 Chr.4: 20727628-20727628 on Build GRCh38
rs767131973 Chr.4: 20727315-20727315 on Build GRCh38
rs149241470 Chr.4: 20727633-20727633 on Build GRCh38
rs548079103 Chr.4: 20727569-20727569 on Build GRCh38
rs930271830 Chr.4: 20727589-20727589 on Build GRCh38
rs531733117 Chr.4: 20727496-20727496 on Build GRCh38
rs920272677 Chr.4: 20727588-20727588 on Build GRCh38
rs1000286376 Chr.4: 20727468-20727468 on Build GRCh38
rs566372151 Chr.4: 20727570-20727570 on Build GRCh38
rs1028768164 Chr.4: 20727455-20727455 on Build GRCh38
rs980014125 Chr.4: 20727580-20727580 on Build GRCh38
rs915131589 Chr.4: 20727329-20727329 on Build GRCh38
rs745876938 Chr.4: 20727367-20727367 on Build GRCh38
rs758200971 Chr.4: 20727355-20727355 on Build GRCh38
rs772911842 Chr.4: 20727316-20727316 on Build GRCh38

View Full Product Details