Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25351510-25352022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCCHC4
Entrez Gene ID
29063
Gene Name
zinc finger CCHC-type containing 4
Gene Aliases
HSPC052, ZGRF4
UniGene
Hs.278945
Gene Chromosome Location
Chr.4: 25312774-25373176 on Build GRCh38
Transcripts
NM_001318148.1 NM_024936.2 XM_011513835.1 XM_017008128.1 XM_017008129.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4891746 c.919C>T p.H307Y Chr.4: 25351597-25351597 on Build GRCh38
6825966 c.968G>A p.R323Q Chr.4: 25351646-25351646 on Build GRCh38
5770385 c.1011+4G>C p.? Chr.4: 25351693-25351693 on Build GRCh38
3133260 c.911-1G>T p.? Chr.4: 25351588-25351588 on Build GRCh38
5866228 c.942G>A p.W314* Chr.4: 25351620-25351620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748621706 Chr.4: 25351595-25351595 on Build GRCh38
rs894827982 Chr.4: 25351803-25351803 on Build GRCh38
rs769907546 Chr.4: 25351734-25351734 on Build GRCh38
rs560401134 Chr.4: 25351795-25351795 on Build GRCh38
rs1046119125 Chr.4: 25351820-25351820 on Build GRCh38
rs866910141 Chr.4: 25351628-25351628 on Build GRCh38
rs745354498 Chr.4: 25351552-25351552 on Build GRCh38
rs755637965 Chr.4: 25351646-25351646 on Build GRCh38
rs763895966 Chr.4: 25351612-25351612 on Build GRCh38
rs577069311 Chr.4: 25351851-25351851 on Build GRCh38
rs759742394 Chr.4: 25351732-25351732 on Build GRCh38
rs570683425 Chr.4: 25351541-25351541 on Build GRCh38
rs200632828 Chr.4: 25351710-25351710 on Build GRCh38
rs1018532936 Chr.4: 25351950-25351950 on Build GRCh38
rs779158515 Chr.4: 25351592-25351592 on Build GRCh38
rs201086181 Chr.4: 25351704-25351704 on Build GRCh38
rs965356261 Chr.4: 25351975-25351975 on Build GRCh38
rs546321206 Chr.4: 25351886-25351886 on Build GRCh38
rs892125984 Chr.4: 25351976-25351976 on Build GRCh38
rs976813480 Chr.4: 25351680-25351680 on Build GRCh38
rs753464905 Chr.4: 25351682-25351682 on Build GRCh38
rs779743271 Chr.4: 25351663-25351663 on Build GRCh38
rs958501164 Chr.4: 25351895-25351895 on Build GRCh38
rs531701968 Chr.4: 25351984-25351984 on Build GRCh38
rs771042100 Chr.4: 25351600-25351600 on Build GRCh38
rs751468207 Chr.4: 25351540-25351540 on Build GRCh38
rs373521805 Chr.4: 25351610-25351610 on Build GRCh38
rs1050376509 Chr.4: 25351919-25351919 on Build GRCh38
rs760723180 Chr.4: 25351599-25351599 on Build GRCh38
rs751303036 Chr.4: 25351629-25351629 on Build GRCh38
rs559806537 Chr.4: 25351656-25351657 on Build GRCh38
rs538363295 Chr.4: 25351597-25351597 on Build GRCh38
rs1018670648 Chr.4: 25351627-25351627 on Build GRCh38
rs182505131 Chr.4: 25351645-25351645 on Build GRCh38
rs908635590 Chr.4: 25351849-25351849 on Build GRCh38
rs755731692 Chr.4: 25351567-25351567 on Build GRCh38
rs375141562 Chr.4: 25351701-25351701 on Build GRCh38
rs911234930 Chr.4: 25351631-25351631 on Build GRCh38
rs57521858 Chr.4: 25351693-25351693 on Build GRCh38
rs781341865 Chr.4: 25351548-25351548 on Build GRCh38
rs185100272 Chr.4: 25351940-25351940 on Build GRCh38
rs753366111 Chr.4: 25351708-25351711 on Build GRCh38
rs776573644 Chr.4: 25351718-25351718 on Build GRCh38
rs763855383 Chr.4: 25351656-25351658 on Build GRCh38
rs748923072 Chr.4: 25351577-25351577 on Build GRCh38
rs1032548397 Chr.4: 25351533-25351533 on Build GRCh38
rs899020499 Chr.4: 25351878-25351878 on Build GRCh38
rs554261131 Chr.4: 25351699-25351699 on Build GRCh38
rs770954617 Chr.4: 25351716-25351716 on Build GRCh38
rs776797550 Chr.4: 25351603-25351603 on Build GRCh38
rs902305347 Chr.4: 25351835-25351835 on Build GRCh38
rs748456903 Chr.4: 25351694-25351694 on Build GRCh38
rs531180947 Chr.4: 25351751-25351751 on Build GRCh38
rs866352550 Chr.4: 25351588-25351588 on Build GRCh38
rs575136135 Chr.4: 25351692-25351692 on Build GRCh38
rs1038858244 Chr.4: 25351856-25351856 on Build GRCh38
rs761737766 Chr.4: 25351632-25351632 on Build GRCh38
rs939346341 Chr.4: 25351763-25351763 on Build GRCh38
rs1013229875 Chr.4: 25351816-25351816 on Build GRCh38
rs760215323 Chr.4: 25351632-25351632 on Build GRCh38
rs985827827 Chr.4: 25351626-25351626 on Build GRCh38
rs781424645 Chr.4: 25351983-25351983 on Build GRCh38
rs370704925 Chr.4: 25351586-25351586 on Build GRCh38
rs889006913 Chr.4: 25351924-25351924 on Build GRCh38
rs779820555 Chr.4: 25351571-25351571 on Build GRCh38
rs767533921 Chr.4: 25351642-25351642 on Build GRCh38

View Full Product Details