Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41603784-41604286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIMCH1
Entrez Gene ID
22998
Gene Name
LIM and calponin homology domains 1
Gene Aliases
LIMCH1A, LMO7B
UniGene
Hs.335163
Gene Chromosome Location
Chr.4: 41359607-41700044 on Build GRCh38
Transcripts
NM_001112717.2 NM_001112718.2 NM_001289122.1 NM_001289124.1 NM_014988.3 XM_005248057.1 XM_005248058.1 XM_005248060.1 XM_005248061.1 XM_005248067.2 XM_005248072.1 XM_005248074.1 XM_005248075.1 XM_006713996.1 XM_011513642.1 XM_011513643.1 XM_011513644.1 XM_011513645.1 XM_011513646.1 XM_011513648.1 XM_011513651.2 XM_011513653.1 XM_011513654.1 XM_011513658.2 XM_017007894.1 XM_017007895.1 XM_017007896.1 XM_017007897.1 XM_017007898.1 XM_017007899.1 XM_017007900.1 XM_017007901.1 XM_017007902.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3661092 c.363G>C p.R121S Chr.4: 41603893-41603893 on Build GRCh38
7146241 c.361A>C p.R121R Chr.4: 41603891-41603891 on Build GRCh38
1618818 c.363G>C p.R121S Chr.4: 41603893-41603893 on Build GRCh38
3603780 c.345G>A p.K115K Chr.4: 41603875-41603875 on Build GRCh38
7146242 c.361A>C p.R121R Chr.4: 41603891-41603891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892646676 Chr.4: 41604177-41604177 on Build GRCh38
rs974200802 Chr.4: 41604013-41604013 on Build GRCh38
rs114753026 Chr.4: 41604139-41604139 on Build GRCh38
rs906821913 Chr.4: 41603841-41603841 on Build GRCh38
rs953089456 Chr.4: 41603960-41603960 on Build GRCh38
rs867998227 Chr.4: 41603875-41603875 on Build GRCh38
rs971852914 Chr.4: 41604044-41604044 on Build GRCh38
rs375058873 Chr.4: 41603866-41603866 on Build GRCh38
rs951517960 Chr.4: 41604073-41604073 on Build GRCh38
rs1024900083 Chr.4: 41604236-41604236 on Build GRCh38
rs982939472 Chr.4: 41604094-41604094 on Build GRCh38
rs763581472 Chr.4: 41603842-41603842 on Build GRCh38
rs768235215 Chr.4: 41604204-41604204 on Build GRCh38
rs965993667 Chr.4: 41603983-41603983 on Build GRCh38
rs948066684 Chr.4: 41604102-41604102 on Build GRCh38
rs370678060 Chr.4: 41604160-41604160 on Build GRCh38
rs928216923 Chr.4: 41604158-41604158 on Build GRCh38
rs1043462962 Chr.4: 41604219-41604219 on Build GRCh38
rs550944519 Chr.4: 41604106-41604106 on Build GRCh38
rs542091990 Chr.4: 41604051-41604051 on Build GRCh38
rs768246453 Chr.4: 41604111-41604111 on Build GRCh38
rs758327781 Chr.4: 41603839-41603839 on Build GRCh38
rs892202649 Chr.4: 41604186-41604186 on Build GRCh38
rs73135464 Chr.4: 41604097-41604097 on Build GRCh38
rs745469963 Chr.4: 41603877-41603877 on Build GRCh38
rs368665156 Chr.4: 41603882-41603882 on Build GRCh38
rs769190649 Chr.4: 41603945-41603945 on Build GRCh38
rs985971644 Chr.4: 41603887-41603887 on Build GRCh38
rs946432378 Chr.4: 41604213-41604213 on Build GRCh38
rs1045314674 Chr.4: 41604119-41604119 on Build GRCh38
rs1052518203 Chr.4: 41604172-41604172 on Build GRCh38
rs756519702 Chr.4: 41603878-41603878 on Build GRCh38
rs1007658821 Chr.4: 41603969-41603969 on Build GRCh38
rs754260659 Chr.4: 41603946-41603947 on Build GRCh38
rs919102013 Chr.4: 41604062-41604062 on Build GRCh38
rs562385166 Chr.4: 41604087-41604087 on Build GRCh38
rs1010661934 Chr.4: 41603924-41603924 on Build GRCh38
rs373056498 Chr.4: 41604082-41604082 on Build GRCh38
rs778276409 Chr.4: 41603944-41603945 on Build GRCh38
rs752628662 Chr.4: 41603837-41603837 on Build GRCh38
rs376896920 Chr.4: 41603915-41603915 on Build GRCh38
rs1022348048 Chr.4: 41603948-41603948 on Build GRCh38
rs751080916 Chr.4: 41603854-41603854 on Build GRCh38
rs1009395388 Chr.4: 41604199-41604199 on Build GRCh38
rs1018868341 Chr.4: 41603978-41603978 on Build GRCh38
rs564151510 Chr.4: 41604112-41604112 on Build GRCh38
rs756687143 Chr.4: 41603860-41603860 on Build GRCh38

View Full Product Details