Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 48497262-48497806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRYL
Entrez Gene ID
285527
Gene Name
FRY like transcription coactivator
Gene Aliases
AF4p12, KIAA0826, MOR2
UniGene
Hs.595553
Gene Chromosome Location
Chr.4: 48497363-48780312 on Build GRCh38
Transcripts
NM_015030.1 XM_005248090.4 XM_005248093.4 XM_011513680.2 XM_011513681.2 XM_011513682.2 XM_011513683.2 XM_011513684.2 XM_017008046.1

SNP Detail

SNP ID Chromosome Location
rs144135465 Chr.4: 48497537-48497537 on Build GRCh38
rs973230129 Chr.4: 48497449-48497449 on Build GRCh38
rs769080396 Chr.4: 48497599-48497599 on Build GRCh38
rs572053021 Chr.4: 48497503-48497503 on Build GRCh38
rs534561355 Chr.4: 48497749-48497749 on Build GRCh38
rs1030752032 Chr.4: 48497329-48497329 on Build GRCh38
rs1053964498 Chr.4: 48497675-48497676 on Build GRCh38
rs538121956 Chr.4: 48497624-48497625 on Build GRCh38
rs927017818 Chr.4: 48497481-48497481 on Build GRCh38
rs779500305 Chr.4: 48497291-48497291 on Build GRCh38
rs996706710 Chr.4: 48497717-48497717 on Build GRCh38
rs949648861 Chr.4: 48497575-48497575 on Build GRCh38
rs368744848 Chr.4: 48497308-48497308 on Build GRCh38
rs558549243 Chr.4: 48497483-48497483 on Build GRCh38
rs994597553 Chr.4: 48497769-48497769 on Build GRCh38
rs924205844 Chr.4: 48497626-48497626 on Build GRCh38
rs556794080 Chr.4: 48497533-48497533 on Build GRCh38
rs147023375 Chr.4: 48497299-48497299 on Build GRCh38
rs890483550 Chr.4: 48497754-48497754 on Build GRCh38
rs916934433 Chr.4: 48497572-48497572 on Build GRCh38
rs748410017 Chr.4: 48497748-48497748 on Build GRCh38
rs954649712 Chr.4: 48497337-48497337 on Build GRCh38
rs1014698180 Chr.4: 48497362-48497364 on Build GRCh38
rs138560426 Chr.4: 48497396-48497396 on Build GRCh38
rs900550182 Chr.4: 48497697-48497697 on Build GRCh38
rs777222882 Chr.4: 48497726-48497726 on Build GRCh38
rs963207138 Chr.4: 48497364-48497364 on Build GRCh38
rs1034183626 Chr.4: 48497778-48497779 on Build GRCh38
rs1005245089 Chr.4: 48497343-48497343 on Build GRCh38
rs143384146 Chr.4: 48497616-48497616 on Build GRCh38
rs371004619 Chr.4: 48497628-48497628 on Build GRCh38
rs868506238 Chr.4: 48497380-48497380 on Build GRCh38
rs1014771847 Chr.4: 48497767-48497767 on Build GRCh38
rs377362439 Chr.4: 48497646-48497647 on Build GRCh38
rs571132240 Chr.4: 48497304-48497304 on Build GRCh38
rs574038890 Chr.4: 48497633-48497633 on Build GRCh38
rs992502893 Chr.4: 48497566-48497567 on Build GRCh38
rs1004784366 Chr.4: 48497755-48497755 on Build GRCh38

View Full Product Details