Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 48998368-48998860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWH43
Entrez Gene ID
80157
Gene Name
cell wall biogenesis 43 C-terminal homolog
Gene Aliases
CWH43-C
UniGene
Hs.479703
Gene Chromosome Location
Chr.4: 48986247-49062079 on Build GRCh38
Transcripts
NM_001286791.1 NM_025087.2 XM_011513755.1 XM_011513756.2 XM_011513757.2 XM_011513758.1 XM_011513759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3974931 c.772C>G p.R258G Chr.4: 48998518-48998518 on Build GRCh38
1055869 c.773G>A p.R258H Chr.4: 48998519-48998519 on Build GRCh38
7363391 c.779C>T p.T260I Chr.4: 48998525-48998525 on Build GRCh38
1055870 c.790T>C p.W264R Chr.4: 48998536-48998536 on Build GRCh38
336778 c.791G>C p.W264S Chr.4: 48998537-48998537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140373008 Chr.4: 48998496-48998496 on Build GRCh38
rs757053654 Chr.4: 48998467-48998467 on Build GRCh38
rs768284538 Chr.4: 48998581-48998581 on Build GRCh38
rs1004886331 Chr.4: 48998803-48998803 on Build GRCh38
rs578049659 Chr.4: 48998430-48998430 on Build GRCh38
rs1016444530 Chr.4: 48998617-48998617 on Build GRCh38
rs775031029 Chr.4: 48998511-48998511 on Build GRCh38
rs868257485 Chr.4: 48998458-48998458 on Build GRCh38
rs747648125 Chr.4: 48998599-48998599 on Build GRCh38
rs886402091 Chr.4: 48998772-48998772 on Build GRCh38
rs766898371 Chr.4: 48998403-48998403 on Build GRCh38
rs960300300 Chr.4: 48998629-48998629 on Build GRCh38
rs1004937294 Chr.4: 48998616-48998616 on Build GRCh38
rs138872325 Chr.4: 48998635-48998635 on Build GRCh38
rs35029152 Chr.4: 48998470-48998471 on Build GRCh38
rs115610972 Chr.4: 48998754-48998754 on Build GRCh38
rs771952137 Chr.4: 48998699-48998699 on Build GRCh38
rs779856872 Chr.4: 48998714-48998714 on Build GRCh38
rs748858371 Chr.4: 48998497-48998497 on Build GRCh38
rs111714807 Chr.4: 48998623-48998623 on Build GRCh38
rs933225650 Chr.4: 48998760-48998760 on Build GRCh38
rs778220012 Chr.4: 48998652-48998652 on Build GRCh38
rs780991037 Chr.4: 48998544-48998544 on Build GRCh38
rs760940270 Chr.4: 48998526-48998526 on Build GRCh38
rs570165324 Chr.4: 48998405-48998405 on Build GRCh38
rs745639219 Chr.4: 48998482-48998482 on Build GRCh38
rs761372860 Chr.4: 48998828-48998828 on Build GRCh38
rs757280793 Chr.4: 48998540-48998540 on Build GRCh38
rs781495269 Chr.4: 48998615-48998615 on Build GRCh38
rs771737965 Chr.4: 48998579-48998580 on Build GRCh38
rs780987356 Chr.4: 48998472-48998472 on Build GRCh38
rs142006288 Chr.4: 48998667-48998667 on Build GRCh38
rs746743613 Chr.4: 48998719-48998719 on Build GRCh38
rs752833301 Chr.4: 48998622-48998622 on Build GRCh38
rs754034849 Chr.4: 48998528-48998528 on Build GRCh38
rs756300031 Chr.4: 48998412-48998412 on Build GRCh38
rs200658416 Chr.4: 48998428-48998428 on Build GRCh38
rs183224687 Chr.4: 48998746-48998746 on Build GRCh38
rs759322134 Chr.4: 48998553-48998553 on Build GRCh38
rs368956715 Chr.4: 48998579-48998579 on Build GRCh38
rs772334679 Chr.4: 48998519-48998519 on Build GRCh38
rs6850199 Chr.4: 48998833-48998833 on Build GRCh38
rs142708462 Chr.4: 48998525-48998525 on Build GRCh38
rs917553222 Chr.4: 48998640-48998640 on Build GRCh38
rs78996420 Chr.4: 48998459-48998459 on Build GRCh38
rs755842629 Chr.4: 48998570-48998570 on Build GRCh38
rs762343467 Chr.4: 48998518-48998518 on Build GRCh38
rs759631753 Chr.4: 48998533-48998533 on Build GRCh38
rs766661541 Chr.4: 48998527-48998527 on Build GRCh38
rs552206176 Chr.4: 48998578-48998578 on Build GRCh38
rs146230326 Chr.4: 48998582-48998582 on Build GRCh38
rs765237802 Chr.4: 48998538-48998538 on Build GRCh38
rs867598172 Chr.4: 48998821-48998821 on Build GRCh38
rs775568185 Chr.4: 48998594-48998602 on Build GRCh38
rs374173676 Chr.4: 48998568-48998568 on Build GRCh38
rs1037822703 Chr.4: 48998813-48998813 on Build GRCh38
rs900406410 Chr.4: 48998550-48998550 on Build GRCh38
rs376158233 Chr.4: 48998455-48998455 on Build GRCh38
rs570009991 Chr.4: 48998799-48998799 on Build GRCh38
rs768233261 Chr.4: 48998510-48998510 on Build GRCh38
rs766495518 Chr.4: 48998426-48998426 on Build GRCh38
rs921863428 Chr.4: 48998759-48998759 on Build GRCh38
rs531938642 Chr.4: 48998768-48998768 on Build GRCh38
rs550198794 Chr.4: 48998792-48998792 on Build GRCh38
rs769560296 Chr.4: 48998490-48998490 on Build GRCh38

View Full Product Details