Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 52594703-52595208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP46
Entrez Gene ID
64854
Gene Name
ubiquitin specific peptidase 46
Gene Aliases
-
UniGene
Hs.7966
Gene Chromosome Location
Chr.4: 52590960-52659335 on Build GRCh38
Transcripts
NM_001134223.1 NM_001286767.1 NM_001286768.1 NM_022832.3 XM_017008557.1

SNP Detail

SNP ID Chromosome Location
rs11351768 Chr.4: 52595043-52595043 on Build GRCh38
rs980964932 Chr.4: 52594787-52594787 on Build GRCh38
rs1045181440 Chr.4: 52594821-52594821 on Build GRCh38
rs1052047776 Chr.4: 52595105-52595105 on Build GRCh38
rs984408096 Chr.4: 52595029-52595032 on Build GRCh38
rs939137410 Chr.4: 52595102-52595102 on Build GRCh38
rs561253269 Chr.4: 52594858-52594858 on Build GRCh38
rs758607484 Chr.4: 52594878-52594878 on Build GRCh38
rs530518812 Chr.4: 52595176-52595176 on Build GRCh38
rs766543375 Chr.4: 52594858-52594859 on Build GRCh38
rs1019015574 Chr.4: 52594957-52594957 on Build GRCh38
rs960872956 Chr.4: 52595177-52595177 on Build GRCh38
rs980591937 Chr.4: 52595062-52595062 on Build GRCh38
rs968159782 Chr.4: 52595129-52595129 on Build GRCh38
rs190662232 Chr.4: 52594758-52594758 on Build GRCh38
rs868772807 Chr.4: 52594855-52594855 on Build GRCh38
rs1039544211 Chr.4: 52594925-52594925 on Build GRCh38
rs909809584 Chr.4: 52595033-52595034 on Build GRCh38
rs4865390 Chr.4: 52594908-52594908 on Build GRCh38
rs560865370 Chr.4: 52594956-52594956 on Build GRCh38
rs927731206 Chr.4: 52594849-52594849 on Build GRCh38
rs144121892 Chr.4: 52595183-52595183 on Build GRCh38
rs752823694 Chr.4: 52594865-52594865 on Build GRCh38
rs759290244 Chr.4: 52595164-52595164 on Build GRCh38
rs531364557 Chr.4: 52595094-52595094 on Build GRCh38
rs544762054 Chr.4: 52594762-52594762 on Build GRCh38
rs951329292 Chr.4: 52595021-52595021 on Build GRCh38
rs796346202 Chr.4: 52595034-52595034 on Build GRCh38
rs1011613800 Chr.4: 52594892-52594892 on Build GRCh38
rs1002286538 Chr.4: 52595167-52595167 on Build GRCh38
rs386674392 Chr.4: 52594961-52594963 on Build GRCh38
rs561217922 Chr.4: 52594773-52594773 on Build GRCh38
rs1043984915 Chr.4: 52594897-52594897 on Build GRCh38
rs1044338193 Chr.4: 52595149-52595149 on Build GRCh38
rs966139279 Chr.4: 52594963-52594963 on Build GRCh38
rs558820500 Chr.4: 52594756-52594756 on Build GRCh38
rs562783843 Chr.4: 52595100-52595100 on Build GRCh38
rs1034266755 Chr.4: 52595170-52595170 on Build GRCh38
rs899660344 Chr.4: 52594938-52594938 on Build GRCh38
rs909748041 Chr.4: 52594732-52594732 on Build GRCh38
rs892474719 Chr.4: 52595071-52595071 on Build GRCh38
rs946410051 Chr.4: 52595131-52595131 on Build GRCh38
rs545166307 Chr.4: 52594736-52594736 on Build GRCh38
rs941098924 Chr.4: 52594915-52594915 on Build GRCh38
rs532724271 Chr.4: 52595034-52595034 on Build GRCh38
rs1022286244 Chr.4: 52595125-52595125 on Build GRCh38
rs1050989876 Chr.4: 52594968-52594968 on Build GRCh38
rs762504088 Chr.4: 52594800-52594800 on Build GRCh38
rs926803591 Chr.4: 52594789-52594789 on Build GRCh38
rs1026355236 Chr.4: 52595000-52595000 on Build GRCh38
rs183255917 Chr.4: 52594904-52594904 on Build GRCh38
rs992384891 Chr.4: 52594808-52594808 on Build GRCh38

View Full Product Details