Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 53498558-53499065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LNX1
Entrez Gene ID
84708
Gene Name
ligand of numb-protein X 1
Gene Aliases
LNX, MPDZ, PDZRN2
UniGene
Hs.518760
Gene Chromosome Location
Chr.4: 53458264-53652524 on Build GRCh38
Transcripts
NM_001126328.2 NM_032622.2 XM_005265785.4 XM_017008776.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1319332 c.966C>T p.D322D Chr.4: 53498653-53498653 on Build GRCh38
1291924 c.903C>T p.I301I Chr.4: 53498716-53498716 on Build GRCh38
5866516 c.526A>T p.I176F Chr.4: 53498805-53498805 on Build GRCh38
3825975 c.528T>A p.I176I Chr.4: 53498803-53498803 on Build GRCh38
3725664 c.894G>T p.L298L Chr.4: 53498725-53498725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913138230 Chr.4: 53498967-53498967 on Build GRCh38
rs557504224 Chr.4: 53498938-53498938 on Build GRCh38
rs199552515 Chr.4: 53498727-53498727 on Build GRCh38
rs777810241 Chr.4: 53498731-53498731 on Build GRCh38
rs768621160 Chr.4: 53498765-53498765 on Build GRCh38
rs1042876052 Chr.4: 53499007-53499007 on Build GRCh38
rs773313418 Chr.4: 53498651-53498651 on Build GRCh38
rs746409231 Chr.4: 53498608-53498608 on Build GRCh38
rs774162598 Chr.4: 53498780-53498780 on Build GRCh38
rs752763825 Chr.4: 53498812-53498812 on Build GRCh38
rs767219285 Chr.4: 53498784-53498784 on Build GRCh38
rs766974602 Chr.4: 53498683-53498683 on Build GRCh38
rs145916448 Chr.4: 53498695-53498695 on Build GRCh38
rs747100679 Chr.4: 53498736-53498736 on Build GRCh38
rs753131561 Chr.4: 53498836-53498836 on Build GRCh38
rs377045257 Chr.4: 53498830-53498830 on Build GRCh38
rs768948566 Chr.4: 53498624-53498624 on Build GRCh38
rs979826855 Chr.4: 53498753-53498753 on Build GRCh38
rs372851227 Chr.4: 53498674-53498674 on Build GRCh38
rs754271968 Chr.4: 53498697-53498697 on Build GRCh38
rs765866296 Chr.4: 53498822-53498822 on Build GRCh38
rs760179311 Chr.4: 53498803-53498803 on Build GRCh38
rs758633692 Chr.4: 53498838-53498838 on Build GRCh38
rs963143390 Chr.4: 53498746-53498746 on Build GRCh38
rs768209163 Chr.4: 53498592-53498592 on Build GRCh38
rs368391218 Chr.4: 53498592-53498592 on Build GRCh38
rs138813024 Chr.4: 53498737-53498737 on Build GRCh38
rs780495550 Chr.4: 53498612-53498612 on Build GRCh38
rs749983896 Chr.4: 53498786-53498786 on Build GRCh38
rs762970381 Chr.4: 53498760-53498760 on Build GRCh38
rs761474282 Chr.4: 53498781-53498781 on Build GRCh38
rs762846749 Chr.4: 53498673-53498673 on Build GRCh38
rs535013065 Chr.4: 53498698-53498698 on Build GRCh38
rs946821493 Chr.4: 53499031-53499031 on Build GRCh38
rs569711152 Chr.4: 53498940-53498940 on Build GRCh38
rs757471467 Chr.4: 53498883-53498883 on Build GRCh38
rs542109970 Chr.4: 53498696-53498696 on Build GRCh38
rs778062890 Chr.4: 53498859-53498859 on Build GRCh38
rs866500759 Chr.4: 53498710-53498710 on Build GRCh38
rs529508524 Chr.4: 53498625-53498625 on Build GRCh38
rs772097288 Chr.4: 53498649-53498649 on Build GRCh38
rs760634604 Chr.4: 53498661-53498661 on Build GRCh38
rs749673105 Chr.4: 53498621-53498621 on Build GRCh38
rs776207255 Chr.4: 53498591-53498592 on Build GRCh38
rs189189345 Chr.4: 53498669-53498669 on Build GRCh38
rs147847312 Chr.4: 53498599-53498599 on Build GRCh38
rs200237039 Chr.4: 53498826-53498826 on Build GRCh38
rs757665685 Chr.4: 53498952-53498952 on Build GRCh38
rs142190654 Chr.4: 53498668-53498668 on Build GRCh38
rs200781307 Chr.4: 53498857-53498858 on Build GRCh38
rs945897549 Chr.4: 53499006-53499006 on Build GRCh38
rs775636773 Chr.4: 53498739-53498739 on Build GRCh38
rs201157691 Chr.4: 53498682-53498682 on Build GRCh38
rs772373954 Chr.4: 53498730-53498730 on Build GRCh38
rs535641298 Chr.4: 53499039-53499039 on Build GRCh38
rs747229435 Chr.4: 53498863-53498863 on Build GRCh38
rs374448115 Chr.4: 53498688-53498688 on Build GRCh38
rs756580885 Chr.4: 53498609-53498609 on Build GRCh38
rs181555457 Chr.4: 53498899-53498899 on Build GRCh38
rs748392876 Chr.4: 53498627-53498627 on Build GRCh38
rs572184157 Chr.4: 53498654-53498654 on Build GRCh38
rs35082655 Chr.4: 53498823-53498823 on Build GRCh38
rs926961256 Chr.4: 53498757-53498757 on Build GRCh38

View Full Product Details