Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 67833115-67833614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11D
Entrez Gene ID
9407
Gene Name
transmembrane protease, serine 11D
Gene Aliases
HAT
UniGene
Hs.132195
Gene Chromosome Location
Chr.4: 67820876-67885063 on Build GRCh38
Transcripts
NM_004262.2 XM_005265710.4 XM_017008851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7308951 c.651C>A p.I217I Chr.4: 67833245-67833245 on Build GRCh38
1056779 c.595C>T p.P199S Chr.4: 67833301-67833301 on Build GRCh38
3918008 c.578C>T p.A193V Chr.4: 67833318-67833318 on Build GRCh38
5401424 c.541A>G p.T181A Chr.4: 67833355-67833355 on Build GRCh38
5355834 c.515-8C>A p.? Chr.4: 67833389-67833389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765604949 Chr.4: 67833221-67833221 on Build GRCh38
rs958160824 Chr.4: 67833494-67833494 on Build GRCh38
rs953909639 Chr.4: 67833562-67833562 on Build GRCh38
rs556667530 Chr.4: 67833491-67833491 on Build GRCh38
rs191606172 Chr.4: 67833463-67833463 on Build GRCh38
rs115858582 Chr.4: 67833217-67833217 on Build GRCh38
rs1014609472 Chr.4: 67833365-67833365 on Build GRCh38
rs377183393 Chr.4: 67833257-67833257 on Build GRCh38
rs745530529 Chr.4: 67833164-67833164 on Build GRCh38
rs989661968 Chr.4: 67833518-67833518 on Build GRCh38
rs753109563 Chr.4: 67833222-67833222 on Build GRCh38
rs369425744 Chr.4: 67833194-67833194 on Build GRCh38
rs752075532 Chr.4: 67833239-67833239 on Build GRCh38
rs201995425 Chr.4: 67833349-67833349 on Build GRCh38
rs765883802 Chr.4: 67833413-67833413 on Build GRCh38
rs780598557 Chr.4: 67833251-67833251 on Build GRCh38
rs944769684 Chr.4: 67833192-67833192 on Build GRCh38
rs768774658 Chr.4: 67833174-67833174 on Build GRCh38
rs755776110 Chr.4: 67833260-67833260 on Build GRCh38
rs372779145 Chr.4: 67833238-67833238 on Build GRCh38
rs373004299 Chr.4: 67833283-67833283 on Build GRCh38
rs376485314 Chr.4: 67833276-67833276 on Build GRCh38
rs767035472 Chr.4: 67833374-67833374 on Build GRCh38
rs183866667 Chr.4: 67833482-67833482 on Build GRCh38
rs762267596 Chr.4: 67833371-67833371 on Build GRCh38
rs1033034590 Chr.4: 67833141-67833141 on Build GRCh38
rs555212029 Chr.4: 67833403-67833403 on Build GRCh38
rs1032766794 Chr.4: 67833490-67833490 on Build GRCh38
rs914167088 Chr.4: 67833253-67833253 on Build GRCh38
rs902325060 Chr.4: 67833481-67833481 on Build GRCh38
rs780840425 Chr.4: 67833163-67833163 on Build GRCh38
rs770926330 Chr.4: 67833197-67833197 on Build GRCh38
rs763076296 Chr.4: 67833414-67833414 on Build GRCh38
rs763436098 Chr.4: 67833235-67833235 on Build GRCh38
rs868830460 Chr.4: 67833309-67833309 on Build GRCh38
rs976181566 Chr.4: 67833586-67833586 on Build GRCh38
rs897095759 Chr.4: 67833353-67833353 on Build GRCh38
rs371200759 Chr.4: 67833244-67833244 on Build GRCh38
rs1024450544 Chr.4: 67833153-67833153 on Build GRCh38
rs114055011 Chr.4: 67833299-67833299 on Build GRCh38
rs369417549 Chr.4: 67833300-67833300 on Build GRCh38
rs1054757379 Chr.4: 67833339-67833339 on Build GRCh38
rs766967202 Chr.4: 67833581-67833581 on Build GRCh38
rs759781505 Chr.4: 67833311-67833311 on Build GRCh38
rs764466422 Chr.4: 67833352-67833352 on Build GRCh38
rs986249434 Chr.4: 67833169-67833169 on Build GRCh38
rs761291610 Chr.4: 67833379-67833379 on Build GRCh38
rs1032253266 Chr.4: 67833493-67833493 on Build GRCh38
rs775827190 Chr.4: 67833343-67833343 on Build GRCh38
rs372118955 Chr.4: 67833179-67833179 on Build GRCh38
rs1017581496 Chr.4: 67833578-67833578 on Build GRCh38
rs188760816 Chr.4: 67833496-67833496 on Build GRCh38
rs756766520 Chr.4: 67833159-67833159 on Build GRCh38
rs779888666 Chr.4: 67833266-67833266 on Build GRCh38
rs769591340 Chr.4: 67833167-67833167 on Build GRCh38
rs768636560 Chr.4: 67833295-67833295 on Build GRCh38
rs914135007 Chr.4: 67833545-67833545 on Build GRCh38
rs1023959018 Chr.4: 67833424-67833424 on Build GRCh38
rs769965359 Chr.4: 67833314-67833314 on Build GRCh38
rs114074287 Chr.4: 67833181-67833181 on Build GRCh38
rs141438601 Chr.4: 67833282-67833282 on Build GRCh38
rs749218787 Chr.4: 67833173-67833173 on Build GRCh38
rs376245737 Chr.4: 67833370-67833370 on Build GRCh38
rs768572405 Chr.4: 67833281-67833281 on Build GRCh38
rs192465409 Chr.4: 67833550-67833550 on Build GRCh38
rs114503550 Chr.4: 67833209-67833209 on Build GRCh38
rs976675050 Chr.4: 67833233-67833233 on Build GRCh38
rs754768106 Chr.4: 67833430-67833430 on Build GRCh38
rs1007569373 Chr.4: 67833563-67833563 on Build GRCh38
rs138505736 Chr.4: 67833517-67833517 on Build GRCh38

View Full Product Details