Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68189476-68189985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTLP10
Entrez Gene ID
100130017
Gene Name
ferritin light chain pseudogene 10
Gene Aliases
-
UniGene
Hs.582037
Gene Chromosome Location
Chr.4: 68182292-68212470 on Build GRCh38
Transcripts
NR_015446.1 NR_104048.1

Gene Symbol
TMPRSS11BNL
Entrez Gene ID
401136
Gene Name
TMPRSS11B N-terminal like, pseudogene
Gene Aliases
-
UniGene
Hs.652046
Gene Chromosome Location
Chr.4: 68184129-68218080 on Build GRCh38
Transcripts
NR_015446.1 NR_104048.1

SNP Detail

SNP ID Chromosome Location
rs543525591 Chr.4: 68189620-68189620 on Build GRCh38
rs919408005 Chr.4: 68189704-68189704 on Build GRCh38
rs902370449 Chr.4: 68189954-68189954 on Build GRCh38
rs764114211 Chr.4: 68189634-68189634 on Build GRCh38
rs966688621 Chr.4: 68189690-68189690 on Build GRCh38
rs145046864 Chr.4: 68189771-68189771 on Build GRCh38
rs984546195 Chr.4: 68189793-68189793 on Build GRCh38
rs887388673 Chr.4: 68189540-68189540 on Build GRCh38
rs754310045 Chr.4: 68189698-68189698 on Build GRCh38
rs577686077 Chr.4: 68189554-68189554 on Build GRCh38
rs372015140 Chr.4: 68189617-68189617 on Build GRCh38
rs1013125203 Chr.4: 68189943-68189943 on Build GRCh38
rs770132613 Chr.4: 68189583-68189583 on Build GRCh38
rs1044480639 Chr.4: 68189949-68189949 on Build GRCh38
rs368770484 Chr.4: 68189631-68189631 on Build GRCh38
rs12649371 Chr.4: 68189891-68189891 on Build GRCh38
rs1047398732 Chr.4: 68189807-68189807 on Build GRCh38
rs767914661 Chr.4: 68189727-68189727 on Build GRCh38
rs950831656 Chr.4: 68189717-68189717 on Build GRCh38
rs1013427590 Chr.4: 68189584-68189584 on Build GRCh38
rs1051925754 Chr.4: 68189890-68189890 on Build GRCh38
rs140139754 Chr.4: 68189586-68189586 on Build GRCh38
rs887455044 Chr.4: 68189514-68189514 on Build GRCh38
rs373450166 Chr.4: 68189780-68189780 on Build GRCh38
rs556775379 Chr.4: 68189813-68189813 on Build GRCh38
rs779349313 Chr.4: 68189953-68189953 on Build GRCh38
rs930296359 Chr.4: 68189803-68189803 on Build GRCh38
rs964608415 Chr.4: 68189711-68189711 on Build GRCh38
rs949285171 Chr.4: 68189892-68189892 on Build GRCh38
rs1020729827 Chr.4: 68189591-68189591 on Build GRCh38
rs903299074 Chr.4: 68189592-68189592 on Build GRCh38
rs1000221000 Chr.4: 68189622-68189622 on Build GRCh38
rs937639133 Chr.4: 68189526-68189526 on Build GRCh38
rs763486213 Chr.4: 68189628-68189628 on Build GRCh38
rs1055053402 Chr.4: 68189527-68189527 on Build GRCh38
rs1042263410 Chr.4: 68189927-68189927 on Build GRCh38
rs930348816 Chr.4: 68189522-68189522 on Build GRCh38
rs867893378 Chr.4: 68189600-68189600 on Build GRCh38
rs537925966 Chr.4: 68189505-68189505 on Build GRCh38
rs1038662154 Chr.4: 68189502-68189502 on Build GRCh38
rs956166324 Chr.4: 68189665-68189665 on Build GRCh38
rs750658895 Chr.4: 68189886-68189886 on Build GRCh38
rs576241268 Chr.4: 68189832-68189832 on Build GRCh38
rs974894744 Chr.4: 68189728-68189728 on Build GRCh38
rs940268923 Chr.4: 68189878-68189878 on Build GRCh38
rs908915844 Chr.4: 68189805-68189805 on Build GRCh38
rs1004355523 Chr.4: 68189564-68189564 on Build GRCh38
rs775910194 Chr.4: 68189612-68189612 on Build GRCh38
rs150297941 Chr.4: 68189608-68189608 on Build GRCh38
rs551395580 Chr.4: 68189576-68189576 on Build GRCh38
rs1017895479 Chr.4: 68189664-68189664 on Build GRCh38
rs554456659 Chr.4: 68189647-68189647 on Build GRCh38
rs376749045 Chr.4: 68189595-68189595 on Build GRCh38
rs908774850 Chr.4: 68189811-68189811 on Build GRCh38
rs545143004 Chr.4: 68189887-68189887 on Build GRCh38
rs563325978 Chr.4: 68189703-68189703 on Build GRCh38
rs986499988 Chr.4: 68189639-68189639 on Build GRCh38

View Full Product Details