Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 69107885-69108398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGT2B7
Entrez Gene ID
7364
Gene Name
UDP glucuronosyltransferase family 2 member B7
Gene Aliases
UDPGT 2B9, UDPGT2B7, UDPGTH2, UGT2B9
UniGene
Hs.654424
Gene Chromosome Location
Chr.4: 69051264-69112987 on Build GRCh38
Transcripts
NM_001074.2 XM_005265702.3 XM_011532229.2 XM_011532230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6337062 c.1292G>C p.R431T Chr.4: 69108304-69108304 on Build GRCh38
5918553 c.1310C>T p.S437L Chr.4: 69108322-69108322 on Build GRCh38
86528 c.1310+1G>C p.? Chr.4: 69108323-69108323 on Build GRCh38
1056924 c.1112T>G p.F371C Chr.4: 69108124-69108124 on Build GRCh38
6448946 c.1290G>C p.K430N Chr.4: 69108302-69108302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182594363 Chr.4: 69107926-69107926 on Build GRCh38
rs191158058 Chr.4: 69107984-69107984 on Build GRCh38
rs745638449 Chr.4: 69108134-69108134 on Build GRCh38
rs200539544 Chr.4: 69108229-69108229 on Build GRCh38
rs545832745 Chr.4: 69108209-69108209 on Build GRCh38
rs138302870 Chr.4: 69108213-69108213 on Build GRCh38
rs114113997 Chr.4: 69108363-69108363 on Build GRCh38
rs768744149 Chr.4: 69108069-69108069 on Build GRCh38
rs780789575 Chr.4: 69108192-69108192 on Build GRCh38
rs142806461 Chr.4: 69108214-69108214 on Build GRCh38
rs749670128 Chr.4: 69108356-69108356 on Build GRCh38
rs921213070 Chr.4: 69108365-69108365 on Build GRCh38
rs536762972 Chr.4: 69107982-69107982 on Build GRCh38
rs753133394 Chr.4: 69108172-69108172 on Build GRCh38
rs762542364 Chr.4: 69108176-69108176 on Build GRCh38
rs560640435 Chr.4: 69107921-69107921 on Build GRCh38
rs543169852 Chr.4: 69108201-69108201 on Build GRCh38
rs547979452 Chr.4: 69108268-69108268 on Build GRCh38
rs760271390 Chr.4: 69108307-69108307 on Build GRCh38
rs528028669 Chr.4: 69108242-69108242 on Build GRCh38
rs571768346 Chr.4: 69107940-69107940 on Build GRCh38
rs10022440 Chr.4: 69108008-69108008 on Build GRCh38
rs369355939 Chr.4: 69108058-69108058 on Build GRCh38
rs369679234 Chr.4: 69108073-69108073 on Build GRCh38
rs748315310 Chr.4: 69108147-69108147 on Build GRCh38
rs769760607 Chr.4: 69108139-69108139 on Build GRCh38
rs139693571 Chr.4: 69108322-69108322 on Build GRCh38
rs1052342746 Chr.4: 69108034-69108034 on Build GRCh38
rs760768829 Chr.4: 69108155-69108155 on Build GRCh38
rs769107320 Chr.4: 69108277-69108277 on Build GRCh38
rs952696846 Chr.4: 69107933-69107933 on Build GRCh38
rs141309270 Chr.4: 69108286-69108286 on Build GRCh38
rs936360483 Chr.4: 69108014-69108014 on Build GRCh38
rs1008501089 Chr.4: 69107923-69107923 on Build GRCh38
rs952673980 Chr.4: 69108107-69108107 on Build GRCh38
rs774841677 Chr.4: 69108369-69108369 on Build GRCh38
rs752295609 Chr.4: 69108100-69108100 on Build GRCh38
rs747153786 Chr.4: 69108264-69108264 on Build GRCh38
rs754799236 Chr.4: 69108326-69108326 on Build GRCh38
rs746543685 Chr.4: 69107961-69107961 on Build GRCh38
rs753837836 Chr.4: 69108254-69108254 on Build GRCh38
rs1004652217 Chr.4: 69108108-69108108 on Build GRCh38
rs765914901 Chr.4: 69108166-69108166 on Build GRCh38
rs908812697 Chr.4: 69107948-69107948 on Build GRCh38
rs34308305 Chr.4: 69108238-69108238 on Build GRCh38
rs754920475 Chr.4: 69108255-69108255 on Build GRCh38
rs771240942 Chr.4: 69108097-69108097 on Build GRCh38
rs761112303 Chr.4: 69108086-69108086 on Build GRCh38
rs541946259 Chr.4: 69108012-69108012 on Build GRCh38
rs4588522 Chr.4: 69107949-69107949 on Build GRCh38
rs377264995 Chr.4: 69108353-69108353 on Build GRCh38
rs781384871 Chr.4: 69108341-69108341 on Build GRCh38
rs763316866 Chr.4: 69108228-69108228 on Build GRCh38
rs774459890 Chr.4: 69108071-69108071 on Build GRCh38
rs1000705906 Chr.4: 69107911-69107911 on Build GRCh38
rs770621742 Chr.4: 69108053-69108053 on Build GRCh38
rs61736964 Chr.4: 69108218-69108218 on Build GRCh38
rs150248109 Chr.4: 69108197-69108197 on Build GRCh38
rs775763044 Chr.4: 69108275-69108275 on Build GRCh38
rs892405684 Chr.4: 69108046-69108046 on Build GRCh38
rs34851008 Chr.4: 69108300-69108300 on Build GRCh38
rs757420843 Chr.4: 69108332-69108332 on Build GRCh38
rs572047837 Chr.4: 69108096-69108096 on Build GRCh38
rs765148488 Chr.4: 69108330-69108330 on Build GRCh38
rs756326976 Chr.4: 69108347-69108347 on Build GRCh38
rs200100430 Chr.4: 69108164-69108164 on Build GRCh38
rs78965261 Chr.4: 69107951-69107951 on Build GRCh38
rs746024489 Chr.4: 69108274-69108274 on Build GRCh38
rs751243157 Chr.4: 69108186-69108186 on Build GRCh38
rs745917758 Chr.4: 69108346-69108346 on Build GRCh38
rs1044801823 Chr.4: 69107996-69107996 on Build GRCh38
rs767214326 Chr.4: 69108240-69108240 on Build GRCh38
rs35327868 Chr.4: 69108149-69108149 on Build GRCh38
rs763449768 Chr.4: 69108184-69108184 on Build GRCh38
rs913522391 Chr.4: 69108352-69108352 on Build GRCh38
rs201964275 Chr.4: 69108222-69108222 on Build GRCh38
rs776991562 Chr.4: 69108204-69108204 on Build GRCh38
rs761324309 Chr.4: 69108280-69108280 on Build GRCh38
rs766047432 Chr.4: 69108253-69108253 on Build GRCh38
rs57913007 Chr.4: 69108203-69108203 on Build GRCh38
rs148274136 Chr.4: 69108270-69108270 on Build GRCh38
rs757071222 Chr.4: 69108127-69108127 on Build GRCh38
rs1052564131 Chr.4: 69108158-69108158 on Build GRCh38
rs774905521 Chr.4: 69108279-69108279 on Build GRCh38
rs61735558 Chr.4: 69108188-69108188 on Build GRCh38
rs185863914 Chr.4: 69107929-69107929 on Build GRCh38
rs749629452 Chr.4: 69108276-69108276 on Build GRCh38
rs753618139 Chr.4: 69108323-69108323 on Build GRCh38
rs563256432 Chr.4: 69108202-69108202 on Build GRCh38
rs373107309 Chr.4: 69108088-69108088 on Build GRCh38
rs767847311 Chr.4: 69108074-69108074 on Build GRCh38
rs796403790 Chr.4: 69107949-69107951 on Build GRCh38
rs151128457 Chr.4: 69108114-69108114 on Build GRCh38
rs780912290 Chr.4: 69108131-69108131 on Build GRCh38
rs35590824 Chr.4: 69108145-69108145 on Build GRCh38
rs372595294 Chr.4: 69107927-69107927 on Build GRCh38
rs4364327 Chr.4: 69107974-69107974 on Build GRCh38
rs755908336 Chr.4: 69108246-69108246 on Build GRCh38
rs757844749 Chr.4: 69108105-69108105 on Build GRCh38
rs773292514 Chr.4: 69108082-69108082 on Build GRCh38
rs750282415 Chr.4: 69108195-69108195 on Build GRCh38
rs144232904 Chr.4: 69108256-69108256 on Build GRCh38
rs983024831 Chr.4: 69107938-69107938 on Build GRCh38
rs771987274 Chr.4: 69108148-69108148 on Build GRCh38
rs993097202 Chr.4: 69108133-69108133 on Build GRCh38
rs58997816 Chr.4: 69107930-69107930 on Build GRCh38
rs771444554 Chr.4: 69108159-69108159 on Build GRCh38
rs751118884 Chr.4: 69108121-69108121 on Build GRCh38
rs374766685 Chr.4: 69108314-69108314 on Build GRCh38
rs138934444 Chr.4: 69108200-69108200 on Build GRCh38
rs758725646 Chr.4: 69108317-69108320 on Build GRCh38
rs960076700 Chr.4: 69108258-69108258 on Build GRCh38
rs4314345 Chr.4: 69107922-69107922 on Build GRCh38
rs34571475 Chr.4: 69108035-69108036 on Build GRCh38

View Full Product Details