Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 69480344-69480872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGT2B4
Entrez Gene ID
7363
Gene Name
UDP glucuronosyltransferase family 2 member B4
Gene Aliases
HLUG25, UDPGT2B4, UDPGTH1, UDPGTh-1, UGT2B11
UniGene
Hs.285887
Gene Chromosome Location
Chr.4: 69480165-69526014 on Build GRCh38
Transcripts
NM_001297615.1 NM_001297616.1 NM_021139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1430699 c.1406G>A p.R469H Chr.4: 69480815-69480815 on Build GRCh38
286285 c.1378G>A p.A460T Chr.4: 69480843-69480843 on Build GRCh38
305606 c.1573G>A p.G525R Chr.4: 69480648-69480648 on Build GRCh38
6100509 c.1386C>A p.F462L Chr.4: 69480835-69480835 on Build GRCh38
7094446 c.1530A>G p.K510K Chr.4: 69480691-69480691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs13119049 Chr.4: 69480847-69480847 on Build GRCh38
rs41298245 Chr.4: 69480690-69480690 on Build GRCh38
rs527905109 Chr.4: 69480739-69480739 on Build GRCh38
rs67904882 Chr.4: 69480846-69480847 on Build GRCh38
rs969576747 Chr.4: 69480504-69480504 on Build GRCh38
rs1004360903 Chr.4: 69480452-69480452 on Build GRCh38
rs574577022 Chr.4: 69480419-69480419 on Build GRCh38
rs968143526 Chr.4: 69480619-69480619 on Build GRCh38
rs767904806 Chr.4: 69480664-69480664 on Build GRCh38
rs762114586 Chr.4: 69480776-69480776 on Build GRCh38
rs748960146 Chr.4: 69480602-69480602 on Build GRCh38
rs1028919761 Chr.4: 69480496-69480496 on Build GRCh38
rs377673869 Chr.4: 69480675-69480675 on Build GRCh38
rs758118729 Chr.4: 69480740-69480740 on Build GRCh38
rs376460975 Chr.4: 69480378-69480378 on Build GRCh38
rs550152016 Chr.4: 69480836-69480836 on Build GRCh38
rs752000647 Chr.4: 69480656-69480656 on Build GRCh38
rs1008351318 Chr.4: 69480721-69480721 on Build GRCh38
rs1966151 Chr.4: 69480409-69480409 on Build GRCh38
rs772384251 Chr.4: 69480753-69480753 on Build GRCh38
rs1031022131 Chr.4: 69480384-69480384 on Build GRCh38
rs200280202 Chr.4: 69480762-69480762 on Build GRCh38
rs367718795 Chr.4: 69480439-69480439 on Build GRCh38
rs375773104 Chr.4: 69480596-69480596 on Build GRCh38
rs770843089 Chr.4: 69480747-69480747 on Build GRCh38
rs751569355 Chr.4: 69480572-69480574 on Build GRCh38
rs771821155 Chr.4: 69480627-69480627 on Build GRCh38
rs867758927 Chr.4: 69480642-69480642 on Build GRCh38
rs764240029 Chr.4: 69480764-69480764 on Build GRCh38
rs367620350 Chr.4: 69480841-69480841 on Build GRCh38
rs767837637 Chr.4: 69480777-69480777 on Build GRCh38
rs760647912 Chr.4: 69480629-69480629 on Build GRCh38
rs757739776 Chr.4: 69480662-69480662 on Build GRCh38
rs768231044 Chr.4: 69480610-69480610 on Build GRCh38
rs1043998951 Chr.4: 69480814-69480814 on Build GRCh38
rs901641166 Chr.4: 69480749-69480750 on Build GRCh38
rs200320318 Chr.4: 69480816-69480816 on Build GRCh38
rs745780826 Chr.4: 69480835-69480835 on Build GRCh38
rs771911170 Chr.4: 69480720-69480720 on Build GRCh38
rs1028890192 Chr.4: 69480782-69480782 on Build GRCh38
rs750830787 Chr.4: 69480393-69480393 on Build GRCh38
rs370143836 Chr.4: 69480616-69480616 on Build GRCh38
rs1035598068 Chr.4: 69480542-69480542 on Build GRCh38
rs770803752 Chr.4: 69480829-69480829 on Build GRCh38
rs748927770 Chr.4: 69480667-69480668 on Build GRCh38
rs773071003 Chr.4: 69480628-69480628 on Build GRCh38
rs373908353 Chr.4: 69480649-69480649 on Build GRCh38
rs41298247 Chr.4: 69480502-69480502 on Build GRCh38
rs951662136 Chr.4: 69480499-69480499 on Build GRCh38
rs951964641 Chr.4: 69480791-69480791 on Build GRCh38
rs774765310 Chr.4: 69480775-69480775 on Build GRCh38
rs368535155 Chr.4: 69480827-69480827 on Build GRCh38
rs1016713751 Chr.4: 69480455-69480455 on Build GRCh38
rs762512901 Chr.4: 69480511-69480511 on Build GRCh38
rs778378649 Chr.4: 69480792-69480792 on Build GRCh38
rs879463290 Chr.4: 69480846-69480847 on Build GRCh38
rs1018812686 Chr.4: 69480454-69480454 on Build GRCh38
rs760028353 Chr.4: 69480850-69480850 on Build GRCh38
rs547579888 Chr.4: 69480778-69480778 on Build GRCh38
rs763278148 Chr.4: 69480641-69480641 on Build GRCh38
rs781751752 Chr.4: 69480586-69480586 on Build GRCh38
rs752751624 Chr.4: 69480640-69480640 on Build GRCh38
rs756644070 Chr.4: 69480678-69480678 on Build GRCh38
rs748186522 Chr.4: 69480566-69480566 on Build GRCh38
rs746357168 Chr.4: 69480591-69480591 on Build GRCh38
rs747860734 Chr.4: 69480807-69480807 on Build GRCh38
rs201317023 Chr.4: 69480719-69480719 on Build GRCh38
rs904133685 Chr.4: 69480623-69480623 on Build GRCh38
rs1051054701 Chr.4: 69480648-69480648 on Build GRCh38
rs761549102 Chr.4: 69480624-69480624 on Build GRCh38
rs756596732 Chr.4: 69480779-69480779 on Build GRCh38
rs567459900 Chr.4: 69480787-69480787 on Build GRCh38
rs778681610 Chr.4: 69480526-69480526 on Build GRCh38
rs769240107 Chr.4: 69480594-69480594 on Build GRCh38
rs563092551 Chr.4: 69480555-69480555 on Build GRCh38
rs773055119 Chr.4: 69480729-69480729 on Build GRCh38
rs374388378 Chr.4: 69480840-69480840 on Build GRCh38
rs1005885167 Chr.4: 69480798-69480798 on Build GRCh38
rs376320686 Chr.4: 69480812-69480812 on Build GRCh38
rs754650230 Chr.4: 69480714-69480714 on Build GRCh38
rs753918506 Chr.4: 69480491-69480491 on Build GRCh38
rs13142440 Chr.4: 69480846-69480846 on Build GRCh38
rs755153405 Chr.4: 69480508-69480508 on Build GRCh38
rs892573766 Chr.4: 69480386-69480386 on Build GRCh38
rs898147687 Chr.4: 69480472-69480472 on Build GRCh38
rs776668037 Chr.4: 69480832-69480832 on Build GRCh38
rs766863857 Chr.4: 69480781-69480781 on Build GRCh38
rs966353707 Chr.4: 69480485-69480485 on Build GRCh38
rs192265916 Chr.4: 69480717-69480717 on Build GRCh38
rs748775080 Chr.4: 69480696-69480696 on Build GRCh38
rs532055799 Chr.4: 69480638-69480638 on Build GRCh38

View Full Product Details