Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 69589476-69590005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGT2A1
Entrez Gene ID
10941
Gene Name
UDP glucuronosyltransferase family 2 member A1 complex locus
Gene Aliases
UDPGT2A1
UniGene
Hs.225950
Gene Chromosome Location
Chr.4: 69588417-69653249 on Build GRCh38
Transcripts
NM_001252274.2 NM_001252275.2 NM_001301239.1 NM_006798.4 NM_001105677.2 NM_001301233.1

Gene Symbol
UGT2A2
Entrez Gene ID
574537
Gene Name
UDP glucuronosyltransferase family 2 member A2
Gene Aliases
-
UniGene
Hs.225950
Gene Chromosome Location
Chr.4: 69588417-69639642 on Build GRCh38
Transcripts
NM_001252274.2 NM_001252275.2 NM_001301239.1 NM_006798.4 NM_001105677.2 NM_001301233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3605325 c.1396C>T p.R466* Chr.4: 69589587-69589587 on Build GRCh38
2829784 c.1331C>G p.S444* Chr.4: 69589625-69589625 on Build GRCh38
4961144 c.1435G>A p.G479R Chr.4: 69589548-69589548 on Build GRCh38
3605328 c.1388C>T p.P463L Chr.4: 69589595-69589595 on Build GRCh38
5032187 c.1367A>G p.H456R Chr.4: 69589616-69589616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022618696 Chr.4: 69589903-69589903 on Build GRCh38
rs760575332 Chr.4: 69589519-69589519 on Build GRCh38
rs767856867 Chr.4: 69589557-69589557 on Build GRCh38
rs922324482 Chr.4: 69589693-69589693 on Build GRCh38
rs375760530 Chr.4: 69589691-69589691 on Build GRCh38
rs776556764 Chr.4: 69589573-69589573 on Build GRCh38
rs748837836 Chr.4: 69589507-69589507 on Build GRCh38
rs781529026 Chr.4: 69589592-69589593 on Build GRCh38
rs1039025626 Chr.4: 69589789-69589789 on Build GRCh38
rs923568531 Chr.4: 69589717-69589717 on Build GRCh38
rs990636185 Chr.4: 69589675-69589675 on Build GRCh38
rs893368905 Chr.4: 69589770-69589770 on Build GRCh38
rs113968234 Chr.4: 69589946-69589946 on Build GRCh38
rs966768654 Chr.4: 69589671-69589671 on Build GRCh38
rs772703720 Chr.4: 69589568-69589568 on Build GRCh38
rs201031939 Chr.4: 69589581-69589581 on Build GRCh38
rs760157773 Chr.4: 69589701-69589701 on Build GRCh38
rs768231232 Chr.4: 69589508-69589508 on Build GRCh38
rs557241555 Chr.4: 69589885-69589885 on Build GRCh38
rs897189379 Chr.4: 69589794-69589794 on Build GRCh38
rs541720302 Chr.4: 69589562-69589562 on Build GRCh38
rs1016213330 Chr.4: 69589877-69589877 on Build GRCh38
rs757489284 Chr.4: 69589549-69589549 on Build GRCh38
rs137925293 Chr.4: 69589892-69589892 on Build GRCh38
rs41292305 Chr.4: 69589513-69589513 on Build GRCh38
rs767821027 Chr.4: 69589550-69589550 on Build GRCh38
rs777499588 Chr.4: 69589635-69589635 on Build GRCh38
rs139303872 Chr.4: 69589587-69589587 on Build GRCh38
rs766782549 Chr.4: 69589596-69589596 on Build GRCh38
rs149562826 Chr.4: 69589590-69589590 on Build GRCh38
rs771987229 Chr.4: 69589666-69589666 on Build GRCh38
rs914839308 Chr.4: 69589962-69589962 on Build GRCh38
rs936356020 Chr.4: 69589943-69589943 on Build GRCh38
rs374546120 Chr.4: 69589680-69589680 on Build GRCh38
rs774329824 Chr.4: 69589847-69589847 on Build GRCh38
rs747808890 Chr.4: 69589566-69589566 on Build GRCh38
rs546585927 Chr.4: 69589714-69589714 on Build GRCh38
rs929954716 Chr.4: 69589820-69589820 on Build GRCh38
rs111934444 Chr.4: 69589857-69589857 on Build GRCh38
rs1029303964 Chr.4: 69589958-69589958 on Build GRCh38
rs959283062 Chr.4: 69589586-69589586 on Build GRCh38
rs377162913 Chr.4: 69589729-69589729 on Build GRCh38
rs1039148544 Chr.4: 69589798-69589798 on Build GRCh38
rs760973572 Chr.4: 69589683-69589683 on Build GRCh38
rs767812077 Chr.4: 69589589-69589589 on Build GRCh38
rs778611239 Chr.4: 69589616-69589616 on Build GRCh38
rs4148322 Chr.4: 69589692-69589692 on Build GRCh38
rs756512694 Chr.4: 69589594-69589594 on Build GRCh38
rs1026709013 Chr.4: 69589608-69589608 on Build GRCh38
rs764218838 Chr.4: 69589577-69589577 on Build GRCh38
rs759466309 Chr.4: 69589575-69589575 on Build GRCh38
rs764097677 Chr.4: 69589539-69589539 on Build GRCh38
rs1042074422 Chr.4: 69589689-69589689 on Build GRCh38
rs959265091 Chr.4: 69589880-69589880 on Build GRCh38
rs769063071 Chr.4: 69589552-69589553 on Build GRCh38
rs67327460 Chr.4: 69589530-69589530 on Build GRCh38
rs757801646 Chr.4: 69589630-69589630 on Build GRCh38
rs745857048 Chr.4: 69589651-69589651 on Build GRCh38
rs2288740 Chr.4: 69589861-69589861 on Build GRCh38
rs145385081 Chr.4: 69589570-69589570 on Build GRCh38
rs769687875 Chr.4: 69589654-69589654 on Build GRCh38
rs997969362 Chr.4: 69589843-69589843 on Build GRCh38
rs553370248 Chr.4: 69589942-69589942 on Build GRCh38
rs370288222 Chr.4: 69589891-69589891 on Build GRCh38
rs770232779 Chr.4: 69589691-69589691 on Build GRCh38
rs1438537 Chr.4: 69589932-69589932 on Build GRCh38
rs750770233 Chr.4: 69589551-69589551 on Build GRCh38
rs755526328 Chr.4: 69589615-69589615 on Build GRCh38
rs774158000 Chr.4: 69589512-69589512 on Build GRCh38
rs2288741 Chr.4: 69589760-69589760 on Build GRCh38
rs1045839245 Chr.4: 69589830-69589830 on Build GRCh38
rs746847554 Chr.4: 69589569-69589569 on Build GRCh38
rs766375229 Chr.4: 69589520-69589520 on Build GRCh38
rs761974484 Chr.4: 69589664-69589664 on Build GRCh38
rs756392602 Chr.4: 69589553-69589553 on Build GRCh38
rs141510881 Chr.4: 69589535-69589535 on Build GRCh38
rs936340637 Chr.4: 69589758-69589758 on Build GRCh38
rs771825937 Chr.4: 69589516-69589516 on Build GRCh38
rs901953106 Chr.4: 69589842-69589842 on Build GRCh38
rs570549689 Chr.4: 69589642-69589642 on Build GRCh38
rs771730135 Chr.4: 69589567-69589567 on Build GRCh38
rs754459546 Chr.4: 69589599-69589599 on Build GRCh38
rs746578636 Chr.4: 69589639-69589639 on Build GRCh38
rs532842168 Chr.4: 69589686-69589686 on Build GRCh38
rs955135621 Chr.4: 69589961-69589961 on Build GRCh38
rs188420216 Chr.4: 69589882-69589882 on Build GRCh38
rs545889676 Chr.4: 69589881-69589881 on Build GRCh38
rs761485544 Chr.4: 69589578-69589578 on Build GRCh38
rs775251542 Chr.4: 69589662-69589662 on Build GRCh38
rs371649258 Chr.4: 69589618-69589618 on Build GRCh38
rs1007710677 Chr.4: 69589796-69589796 on Build GRCh38
rs150861593 Chr.4: 69589556-69589556 on Build GRCh38
rs199765285 Chr.4: 69589548-69589548 on Build GRCh38
rs748662922 Chr.4: 69589661-69589664 on Build GRCh38
rs994301586 Chr.4: 69589537-69589537 on Build GRCh38
rs562684415 Chr.4: 69589883-69589883 on Build GRCh38
rs780953199 Chr.4: 69589648-69589648 on Build GRCh38
rs959232605 Chr.4: 69589674-69589674 on Build GRCh38

View Full Product Details