Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70031956-70032468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTN3
Entrez Gene ID
3347
Gene Name
histatin 3
Gene Aliases
HIS2, HTN2, HTN5
UniGene
Hs.654442
Gene Chromosome Location
Chr.4: 70028413-70036538 on Build GRCh38
Transcripts
NM_000200.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3780328 c.52G>C p.G18R Chr.4: 70031979-70031979 on Build GRCh38
387458 c.102+1G>T p.? Chr.4: 70032108-70032108 on Build GRCh38
109097 c.62C>T p.S21L Chr.4: 70031989-70031989 on Build GRCh38
1254486 c.53G>T p.G18V Chr.4: 70031980-70031980 on Build GRCh38
4691442 c.75A>C p.R25S Chr.4: 70032080-70032080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751461581 Chr.4: 70032090-70032090 on Build GRCh38
rs115176279 Chr.4: 70032123-70032123 on Build GRCh38
rs557360175 Chr.4: 70032098-70032098 on Build GRCh38
rs775471285 Chr.4: 70032146-70032146 on Build GRCh38
rs991605133 Chr.4: 70032368-70032368 on Build GRCh38
rs947609351 Chr.4: 70032161-70032161 on Build GRCh38
rs1041944820 Chr.4: 70032408-70032408 on Build GRCh38
rs879028867 Chr.4: 70032117-70032117 on Build GRCh38
rs764955288 Chr.4: 70032141-70032141 on Build GRCh38
rs375236308 Chr.4: 70032031-70032031 on Build GRCh38
rs1007304974 Chr.4: 70032216-70032216 on Build GRCh38
rs903442472 Chr.4: 70032419-70032419 on Build GRCh38
rs772295668 Chr.4: 70032153-70032156 on Build GRCh38
rs534081851 Chr.4: 70032187-70032187 on Build GRCh38
rs375542985 Chr.4: 70032222-70032222 on Build GRCh38
rs997605206 Chr.4: 70032185-70032185 on Build GRCh38
rs947556103 Chr.4: 70032389-70032389 on Build GRCh38
rs796416049 Chr.4: 70032383-70032383 on Build GRCh38
rs184638013 Chr.4: 70032023-70032023 on Build GRCh38
rs776561503 Chr.4: 70032288-70032288 on Build GRCh38
rs116486629 Chr.4: 70032234-70032234 on Build GRCh38
rs759331701 Chr.4: 70032130-70032130 on Build GRCh38
rs762860733 Chr.4: 70032157-70032157 on Build GRCh38
rs28491522 Chr.4: 70032104-70032104 on Build GRCh38
rs78169960 Chr.4: 70032094-70032094 on Build GRCh38
rs770364282 Chr.4: 70032127-70032127 on Build GRCh38
rs745712748 Chr.4: 70032294-70032294 on Build GRCh38
rs551658548 Chr.4: 70032356-70032356 on Build GRCh38
rs186993306 Chr.4: 70032428-70032428 on Build GRCh38
rs200920872 Chr.4: 70032063-70032063 on Build GRCh38
rs888869630 Chr.4: 70032212-70032212 on Build GRCh38
rs145531373 Chr.4: 70032004-70032004 on Build GRCh38
rs182481150 Chr.4: 70032353-70032353 on Build GRCh38
rs765186297 Chr.4: 70032044-70032044 on Build GRCh38
rs772562109 Chr.4: 70032021-70032021 on Build GRCh38
rs561798967 Chr.4: 70032298-70032298 on Build GRCh38
rs997257922 Chr.4: 70031995-70031995 on Build GRCh38
rs776195819 Chr.4: 70032128-70032128 on Build GRCh38
rs773623441 Chr.4: 70032118-70032118 on Build GRCh38
rs762666112 Chr.4: 70032339-70032339 on Build GRCh38
rs537063184 Chr.4: 70032392-70032392 on Build GRCh38
rs1055974842 Chr.4: 70032126-70032126 on Build GRCh38
rs547652131 Chr.4: 70032039-70032039 on Build GRCh38
rs749483616 Chr.4: 70031985-70031985 on Build GRCh38
rs569689246 Chr.4: 70032377-70032377 on Build GRCh38
rs199755562 Chr.4: 70032085-70032085 on Build GRCh38
rs747062376 Chr.4: 70032155-70032155 on Build GRCh38
rs774641106 Chr.4: 70031994-70031994 on Build GRCh38
rs748491364 Chr.4: 70032100-70032100 on Build GRCh38
rs779168727 Chr.4: 70032040-70032041 on Build GRCh38
rs189858152 Chr.4: 70032334-70032334 on Build GRCh38
rs780158080 Chr.4: 70031984-70031984 on Build GRCh38
rs111345058 Chr.4: 70032229-70032229 on Build GRCh38
rs902826543 Chr.4: 70032172-70032172 on Build GRCh38
rs768850611 Chr.4: 70032096-70032096 on Build GRCh38
rs56388012 Chr.4: 70032204-70032204 on Build GRCh38
rs113589683 Chr.4: 70032297-70032297 on Build GRCh38
rs1042386225 Chr.4: 70032168-70032168 on Build GRCh38
rs565957559 Chr.4: 70032338-70032338 on Build GRCh38
rs779174509 Chr.4: 70032091-70032091 on Build GRCh38
rs539815223 Chr.4: 70032064-70032064 on Build GRCh38
rs151219648 Chr.4: 70031989-70031989 on Build GRCh38
rs777827633 Chr.4: 70032077-70032077 on Build GRCh38
rs769012258 Chr.4: 70031992-70031992 on Build GRCh38
rs971990617 Chr.4: 70032089-70032089 on Build GRCh38
rs17147981 Chr.4: 70032310-70032310 on Build GRCh38
rs138043613 Chr.4: 70032199-70032199 on Build GRCh38
rs557909178 Chr.4: 70032092-70032092 on Build GRCh38
rs140992381 Chr.4: 70032263-70032263 on Build GRCh38
rs754168370 Chr.4: 70032037-70032037 on Build GRCh38
rs763803800 Chr.4: 70032346-70032346 on Build GRCh38
rs755304884 Chr.4: 70032058-70032058 on Build GRCh38
rs759211037 Chr.4: 70032042-70032042 on Build GRCh38
rs113261238 Chr.4: 70032033-70032033 on Build GRCh38
rs776120310 Chr.4: 70032158-70032161 on Build GRCh38
rs563879003 Chr.4: 70032277-70032277 on Build GRCh38
rs746205585 Chr.4: 70032112-70032113 on Build GRCh38

View Full Product Details