Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70825198-70825654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRSF1
Entrez Gene ID
2926
Gene Name
G-rich RNA sequence binding factor 1
Gene Aliases
-
UniGene
Hs.309763
Gene Chromosome Location
Chr.4: 70815782-70843274 on Build GRCh38
Transcripts
NM_001098477.1 NM_002092.3 XM_005265681.3 XM_011531897.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4940647 c.1384T>C p.S462P Chr.4: 70825305-70825305 on Build GRCh38
5984855 c.1382G>A p.R461Q Chr.4: 70825307-70825307 on Build GRCh38
1619027 c.1293G>A p.M431I Chr.4: 70825396-70825396 on Build GRCh38
6335538 c.1276C>A p.P426T Chr.4: 70825413-70825413 on Build GRCh38
6031162 c.1349A>G p.H450R Chr.4: 70825340-70825340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532657086 Chr.4: 70825237-70825237 on Build GRCh38
rs780190725 Chr.4: 70825295-70825295 on Build GRCh38
rs377631575 Chr.4: 70825300-70825300 on Build GRCh38
rs374217828 Chr.4: 70825368-70825368 on Build GRCh38
rs1019847555 Chr.4: 70825348-70825348 on Build GRCh38
rs980299382 Chr.4: 70825621-70825621 on Build GRCh38
rs368308484 Chr.4: 70825454-70825454 on Build GRCh38
rs961569221 Chr.4: 70825596-70825596 on Build GRCh38
rs761791649 Chr.4: 70825494-70825494 on Build GRCh38
rs751771246 Chr.4: 70825581-70825581 on Build GRCh38
rs386675830 Chr.4: 70825565-70825566 on Build GRCh38
rs773565301 Chr.4: 70825307-70825307 on Build GRCh38
rs565287098 Chr.4: 70825228-70825228 on Build GRCh38
rs748297375 Chr.4: 70825366-70825366 on Build GRCh38
rs1041622996 Chr.4: 70825560-70825560 on Build GRCh38
rs770059272 Chr.4: 70825224-70825224 on Build GRCh38
rs931352432 Chr.4: 70825416-70825416 on Build GRCh38
rs767232395 Chr.4: 70825439-70825439 on Build GRCh38
rs200822254 Chr.4: 70825537-70825537 on Build GRCh38
rs765294838 Chr.4: 70825530-70825530 on Build GRCh38
rs772360700 Chr.4: 70825304-70825304 on Build GRCh38
rs549084693 Chr.4: 70825463-70825463 on Build GRCh38
rs183345000 Chr.4: 70825568-70825568 on Build GRCh38
rs952688475 Chr.4: 70825602-70825602 on Build GRCh38
rs373247711 Chr.4: 70825299-70825299 on Build GRCh38
rs973308032 Chr.4: 70825598-70825598 on Build GRCh38
rs6843245 Chr.4: 70825565-70825565 on Build GRCh38
rs775508165 Chr.4: 70825322-70825322 on Build GRCh38
rs755031031 Chr.4: 70825350-70825350 on Build GRCh38
rs200367912 Chr.4: 70825336-70825336 on Build GRCh38
rs766585930 Chr.4: 70825343-70825343 on Build GRCh38
rs774187379 Chr.4: 70825435-70825435 on Build GRCh38
rs755351964 Chr.4: 70825609-70825609 on Build GRCh38
rs938798935 Chr.4: 70825470-70825470 on Build GRCh38
rs769361024 Chr.4: 70825417-70825417 on Build GRCh38
rs1008382777 Chr.4: 70825541-70825541 on Build GRCh38
rs535101121 Chr.4: 70825460-70825460 on Build GRCh38
rs952951717 Chr.4: 70825618-70825618 on Build GRCh38
rs903766935 Chr.4: 70825540-70825540 on Build GRCh38
rs1019745991 Chr.4: 70825592-70825592 on Build GRCh38
rs745518563 Chr.4: 70825415-70825415 on Build GRCh38
rs776184513 Chr.4: 70825315-70825315 on Build GRCh38
rs948662406 Chr.4: 70825490-70825490 on Build GRCh38
rs755046329 Chr.4: 70825298-70825298 on Build GRCh38
rs973016020 Chr.4: 70825384-70825384 on Build GRCh38
rs534597893 Chr.4: 70825539-70825539 on Build GRCh38
rs6813986 Chr.4: 70825566-70825566 on Build GRCh38
rs761622831 Chr.4: 70825437-70825437 on Build GRCh38
rs768684523 Chr.4: 70825434-70825434 on Build GRCh38
rs374437339 Chr.4: 70825321-70825321 on Build GRCh38
rs533668542 Chr.4: 70825450-70825450 on Build GRCh38
rs998566844 Chr.4: 70825335-70825335 on Build GRCh38
rs915800049 Chr.4: 70825483-70825483 on Build GRCh38
rs756353284 Chr.4: 70825282-70825282 on Build GRCh38
rs374413394 Chr.4: 70825420-70825420 on Build GRCh38
rs747387977 Chr.4: 70825310-70825310 on Build GRCh38
rs1057455839 Chr.4: 70825473-70825473 on Build GRCh38
rs759754289 Chr.4: 70825457-70825457 on Build GRCh38
rs750461303 Chr.4: 70825549-70825549 on Build GRCh38
rs370109008 Chr.4: 70825319-70825319 on Build GRCh38
rs904207332 Chr.4: 70825520-70825520 on Build GRCh38
rs953761072 Chr.4: 70825362-70825362 on Build GRCh38
rs200027347 Chr.4: 70825439-70825442 on Build GRCh38
rs762608883 Chr.4: 70825427-70825427 on Build GRCh38
rs757043739 Chr.4: 70825341-70825341 on Build GRCh38
rs376559778 Chr.4: 70825372-70825372 on Build GRCh38
rs1042369129 Chr.4: 70825533-70825533 on Build GRCh38
rs763994545 Chr.4: 70825260-70825260 on Build GRCh38
rs370663108 Chr.4: 70825393-70825393 on Build GRCh38
rs903524881 Chr.4: 70825329-70825329 on Build GRCh38
rs186328559 Chr.4: 70825375-70825375 on Build GRCh38
rs896285186 Chr.4: 70825308-70825308 on Build GRCh38
rs776295787 Chr.4: 70825394-70825394 on Build GRCh38
rs758508726 Chr.4: 70825472-70825472 on Build GRCh38
rs964674991 Chr.4: 70825371-70825371 on Build GRCh38
rs900041879 Chr.4: 70825562-70825562 on Build GRCh38
rs778971404 Chr.4: 70825365-70825365 on Build GRCh38
rs770591540 Chr.4: 70825314-70825314 on Build GRCh38
rs192040893 Chr.4: 70825465-70825465 on Build GRCh38
rs201134200 Chr.4: 70825327-70825327 on Build GRCh38
rs754031422 Chr.4: 70825344-70825344 on Build GRCh38
rs749356964 Chr.4: 70825296-70825296 on Build GRCh38
rs934363543 Chr.4: 70825522-70825522 on Build GRCh38
rs1026989462 Chr.4: 70825386-70825386 on Build GRCh38
rs199887449 Chr.4: 70825340-70825340 on Build GRCh38
rs775135425 Chr.4: 70825422-70825422 on Build GRCh38

View Full Product Details