Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70976219-70976716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOB1B
Entrez Gene ID
92597
Gene Name
MOB kinase activator 1B
Gene Aliases
MATS2, MOB4A, MOBKL1A
UniGene
Hs.691454
Gene Chromosome Location
Chr.4: 70901970-70988174 on Build GRCh38
Transcripts
NM_001244766.1 NM_001244767.1 NM_173468.3 XM_005265709.2 XM_011532412.2 XM_017008837.1

SNP Detail

SNP ID Chromosome Location
rs770320033 Chr.4: 70976339-70976339 on Build GRCh38
rs930087003 Chr.4: 70976414-70976414 on Build GRCh38
rs114607241 Chr.4: 70976593-70976593 on Build GRCh38
rs907072903 Chr.4: 70976468-70976469 on Build GRCh38
rs574686951 Chr.4: 70976620-70976620 on Build GRCh38
rs978236489 Chr.4: 70976290-70976290 on Build GRCh38
rs944763569 Chr.4: 70976405-70976405 on Build GRCh38
rs184166587 Chr.4: 70976371-70976371 on Build GRCh38
rs1018501366 Chr.4: 70976608-70976608 on Build GRCh38
rs914476986 Chr.4: 70976688-70976688 on Build GRCh38
rs763688813 Chr.4: 70976571-70976571 on Build GRCh38
rs550406983 Chr.4: 70976562-70976562 on Build GRCh38
rs893053703 Chr.4: 70976512-70976512 on Build GRCh38
rs570030732 Chr.4: 70976603-70976603 on Build GRCh38
rs1019984799 Chr.4: 70976526-70976526 on Build GRCh38
rs1055679413 Chr.4: 70976510-70976510 on Build GRCh38
rs188300899 Chr.4: 70976442-70976442 on Build GRCh38
rs538437744 Chr.4: 70976516-70976516 on Build GRCh38
rs532358551 Chr.4: 70976254-70976254 on Build GRCh38
rs1036460941 Chr.4: 70976411-70976411 on Build GRCh38
rs564747129 Chr.4: 70976325-70976325 on Build GRCh38
rs548995143 Chr.4: 70976682-70976683 on Build GRCh38
rs777011686 Chr.4: 70976271-70976271 on Build GRCh38
rs974028493 Chr.4: 70976624-70976624 on Build GRCh38
rs955529058 Chr.4: 70976598-70976598 on Build GRCh38
rs1027694509 Chr.4: 70976465-70976465 on Build GRCh38
rs911466413 Chr.4: 70976376-70976376 on Build GRCh38
rs766812451 Chr.4: 70976428-70976428 on Build GRCh38
rs773849627 Chr.4: 70976340-70976340 on Build GRCh38
rs981450896 Chr.4: 70976641-70976641 on Build GRCh38
rs191364030 Chr.4: 70976261-70976261 on Build GRCh38
rs955806964 Chr.4: 70976335-70976335 on Build GRCh38
rs868544374 Chr.4: 70976623-70976623 on Build GRCh38
rs763576054 Chr.4: 70976391-70976391 on Build GRCh38
rs928717396 Chr.4: 70976645-70976647 on Build GRCh38
rs781298842 Chr.4: 70976281-70976281 on Build GRCh38
rs937172695 Chr.4: 70976475-70976475 on Build GRCh38
rs548699512 Chr.4: 70976305-70976305 on Build GRCh38
rs533363860 Chr.4: 70976503-70976503 on Build GRCh38
rs139882446 Chr.4: 70976286-70976286 on Build GRCh38
rs556442978 Chr.4: 70976540-70976540 on Build GRCh38
rs978428158 Chr.4: 70976269-70976269 on Build GRCh38
rs186747930 Chr.4: 70976257-70976257 on Build GRCh38
rs542340538 Chr.4: 70976646-70976646 on Build GRCh38
rs974059913 Chr.4: 70976412-70976412 on Build GRCh38
rs951461833 Chr.4: 70976632-70976632 on Build GRCh38
rs890630433 Chr.4: 70976387-70976387 on Build GRCh38
rs918618371 Chr.4: 70976625-70976625 on Build GRCh38
rs1045530483 Chr.4: 70976452-70976452 on Build GRCh38
rs985770372 Chr.4: 70976357-70976357 on Build GRCh38
rs145865418 Chr.4: 70976687-70976687 on Build GRCh38
rs1001468968 Chr.4: 70976519-70976519 on Build GRCh38
rs149840362 Chr.4: 70976413-70976413 on Build GRCh38
rs937159403 Chr.4: 70976674-70976674 on Build GRCh38
rs997302221 Chr.4: 70976590-70976590 on Build GRCh38
rs201518216 Chr.4: 70976622-70976623 on Build GRCh38
rs757016107 Chr.4: 70976678-70976678 on Build GRCh38
rs1020015945 Chr.4: 70976276-70976276 on Build GRCh38

View Full Product Details