Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87490618-87491132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPARCL1
Entrez Gene ID
8404
Gene Name
SPARC like 1
Gene Aliases
MAST 9, MAST9, PIG33, SC1
UniGene
Hs.62886
Gene Chromosome Location
Chr.4: 87473330-87529503 on Build GRCh38
Transcripts
NM_001128310.2 NM_001291976.1 NM_001291977.1 NM_004684.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5030759 c.1409_1410+2delAAGT p.? Chr.4: 87490758-87490761 on Build GRCh38
2957090 c.1337C>T p.A446V Chr.4: 87490833-87490833 on Build GRCh38
6788028 c.1323C>G p.G441G Chr.4: 87490847-87490847 on Build GRCh38
2957091 c.1323C>T p.G441G Chr.4: 87490847-87490847 on Build GRCh38
4126576 c.1387C>A p.P463T Chr.4: 87490783-87490783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779374096 Chr.4: 87490762-87490762 on Build GRCh38
rs752120364 Chr.4: 87490905-87490905 on Build GRCh38
rs966176742 Chr.4: 87490698-87490698 on Build GRCh38
rs532593633 Chr.4: 87490660-87490660 on Build GRCh38
rs930553725 Chr.4: 87490944-87490944 on Build GRCh38
rs202053780 Chr.4: 87490999-87490999 on Build GRCh38
rs900905778 Chr.4: 87491085-87491085 on Build GRCh38
rs909186225 Chr.4: 87490668-87490668 on Build GRCh38
rs754412496 Chr.4: 87490787-87490788 on Build GRCh38
rs200212509 Chr.4: 87490798-87490798 on Build GRCh38
rs547879623 Chr.4: 87490921-87490921 on Build GRCh38
rs747450487 Chr.4: 87490815-87490815 on Build GRCh38
rs757768682 Chr.4: 87490923-87490923 on Build GRCh38
rs1055961684 Chr.4: 87490779-87490779 on Build GRCh38
rs968277119 Chr.4: 87491099-87491100 on Build GRCh38
rs112902367 Chr.4: 87490880-87490880 on Build GRCh38
rs552144509 Chr.4: 87490744-87490744 on Build GRCh38
rs756828791 Chr.4: 87490709-87490709 on Build GRCh38
rs940693818 Chr.4: 87490690-87490690 on Build GRCh38
rs761826368 Chr.4: 87490845-87490845 on Build GRCh38
rs112577256 Chr.4: 87490870-87490870 on Build GRCh38
rs781291932 Chr.4: 87490914-87490914 on Build GRCh38
rs1019010936 Chr.4: 87490996-87490996 on Build GRCh38
rs1007582768 Chr.4: 87490970-87490970 on Build GRCh38
rs765969060 Chr.4: 87490891-87490891 on Build GRCh38
rs1028073817 Chr.4: 87490653-87490653 on Build GRCh38
rs754230942 Chr.4: 87490967-87490967 on Build GRCh38
rs751863084 Chr.4: 87490956-87490956 on Build GRCh38
rs894673170 Chr.4: 87490839-87490839 on Build GRCh38
rs776681189 Chr.4: 87490828-87490828 on Build GRCh38
rs1043342296 Chr.4: 87490950-87490950 on Build GRCh38
rs1021183439 Chr.4: 87490720-87490720 on Build GRCh38
rs769873953 Chr.4: 87490926-87490926 on Build GRCh38
rs979745828 Chr.4: 87490789-87490789 on Build GRCh38
rs779189091 Chr.4: 87490735-87490736 on Build GRCh38
rs1033961799 Chr.4: 87491101-87491101 on Build GRCh38
rs762683766 Chr.4: 87490835-87490835 on Build GRCh38
rs980077357 Chr.4: 87491099-87491099 on Build GRCh38
rs748438717 Chr.4: 87490777-87490777 on Build GRCh38
rs755838913 Chr.4: 87490885-87490885 on Build GRCh38
rs761282479 Chr.4: 87490707-87490707 on Build GRCh38
rs897702435 Chr.4: 87490927-87490927 on Build GRCh38
rs757759703 Chr.4: 87490906-87490906 on Build GRCh38
rs376446939 Chr.4: 87490749-87490749 on Build GRCh38
rs185948869 Chr.4: 87490652-87490652 on Build GRCh38
rs755715073 Chr.4: 87490752-87490752 on Build GRCh38
rs559197818 Chr.4: 87490792-87490792 on Build GRCh38
rs984021642 Chr.4: 87490663-87490663 on Build GRCh38
rs778055578 Chr.4: 87490900-87490900 on Build GRCh38
rs373292481 Chr.4: 87490869-87490869 on Build GRCh38
rs775408684 Chr.4: 87490834-87490834 on Build GRCh38
rs749867653 Chr.4: 87490874-87490874 on Build GRCh38
rs746170845 Chr.4: 87490832-87490832 on Build GRCh38
rs189143813 Chr.4: 87490841-87490841 on Build GRCh38
rs569923541 Chr.4: 87490964-87490964 on Build GRCh38
rs771013611 Chr.4: 87490822-87490822 on Build GRCh38
rs547450567 Chr.4: 87490700-87490700 on Build GRCh38
rs749834935 Chr.4: 87490893-87490893 on Build GRCh38
rs1040748858 Chr.4: 87491060-87491060 on Build GRCh38
rs372337679 Chr.4: 87490713-87490713 on Build GRCh38
rs569046146 Chr.4: 87491037-87491037 on Build GRCh38
rs368683010 Chr.4: 87490894-87490894 on Build GRCh38
rs901935379 Chr.4: 87491007-87491007 on Build GRCh38
rs764214465 Chr.4: 87490892-87490892 on Build GRCh38
rs760388448 Chr.4: 87490896-87490896 on Build GRCh38

View Full Product Details