Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87838503-87839029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEPE
Entrez Gene ID
56955
Gene Name
matrix extracellular phosphoglycoprotein
Gene Aliases
OF45
UniGene
Hs.676357
Gene Chromosome Location
Chr.4: 87821398-87846817 on Build GRCh38
Transcripts
NM_001184694.2 NM_001184695.1 NM_001184696.1 NM_001184697.1 NM_001291183.1 NM_020203.3 XM_006714278.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1431637 c.107G>A p.R36K Chr.4: 87838684-87838684 on Build GRCh38
4821282 c.89G>C p.S30T Chr.4: 87838666-87838666 on Build GRCh38
353301 c.94G>T p.V32L Chr.4: 87838671-87838671 on Build GRCh38
5912108 c.55-8C>T p.? Chr.4: 87838624-87838624 on Build GRCh38
1058681 c.58T>G p.F20V Chr.4: 87838635-87838635 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749138295 Chr.4: 87838608-87838608 on Build GRCh38
rs542505071 Chr.4: 87838859-87838859 on Build GRCh38
rs544689618 Chr.4: 87838961-87838961 on Build GRCh38
rs952212916 Chr.4: 87838710-87838710 on Build GRCh38
rs555770848 Chr.4: 87838756-87838756 on Build GRCh38
rs1008029249 Chr.4: 87838884-87838884 on Build GRCh38
rs533503256 Chr.4: 87838858-87838858 on Build GRCh38
rs148421186 Chr.4: 87838547-87838547 on Build GRCh38
rs756331103 Chr.4: 87838602-87838602 on Build GRCh38
rs771993657 Chr.4: 87838623-87838623 on Build GRCh38
rs567061562 Chr.4: 87838728-87838728 on Build GRCh38
rs754762635 Chr.4: 87838615-87838615 on Build GRCh38
rs142563589 Chr.4: 87838801-87838801 on Build GRCh38
rs996159689 Chr.4: 87838947-87838947 on Build GRCh38
rs760230951 Chr.4: 87838738-87838741 on Build GRCh38
rs758320935 Chr.4: 87838729-87838729 on Build GRCh38
rs371833970 Chr.4: 87838574-87838574 on Build GRCh38
rs1005352781 Chr.4: 87838776-87838776 on Build GRCh38
rs983525494 Chr.4: 87838748-87838748 on Build GRCh38
rs1054104982 Chr.4: 87838806-87838806 on Build GRCh38
rs745824409 Chr.4: 87838692-87838693 on Build GRCh38
rs983943511 Chr.4: 87838957-87838957 on Build GRCh38
rs1030059922 Chr.4: 87838886-87838886 on Build GRCh38
rs771978563 Chr.4: 87838721-87838722 on Build GRCh38
rs775298238 Chr.4: 87838775-87838776 on Build GRCh38
rs780226197 Chr.4: 87838605-87838605 on Build GRCh38
rs760252046 Chr.4: 87838631-87838631 on Build GRCh38
rs372276204 Chr.4: 87838649-87838649 on Build GRCh38
rs952054581 Chr.4: 87838953-87838953 on Build GRCh38
rs778609861 Chr.4: 87838616-87838616 on Build GRCh38
rs140694518 Chr.4: 87838936-87838936 on Build GRCh38
rs562886172 Chr.4: 87838963-87838963 on Build GRCh38
rs1027623504 Chr.4: 87838952-87838952 on Build GRCh38
rs764568886 Chr.4: 87838659-87838659 on Build GRCh38
rs955207527 Chr.4: 87838542-87838542 on Build GRCh38
rs1010416019 Chr.4: 87838822-87838822 on Build GRCh38
rs759200790 Chr.4: 87838655-87838655 on Build GRCh38
rs6835632 Chr.4: 87838538-87838538 on Build GRCh38
rs756204102 Chr.4: 87838711-87838711 on Build GRCh38
rs778636178 Chr.4: 87838688-87838688 on Build GRCh38
rs150920278 Chr.4: 87838808-87838808 on Build GRCh38
rs1031722149 Chr.4: 87838938-87838938 on Build GRCh38
rs150608883 Chr.4: 87838642-87838642 on Build GRCh38
rs555535372 Chr.4: 87838679-87838679 on Build GRCh38
rs561047257 Chr.4: 87838909-87838909 on Build GRCh38
rs990253704 Chr.4: 87838955-87838955 on Build GRCh38
rs375754391 Chr.4: 87838708-87838708 on Build GRCh38
rs201903368 Chr.4: 87838619-87838619 on Build GRCh38
rs1018183740 Chr.4: 87838890-87838890 on Build GRCh38
rs567060467 Chr.4: 87838629-87838629 on Build GRCh38
rs754061470 Chr.4: 87838723-87838723 on Build GRCh38
rs937029623 Chr.4: 87838803-87838803 on Build GRCh38
rs1046020951 Chr.4: 87838791-87838791 on Build GRCh38
rs955524259 Chr.4: 87838918-87838918 on Build GRCh38
rs534082565 Chr.4: 87838668-87838668 on Build GRCh38
rs56358523 Chr.4: 87838837-87838837 on Build GRCh38
rs775025501 Chr.4: 87838724-87838725 on Build GRCh38
rs545031162 Chr.4: 87838750-87838750 on Build GRCh38
rs577540034 Chr.4: 87838778-87838778 on Build GRCh38
rs746510246 Chr.4: 87838638-87838638 on Build GRCh38
rs963930017 Chr.4: 87838804-87838804 on Build GRCh38
rs768037642 Chr.4: 87838699-87838699 on Build GRCh38
rs367865276 Chr.4: 87838716-87838716 on Build GRCh38

View Full Product Details