Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87839848-87840345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEPE
Entrez Gene ID
56955
Gene Name
matrix extracellular phosphoglycoprotein
Gene Aliases
OF45
UniGene
Hs.676357
Gene Chromosome Location
Chr.4: 87821398-87846817 on Build GRCh38
Transcripts
NM_001184694.2 NM_001184695.1 NM_001184696.1 NM_001184697.1 NM_001291183.1 NM_020203.3 XM_006714278.2

SNP Detail

SNP ID Chromosome Location
rs942666201 Chr.4: 87839916-87839916 on Build GRCh38
rs1027953615 Chr.4: 87840149-87840149 on Build GRCh38
rs894596200 Chr.4: 87840011-87840011 on Build GRCh38
rs939825205 Chr.4: 87839922-87839922 on Build GRCh38
rs912394289 Chr.4: 87840055-87840055 on Build GRCh38
rs561163908 Chr.4: 87839998-87839998 on Build GRCh38
rs753862162 Chr.4: 87839958-87839958 on Build GRCh38
rs879075132 Chr.4: 87840016-87840016 on Build GRCh38
rs765411380 Chr.4: 87840000-87840000 on Build GRCh38
rs1002958041 Chr.4: 87840123-87840123 on Build GRCh38
rs757495462 Chr.4: 87840267-87840267 on Build GRCh38
rs776254551 Chr.4: 87839880-87839880 on Build GRCh38
rs1039942131 Chr.4: 87839920-87839920 on Build GRCh38
rs1055798921 Chr.4: 87839995-87839995 on Build GRCh38
rs903249529 Chr.4: 87840314-87840314 on Build GRCh38
rs189208748 Chr.4: 87840126-87840126 on Build GRCh38
rs370425004 Chr.4: 87840166-87840166 on Build GRCh38
rs982332985 Chr.4: 87840183-87840183 on Build GRCh38
rs759781070 Chr.4: 87839985-87839985 on Build GRCh38
rs373685284 Chr.4: 87840167-87840167 on Build GRCh38
rs773763046 Chr.4: 87839898-87839898 on Build GRCh38
rs934886369 Chr.4: 87840319-87840319 on Build GRCh38
rs751531374 Chr.4: 87840046-87840046 on Build GRCh38
rs765768846 Chr.4: 87840006-87840006 on Build GRCh38
rs1044364037 Chr.4: 87840109-87840109 on Build GRCh38
rs1005130392 Chr.4: 87839978-87839978 on Build GRCh38
rs927851266 Chr.4: 87839957-87839957 on Build GRCh38
rs72655787 Chr.4: 87840117-87840117 on Build GRCh38
rs749596340 Chr.4: 87840164-87840164 on Build GRCh38
rs745341843 Chr.4: 87840073-87840073 on Build GRCh38
rs952442826 Chr.4: 87840162-87840162 on Build GRCh38
rs374074669 Chr.4: 87840178-87840178 on Build GRCh38
rs1027970459 Chr.4: 87840284-87840284 on Build GRCh38
rs1052971926 Chr.4: 87840071-87840071 on Build GRCh38
rs937901651 Chr.4: 87840261-87840261 on Build GRCh38
rs549240621 Chr.4: 87839993-87839993 on Build GRCh38
rs903337618 Chr.4: 87840207-87840207 on Build GRCh38
rs72875497 Chr.4: 87840041-87840041 on Build GRCh38
rs58720415 Chr.4: 87840264-87840264 on Build GRCh38
rs758212941 Chr.4: 87840019-87840019 on Build GRCh38
rs1011667108 Chr.4: 87840035-87840035 on Build GRCh38
rs927903718 Chr.4: 87840196-87840196 on Build GRCh38
rs184435358 Chr.4: 87840062-87840062 on Build GRCh38
rs372621738 Chr.4: 87839918-87839918 on Build GRCh38
rs538718633 Chr.4: 87840102-87840102 on Build GRCh38
rs13130999 Chr.4: 87840107-87840107 on Build GRCh38
rs766457667 Chr.4: 87839950-87839950 on Build GRCh38
rs1016491818 Chr.4: 87839984-87839984 on Build GRCh38
rs753202610 Chr.4: 87840051-87840051 on Build GRCh38
rs1043178210 Chr.4: 87840307-87840307 on Build GRCh38
rs901468620 Chr.4: 87839933-87839933 on Build GRCh38
rs759005068 Chr.4: 87839994-87839994 on Build GRCh38
rs996503607 Chr.4: 87840144-87840144 on Build GRCh38
rs947348481 Chr.4: 87840275-87840275 on Build GRCh38
rs1000366041 Chr.4: 87840315-87840315 on Build GRCh38
rs984243025 Chr.4: 87839907-87839907 on Build GRCh38
rs752497505 Chr.4: 87840012-87840012 on Build GRCh38
rs549468956 Chr.4: 87840050-87840050 on Build GRCh38

View Full Product Details