Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88466967-88467468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC5
Entrez Gene ID
51191
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 5
Gene Aliases
CEB1, CEBP1
UniGene
Hs.26663
Gene Chromosome Location
Chr.4: 88456604-88506170 on Build GRCh38
Transcripts
NM_016323.3 XM_011532022.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5640303 c.1037C>G p.S346* Chr.4: 88467184-88467184 on Build GRCh38
334613 c.1016C>A p.P339Q Chr.4: 88467163-88467163 on Build GRCh38
4126657 c.1034C>T p.S345L Chr.4: 88467181-88467181 on Build GRCh38
311756 c.1030G>A p.V344I Chr.4: 88467177-88467177 on Build GRCh38
4126655 c.1017G>A p.P339P Chr.4: 88467164-88467164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005262121 Chr.4: 88467438-88467438 on Build GRCh38
rs756633895 Chr.4: 88467242-88467242 on Build GRCh38
rs748836362 Chr.4: 88467220-88467220 on Build GRCh38
rs933085789 Chr.4: 88467311-88467311 on Build GRCh38
rs779445657 Chr.4: 88466998-88466998 on Build GRCh38
rs761240422 Chr.4: 88467234-88467234 on Build GRCh38
rs778130580 Chr.4: 88467240-88467240 on Build GRCh38
rs776435312 Chr.4: 88467034-88467034 on Build GRCh38
rs770639048 Chr.4: 88467020-88467020 on Build GRCh38
rs778841083 Chr.4: 88467164-88467164 on Build GRCh38
rs997073052 Chr.4: 88467217-88467217 on Build GRCh38
rs758719606 Chr.4: 88467210-88467210 on Build GRCh38
rs370062005 Chr.4: 88467176-88467176 on Build GRCh38
rs886826039 Chr.4: 88467434-88467435 on Build GRCh38
rs73841944 Chr.4: 88467417-88467417 on Build GRCh38
rs767921967 Chr.4: 88467057-88467058 on Build GRCh38
rs374210638 Chr.4: 88467207-88467207 on Build GRCh38
rs781199302 Chr.4: 88467200-88467200 on Build GRCh38
rs753832149 Chr.4: 88467245-88467245 on Build GRCh38
rs942938702 Chr.4: 88467381-88467381 on Build GRCh38
rs754059043 Chr.4: 88467148-88467148 on Build GRCh38
rs560206017 Chr.4: 88467392-88467392 on Build GRCh38
rs762177633 Chr.4: 88467097-88467097 on Build GRCh38
rs759319386 Chr.4: 88467061-88467061 on Build GRCh38
rs915077930 Chr.4: 88467391-88467391 on Build GRCh38
rs773698058 Chr.4: 88467224-88467224 on Build GRCh38
rs147757779 Chr.4: 88467147-88467147 on Build GRCh38
rs571675016 Chr.4: 88467011-88467011 on Build GRCh38
rs777767660 Chr.4: 88467180-88467180 on Build GRCh38
rs186000202 Chr.4: 88467004-88467004 on Build GRCh38
rs777379231 Chr.4: 88467010-88467010 on Build GRCh38
rs774824003 Chr.4: 88467213-88467213 on Build GRCh38
rs753052752 Chr.4: 88467207-88467207 on Build GRCh38
rs956557663 Chr.4: 88467321-88467321 on Build GRCh38
rs577596736 Chr.4: 88467175-88467175 on Build GRCh38
rs762547084 Chr.4: 88467014-88467016 on Build GRCh38
rs1046805634 Chr.4: 88467434-88467434 on Build GRCh38
rs141001025 Chr.4: 88467182-88467182 on Build GRCh38
rs752839723 Chr.4: 88467174-88467174 on Build GRCh38
rs1045034329 Chr.4: 88467400-88467400 on Build GRCh38
rs955586705 Chr.4: 88467287-88467287 on Build GRCh38
rs201521313 Chr.4: 88467198-88467198 on Build GRCh38
rs772403500 Chr.4: 88467223-88467223 on Build GRCh38
rs906207184 Chr.4: 88467133-88467133 on Build GRCh38
rs547568005 Chr.4: 88467073-88467073 on Build GRCh38
rs911433335 Chr.4: 88467366-88467366 on Build GRCh38
rs1044711442 Chr.4: 88467068-88467068 on Build GRCh38
rs544753299 Chr.4: 88467295-88467295 on Build GRCh38
rs1025687876 Chr.4: 88467103-88467103 on Build GRCh38
rs144890220 Chr.4: 88467204-88467204 on Build GRCh38
rs1023450998 Chr.4: 88467177-88467177 on Build GRCh38
rs750720998 Chr.4: 88467134-88467134 on Build GRCh38
rs760632109 Chr.4: 88467019-88467019 on Build GRCh38
rs867414801 Chr.4: 88467162-88467162 on Build GRCh38
rs887281764 Chr.4: 88467170-88467170 on Build GRCh38
rs755237879 Chr.4: 88467163-88467163 on Build GRCh38
rs746828656 Chr.4: 88467014-88467014 on Build GRCh38
rs761043953 Chr.4: 88467137-88467137 on Build GRCh38
rs201630041 Chr.4: 88467050-88467050 on Build GRCh38
rs755135564 Chr.4: 88467406-88467406 on Build GRCh38
rs977514178 Chr.4: 88467316-88467316 on Build GRCh38
rs772091890 Chr.4: 88467008-88467008 on Build GRCh38
rs775105692 Chr.4: 88467093-88467093 on Build GRCh38
rs200721883 Chr.4: 88467202-88467202 on Build GRCh38
rs768101045 Chr.4: 88467117-88467117 on Build GRCh38
rs747206462 Chr.4: 88467296-88467296 on Build GRCh38
rs988233208 Chr.4: 88467282-88467282 on Build GRCh38
rs1051247258 Chr.4: 88467365-88467365 on Build GRCh38

View Full Product Details