Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99881619-99882128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMTOR3
Entrez Gene ID
8649
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Gene Aliases
MAP2K1IP1, MAPBP, MAPKSP1, MP1, PRO0633, Ragulator3
UniGene
Hs.433332
Gene Chromosome Location
Chr.4: 99878338-99894546 on Build GRCh38
Transcripts
NM_001243736.1 NM_021970.3 NR_024170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6394372 c.343G>T p.E115* Chr.4: 99882026-99882026 on Build GRCh38
3945814 c.371C>A p.S124Y Chr.4: 99881998-99881998 on Build GRCh38
6864990 c.366A>C p.E122D Chr.4: 99882003-99882003 on Build GRCh38
355665 c.314G>T p.S105I Chr.4: 99882055-99882055 on Build GRCh38
5623659 c.336A>G p.P112P Chr.4: 99882033-99882033 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750050276 Chr.4: 99882091-99882091 on Build GRCh38
rs984452668 Chr.4: 99881827-99881827 on Build GRCh38
rs1020164081 Chr.4: 99881728-99881728 on Build GRCh38
rs764582961 Chr.4: 99881968-99881969 on Build GRCh38
rs778168254 Chr.4: 99881959-99881959 on Build GRCh38
rs769136255 Chr.4: 99881908-99881908 on Build GRCh38
rs191940376 Chr.4: 99881929-99881929 on Build GRCh38
rs890756760 Chr.4: 99881732-99881732 on Build GRCh38
rs937297686 Chr.4: 99882033-99882033 on Build GRCh38
rs934523058 Chr.4: 99881966-99881966 on Build GRCh38
rs544393327 Chr.4: 99881821-99881821 on Build GRCh38
rs1049685149 Chr.4: 99882083-99882083 on Build GRCh38
rs1018501218 Chr.4: 99881795-99881795 on Build GRCh38
rs928432376 Chr.4: 99881915-99881915 on Build GRCh38
rs775223542 Chr.4: 99881748-99881748 on Build GRCh38
rs952964129 Chr.4: 99881787-99881787 on Build GRCh38
rs371667534 Chr.4: 99881973-99881973 on Build GRCh38
rs775405019 Chr.4: 99882008-99882008 on Build GRCh38
rs142936049 Chr.4: 99881852-99881852 on Build GRCh38
rs917999116 Chr.4: 99881898-99881898 on Build GRCh38
rs772545504 Chr.4: 99881957-99881957 on Build GRCh38
rs576991769 Chr.4: 99881811-99881811 on Build GRCh38
rs956768853 Chr.4: 99881894-99881894 on Build GRCh38
rs551313105 Chr.4: 99881869-99881869 on Build GRCh38
rs993910452 Chr.4: 99881662-99881662 on Build GRCh38
rs1025820855 Chr.4: 99881717-99881717 on Build GRCh38
rs558801768 Chr.4: 99881739-99881739 on Build GRCh38
rs755690193 Chr.4: 99882104-99882104 on Build GRCh38
rs973368358 Chr.4: 99881740-99881740 on Build GRCh38
rs779317864 Chr.4: 99881947-99881947 on Build GRCh38
rs1045486623 Chr.4: 99881874-99881874 on Build GRCh38
rs988209695 Chr.4: 99881895-99881895 on Build GRCh38
rs776856839 Chr.4: 99881974-99881974 on Build GRCh38
rs34321222 Chr.4: 99881742-99881743 on Build GRCh38
rs763036197 Chr.4: 99882038-99882038 on Build GRCh38
rs377184402 Chr.4: 99881985-99881985 on Build GRCh38
rs767095415 Chr.4: 99882079-99882079 on Build GRCh38
rs745958845 Chr.4: 99881976-99881976 on Build GRCh38
rs747370548 Chr.4: 99881969-99881969 on Build GRCh38
rs774289012 Chr.4: 99882049-99882049 on Build GRCh38
rs778476302 Chr.4: 99881992-99881995 on Build GRCh38
rs920163031 Chr.4: 99881779-99881779 on Build GRCh38
rs1038157535 Chr.4: 99882037-99882037 on Build GRCh38
rs35008249 Chr.4: 99881966-99881966 on Build GRCh38
rs927194698 Chr.4: 99882030-99882030 on Build GRCh38
rs748691947 Chr.4: 99881949-99881949 on Build GRCh38
rs764117490 Chr.4: 99882044-99882044 on Build GRCh38
rs898298061 Chr.4: 99882041-99882041 on Build GRCh38
rs964319828 Chr.4: 99881809-99881809 on Build GRCh38
rs560205572 Chr.4: 99882056-99882056 on Build GRCh38
rs893054645 Chr.4: 99882053-99882053 on Build GRCh38

View Full Product Details