Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105437858-105438357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPA2
Entrez Gene ID
27068
Gene Name
pyrophosphatase (inorganic) 2
Gene Aliases
HSPC124, SID6-306
UniGene
Hs.654957
Gene Chromosome Location
Chr.4: 105369077-105474070 on Build GRCh38
Transcripts
NM_006903.4 NM_176866.2 NM_176867.3 NM_176869.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4862347 c.553G>A p.V185I Chr.4: 105437970-105437970 on Build GRCh38
4864890 c.559G>A p.E187K Chr.4: 105437964-105437964 on Build GRCh38
4970868 c.454C>A p.P152T Chr.4: 105438024-105438024 on Build GRCh38
7284780 c.442-7delA p.? Chr.4: 105438043-105438043 on Build GRCh38
732044 c.508G>A p.V170I Chr.4: 105437970-105437970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745682289 Chr.4: 105438040-105438040 on Build GRCh38
rs922854941 Chr.4: 105438027-105438027 on Build GRCh38
rs1017120064 Chr.4: 105438198-105438198 on Build GRCh38
rs770680674 Chr.4: 105437933-105437933 on Build GRCh38
rs760083632 Chr.4: 105438083-105438083 on Build GRCh38
rs200123475 Chr.4: 105437999-105437999 on Build GRCh38
rs756455141 Chr.4: 105438051-105438052 on Build GRCh38
rs761751672 Chr.4: 105438262-105438262 on Build GRCh38
rs1026097549 Chr.4: 105438188-105438188 on Build GRCh38
rs181437213 Chr.4: 105437931-105437931 on Build GRCh38
rs758961996 Chr.4: 105437909-105437909 on Build GRCh38
rs376523591 Chr.4: 105438206-105438206 on Build GRCh38
rs765643758 Chr.4: 105437996-105437996 on Build GRCh38
rs994131560 Chr.4: 105437934-105437934 on Build GRCh38
rs763546588 Chr.4: 105438085-105438085 on Build GRCh38
rs764567038 Chr.4: 105438052-105438053 on Build GRCh38
rs891860768 Chr.4: 105438117-105438117 on Build GRCh38
rs980284813 Chr.4: 105438012-105438012 on Build GRCh38
rs918313820 Chr.4: 105438042-105438042 on Build GRCh38
rs201990956 Chr.4: 105438066-105438066 on Build GRCh38
rs1001757095 Chr.4: 105438180-105438180 on Build GRCh38
rs1029831443 Chr.4: 105438261-105438261 on Build GRCh38
rs143660369 Chr.4: 105438082-105438082 on Build GRCh38
rs866393040 Chr.4: 105437971-105437971 on Build GRCh38
rs201209617 Chr.4: 105438069-105438069 on Build GRCh38
rs371351245 Chr.4: 105437929-105437929 on Build GRCh38
rs1043314885 Chr.4: 105437901-105437901 on Build GRCh38
rs753176486 Chr.4: 105438042-105438043 on Build GRCh38
rs199503096 Chr.4: 105437915-105437915 on Build GRCh38
rs765766495 Chr.4: 105437902-105437902 on Build GRCh38
rs546693824 Chr.4: 105437978-105437978 on Build GRCh38
rs186403998 Chr.4: 105438086-105438086 on Build GRCh38
rs139651138 Chr.4: 105438001-105438001 on Build GRCh38
rs147396330 Chr.4: 105437922-105437922 on Build GRCh38
rs774181831 Chr.4: 105437940-105437940 on Build GRCh38
rs750869104 Chr.4: 105437907-105437907 on Build GRCh38
rs201997865 Chr.4: 105437920-105437920 on Build GRCh38
rs994732040 Chr.4: 105438229-105438229 on Build GRCh38
rs575938866 Chr.4: 105438200-105438200 on Build GRCh38
rs778534602 Chr.4: 105438035-105438035 on Build GRCh38
rs752062224 Chr.4: 105438002-105438002 on Build GRCh38
rs375410953 Chr.4: 105437919-105437919 on Build GRCh38
rs755844015 Chr.4: 105438005-105438005 on Build GRCh38
rs7659961 Chr.4: 105438235-105438235 on Build GRCh38
rs112923507 Chr.4: 105438285-105438285 on Build GRCh38
rs762171869 Chr.4: 105437985-105437985 on Build GRCh38
rs745745034 Chr.4: 105437961-105437961 on Build GRCh38
rs762422555 Chr.4: 105437925-105437928 on Build GRCh38
rs35079223 Chr.4: 105438270-105438271 on Build GRCh38
rs115990263 Chr.4: 105438071-105438071 on Build GRCh38
rs1024587484 Chr.4: 105437977-105437977 on Build GRCh38
rs575289677 Chr.4: 105438053-105438053 on Build GRCh38
rs2713862 Chr.4: 105438052-105438052 on Build GRCh38
rs764329827 Chr.4: 105437976-105437976 on Build GRCh38
rs979360950 Chr.4: 105438274-105438274 on Build GRCh38
rs986526757 Chr.4: 105438237-105438237 on Build GRCh38
rs551515553 Chr.4: 105438051-105438051 on Build GRCh38
rs146013446 Chr.4: 105437964-105437964 on Build GRCh38
rs763393976 Chr.4: 105438153-105438153 on Build GRCh38
rs1032144565 Chr.4: 105437949-105437949 on Build GRCh38
rs760824971 Chr.4: 105437975-105437975 on Build GRCh38
rs962824637 Chr.4: 105438028-105438028 on Build GRCh38
rs756925958 Chr.4: 105438014-105438014 on Build GRCh38
rs545971431 Chr.4: 105438310-105438310 on Build GRCh38
rs994039690 Chr.4: 105438181-105438181 on Build GRCh38
rs768039900 Chr.4: 105438043-105438043 on Build GRCh38
rs988397421 Chr.4: 105438303-105438303 on Build GRCh38
rs766718377 Chr.4: 105438078-105438078 on Build GRCh38
rs538394501 Chr.4: 105438109-105438109 on Build GRCh38
rs866826492 Chr.4: 105438156-105438156 on Build GRCh38
rs1018371527 Chr.4: 105438219-105438219 on Build GRCh38
rs2713863 Chr.4: 105438236-105438236 on Build GRCh38
rs540577921 Chr.4: 105438335-105438335 on Build GRCh38
rs759504578 Chr.4: 105438321-105438321 on Build GRCh38
rs559192615 Chr.4: 105438119-105438119 on Build GRCh38
rs771850189 Chr.4: 105438047-105438047 on Build GRCh38
rs138826533 Chr.4: 105437965-105437965 on Build GRCh38
rs750827864 Chr.4: 105437997-105437997 on Build GRCh38
rs150745595 Chr.4: 105438013-105438013 on Build GRCh38
rs895776865 Chr.4: 105438167-105438167 on Build GRCh38
rs1046150174 Chr.4: 105438120-105438120 on Build GRCh38
rs910897767 Chr.4: 105438251-105438251 on Build GRCh38

View Full Product Details