Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109712426-109712972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G12A
Entrez Gene ID
81579
Gene Name
phospholipase A2 group XIIA
Gene Aliases
GXII, PLA2G12, ROSSY
UniGene
Hs.389452
Gene Chromosome Location
Chr.4: 109709989-109730086 on Build GRCh38
Transcripts
NM_030821.4

SNP Detail

SNP ID Chromosome Location
rs966952100 Chr.4: 109712616-109712616 on Build GRCh38
rs958428061 Chr.4: 109712796-109712796 on Build GRCh38
rs1000179770 Chr.4: 109712775-109712775 on Build GRCh38
rs957056493 Chr.4: 109712607-109712607 on Build GRCh38
rs971572458 Chr.4: 109712648-109712649 on Build GRCh38
rs76268496 Chr.4: 109712476-109712476 on Build GRCh38
rs928510602 Chr.4: 109712707-109712707 on Build GRCh38
rs751766422 Chr.4: 109712669-109712669 on Build GRCh38
rs902741259 Chr.4: 109712771-109712771 on Build GRCh38
rs192093469 Chr.4: 109712759-109712759 on Build GRCh38
rs113235454 Chr.4: 109712509-109712510 on Build GRCh38
rs540508236 Chr.4: 109712646-109712646 on Build GRCh38
rs1021561354 Chr.4: 109712482-109712482 on Build GRCh38
rs939933168 Chr.4: 109712720-109712720 on Build GRCh38
rs1037288716 Chr.4: 109712455-109712455 on Build GRCh38
rs995671086 Chr.4: 109712474-109712474 on Build GRCh38
rs895412841 Chr.4: 109712820-109712820 on Build GRCh38
rs529798285 Chr.4: 109712657-109712657 on Build GRCh38
rs1050204077 Chr.4: 109712705-109712705 on Build GRCh38
rs559788871 Chr.4: 109712742-109712742 on Build GRCh38
rs183502177 Chr.4: 109712895-109712895 on Build GRCh38
rs761348437 Chr.4: 109712938-109712938 on Build GRCh38
rs889433251 Chr.4: 109712538-109712538 on Build GRCh38
rs552282510 Chr.4: 109712825-109712825 on Build GRCh38
rs575638346 Chr.4: 109712584-109712584 on Build GRCh38
rs781618249 Chr.4: 109712704-109712704 on Build GRCh38
rs1036830352 Chr.4: 109712726-109712726 on Build GRCh38
rs113898153 Chr.4: 109712457-109712457 on Build GRCh38
rs775913208 Chr.4: 109712887-109712887 on Build GRCh38
rs991632135 Chr.4: 109712630-109712630 on Build GRCh38
rs188519742 Chr.4: 109712655-109712655 on Build GRCh38
rs1019244821 Chr.4: 109712767-109712767 on Build GRCh38
rs183525566 Chr.4: 109712637-109712637 on Build GRCh38
rs991030114 Chr.4: 109712810-109712810 on Build GRCh38
rs747232386 Chr.4: 109712553-109712553 on Build GRCh38
rs568751068 Chr.4: 109712847-109712847 on Build GRCh38
rs889546306 Chr.4: 109712745-109712745 on Build GRCh38
rs919908512 Chr.4: 109712698-109712698 on Build GRCh38
rs1032932750 Chr.4: 109712645-109712645 on Build GRCh38
rs996932615 Chr.4: 109712451-109712451 on Build GRCh38
rs1049457295 Chr.4: 109712743-109712743 on Build GRCh38
rs531382827 Chr.4: 109712885-109712885 on Build GRCh38
rs568000690 Chr.4: 109712519-109712520 on Build GRCh38
rs187860533 Chr.4: 109712922-109712922 on Build GRCh38
rs971136933 Chr.4: 109712873-109712873 on Build GRCh38
rs1042528004 Chr.4: 109712749-109712749 on Build GRCh38
rs1032584223 Chr.4: 109712795-109712795 on Build GRCh38
rs907883390 Chr.4: 109712926-109712926 on Build GRCh38
rs1038335580 Chr.4: 109712694-109712694 on Build GRCh38
rs1001082008 Chr.4: 109712510-109712510 on Build GRCh38
rs1032636201 Chr.4: 109712776-109712776 on Build GRCh38
rs898313672 Chr.4: 109712727-109712727 on Build GRCh38
rs1019797868 Chr.4: 109712606-109712606 on Build GRCh38
rs577804544 Chr.4: 109712653-109712653 on Build GRCh38
rs373688542 Chr.4: 109712791-109712791 on Build GRCh38
rs557604677 Chr.4: 109712851-109712851 on Build GRCh38
rs565063581 Chr.4: 109712676-109712676 on Build GRCh38
rs757375174 Chr.4: 109712741-109712741 on Build GRCh38

View Full Product Details