Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109713284-109713793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G12A
Entrez Gene ID
81579
Gene Name
phospholipase A2 group XIIA
Gene Aliases
GXII, PLA2G12, ROSSY
UniGene
Hs.389452
Gene Chromosome Location
Chr.4: 109709989-109730086 on Build GRCh38
Transcripts
NM_030821.4

SNP Detail

SNP ID Chromosome Location
rs866664055 Chr.4: 109713743-109713743 on Build GRCh38
rs184692241 Chr.4: 109713699-109713699 on Build GRCh38
rs559898614 Chr.4: 109713617-109713617 on Build GRCh38
rs1036718071 Chr.4: 109713600-109713600 on Build GRCh38
rs138261217 Chr.4: 109713524-109713524 on Build GRCh38
rs758456542 Chr.4: 109713520-109713520 on Build GRCh38
rs978022831 Chr.4: 109713703-109713703 on Build GRCh38
rs149653102 Chr.4: 109713673-109713673 on Build GRCh38
rs924002634 Chr.4: 109713371-109713371 on Build GRCh38
rs919814193 Chr.4: 109713347-109713347 on Build GRCh38
rs74788225 Chr.4: 109713764-109713764 on Build GRCh38
rs1053472093 Chr.4: 109713607-109713607 on Build GRCh38
rs895336359 Chr.4: 109713465-109713465 on Build GRCh38
rs562604630 Chr.4: 109713698-109713698 on Build GRCh38
rs192849379 Chr.4: 109713693-109713693 on Build GRCh38
rs896869656 Chr.4: 109713605-109713605 on Build GRCh38
rs1053847403 Chr.4: 109713452-109713452 on Build GRCh38
rs557248911 Chr.4: 109713686-109713686 on Build GRCh38
rs911049604 Chr.4: 109713384-109713384 on Build GRCh38
rs952663143 Chr.4: 109713359-109713359 on Build GRCh38
rs189673181 Chr.4: 109713570-109713570 on Build GRCh38
rs567930322 Chr.4: 109713736-109713736 on Build GRCh38
rs746239136 Chr.4: 109713769-109713769 on Build GRCh38
rs995265771 Chr.4: 109713632-109713632 on Build GRCh38
rs544603553 Chr.4: 109713336-109713336 on Build GRCh38
rs373192958 Chr.4: 109713668-109713668 on Build GRCh38
rs954056557 Chr.4: 109713656-109713656 on Build GRCh38
rs774242102 Chr.4: 109713751-109713753 on Build GRCh38
rs962341173 Chr.4: 109713322-109713322 on Build GRCh38
rs936759556 Chr.4: 109713390-109713390 on Build GRCh38
rs752633996 Chr.4: 109713507-109713507 on Build GRCh38
rs943914505 Chr.4: 109713503-109713503 on Build GRCh38
rs1026812675 Chr.4: 109713649-109713649 on Build GRCh38
rs2285713 Chr.4: 109713735-109713735 on Build GRCh38
rs765296482 Chr.4: 109713367-109713367 on Build GRCh38
rs944064291 Chr.4: 109713313-109713313 on Build GRCh38
rs540165639 Chr.4: 109713577-109713577 on Build GRCh38
rs894007133 Chr.4: 109713626-109713626 on Build GRCh38
rs533866174 Chr.4: 109713472-109713473 on Build GRCh38
rs985342519 Chr.4: 109713768-109713768 on Build GRCh38
rs1027665476 Chr.4: 109713760-109713760 on Build GRCh38
rs923950136 Chr.4: 109713727-109713727 on Build GRCh38

View Full Product Details