Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109871300-109871848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRIT3
Entrez Gene ID
345193
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 3
Gene Aliases
CSNB1F, FIGLER4
UniGene
Hs.308127
Gene Chromosome Location
Chr.4: 109848182-109872315 on Build GRCh38
Transcripts
NM_198506.4

SNP Detail

SNP ID Chromosome Location
rs1036225712 Chr.4: 109871401-109871401 on Build GRCh38
rs372231507 Chr.4: 109871713-109871713 on Build GRCh38
rs989173141 Chr.4: 109871639-109871639 on Build GRCh38
rs1025182716 Chr.4: 109871394-109871394 on Build GRCh38
rs4698798 Chr.4: 109871473-109871473 on Build GRCh38
rs920526153 Chr.4: 109871697-109871697 on Build GRCh38
rs886710351 Chr.4: 109871658-109871658 on Build GRCh38
rs999135632 Chr.4: 109871576-109871576 on Build GRCh38
rs191289678 Chr.4: 109871771-109871771 on Build GRCh38
rs986684138 Chr.4: 109871539-109871539 on Build GRCh38
rs781388896 Chr.4: 109871651-109871651 on Build GRCh38
rs564822946 Chr.4: 109871329-109871329 on Build GRCh38
rs930523977 Chr.4: 109871745-109871745 on Build GRCh38
rs971849927 Chr.4: 109871695-109871695 on Build GRCh38
rs912497873 Chr.4: 109871544-109871544 on Build GRCh38
rs982264122 Chr.4: 109871653-109871653 on Build GRCh38
rs542240600 Chr.4: 109871536-109871536 on Build GRCh38
rs6830168 Chr.4: 109871643-109871643 on Build GRCh38
rs1054714290 Chr.4: 109871617-109871617 on Build GRCh38
rs1016737571 Chr.4: 109871431-109871431 on Build GRCh38
rs1019526864 Chr.4: 109871753-109871753 on Build GRCh38
rs925008141 Chr.4: 109871600-109871600 on Build GRCh38
rs868117184 Chr.4: 109871774-109871774 on Build GRCh38
rs111245161 Chr.4: 109871770-109871770 on Build GRCh38
rs10011256 Chr.4: 109871751-109871751 on Build GRCh38
rs76150278 Chr.4: 109871339-109871339 on Build GRCh38
rs1016466951 Chr.4: 109871669-109871669 on Build GRCh38
rs978319370 Chr.4: 109871767-109871767 on Build GRCh38
rs1005064377 Chr.4: 109871419-109871419 on Build GRCh38
rs1029092008 Chr.4: 109871736-109871736 on Build GRCh38
rs1019163749 Chr.4: 109871561-109871561 on Build GRCh38
rs550462658 Chr.4: 109871415-109871415 on Build GRCh38
rs894774899 Chr.4: 109871625-109871625 on Build GRCh38
rs375326266 Chr.4: 109871453-109871453 on Build GRCh38
rs564004924 Chr.4: 109871472-109871472 on Build GRCh38
rs116786378 Chr.4: 109871612-109871612 on Build GRCh38
rs538349764 Chr.4: 109871328-109871328 on Build GRCh38
rs954666885 Chr.4: 109871748-109871748 on Build GRCh38
rs927801319 Chr.4: 109871384-109871384 on Build GRCh38
rs996237022 Chr.4: 109871732-109871732 on Build GRCh38
rs977973361 Chr.4: 109871559-109871559 on Build GRCh38
rs374407364 Chr.4: 109871664-109871664 on Build GRCh38
rs966678270 Chr.4: 109871766-109871766 on Build GRCh38
rs903343695 Chr.4: 109871565-109871565 on Build GRCh38
rs945182598 Chr.4: 109871557-109871557 on Build GRCh38
rs963524146 Chr.4: 109871471-109871471 on Build GRCh38
rs954696528 Chr.4: 109871484-109871484 on Build GRCh38
rs78668024 Chr.4: 109871686-109871686 on Build GRCh38

View Full Product Details