Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 117084259-117084772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAM1L1
Entrez Gene ID
133022
Gene Name
translocation associated membrane protein 1-like 1
Gene Aliases
-
UniGene
Hs.570737
Gene Chromosome Location
Chr.4: 117083554-117085580 on Build GRCh38
Transcripts
NM_152402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3365473 c.877G>T p.V293L Chr.4: 117084517-117084517 on Build GRCh38
4606638 c.730G>T p.E244* Chr.4: 117084664-117084664 on Build GRCh38
1426507 c.947T>A p.I316N Chr.4: 117084447-117084447 on Build GRCh38
3599384 c.655G>A p.G219R Chr.4: 117084739-117084739 on Build GRCh38
7094728 c.688G>A p.E230K Chr.4: 117084706-117084706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781280323 Chr.4: 117084401-117084401 on Build GRCh38
rs780613111 Chr.4: 117084679-117084679 on Build GRCh38
rs898012312 Chr.4: 117084435-117084435 on Build GRCh38
rs770073898 Chr.4: 117084324-117084324 on Build GRCh38
rs746791878 Chr.4: 117084706-117084706 on Build GRCh38
rs1045348072 Chr.4: 117084311-117084311 on Build GRCh38
rs142078992 Chr.4: 117084535-117084535 on Build GRCh38
rs1054688903 Chr.4: 117084310-117084310 on Build GRCh38
rs757517960 Chr.4: 117084650-117084650 on Build GRCh38
rs760262115 Chr.4: 117084363-117084363 on Build GRCh38
rs375243973 Chr.4: 117084328-117084328 on Build GRCh38
rs963691178 Chr.4: 117084521-117084521 on Build GRCh38
rs772059348 Chr.4: 117084384-117084385 on Build GRCh38
rs751520349 Chr.4: 117084528-117084528 on Build GRCh38
rs778712958 Chr.4: 117084284-117084284 on Build GRCh38
rs757713602 Chr.4: 117084561-117084561 on Build GRCh38
rs556393685 Chr.4: 117084700-117084700 on Build GRCh38
rs763630463 Chr.4: 117084367-117084367 on Build GRCh38
rs778050351 Chr.4: 117084417-117084417 on Build GRCh38
rs749454354 Chr.4: 117084557-117084557 on Build GRCh38
rs747686922 Chr.4: 117084576-117084576 on Build GRCh38
rs773583049 Chr.4: 117084330-117084330 on Build GRCh38
rs748978094 Chr.4: 117084701-117084701 on Build GRCh38
rs370338309 Chr.4: 117084406-117084406 on Build GRCh38
rs544392089 Chr.4: 117084383-117084383 on Build GRCh38
rs151149444 Chr.4: 117084573-117084573 on Build GRCh38
rs200676581 Chr.4: 117084671-117084671 on Build GRCh38
rs549346468 Chr.4: 117084575-117084575 on Build GRCh38
rs781396439 Chr.4: 117084304-117084304 on Build GRCh38
rs962128499 Chr.4: 117084673-117084673 on Build GRCh38
rs758285621 Chr.4: 117084374-117084374 on Build GRCh38
rs756351377 Chr.4: 117084548-117084548 on Build GRCh38
rs747412804 Chr.4: 117084682-117084682 on Build GRCh38
rs1015404475 Chr.4: 117084627-117084627 on Build GRCh38
rs754138688 Chr.4: 117084646-117084646 on Build GRCh38
rs763407171 Chr.4: 117084338-117084338 on Build GRCh38
rs778052836 Chr.4: 117084555-117084555 on Build GRCh38
rs746323628 Chr.4: 117084476-117084476 on Build GRCh38
rs1026899495 Chr.4: 117084564-117084564 on Build GRCh38
rs149752420 Chr.4: 117084318-117084318 on Build GRCh38
rs533288308 Chr.4: 117084524-117084524 on Build GRCh38
rs199904946 Chr.4: 117084458-117084458 on Build GRCh38
rs914442549 Chr.4: 117084312-117084312 on Build GRCh38
rs764927567 Chr.4: 117084492-117084492 on Build GRCh38
rs750351492 Chr.4: 117084288-117084288 on Build GRCh38
rs752787981 Chr.4: 117084631-117084631 on Build GRCh38
rs768307094 Chr.4: 117084488-117084488 on Build GRCh38
rs147601937 Chr.4: 117084504-117084504 on Build GRCh38
rs201199191 Chr.4: 117084296-117084296 on Build GRCh38
rs143062720 Chr.4: 117084536-117084536 on Build GRCh38
rs982665500 Chr.4: 117084622-117084622 on Build GRCh38
rs771698987 Chr.4: 117084733-117084733 on Build GRCh38
rs772926593 Chr.4: 117084585-117084585 on Build GRCh38
rs374666796 Chr.4: 117084543-117084543 on Build GRCh38
rs760814608 Chr.4: 117084635-117084635 on Build GRCh38
rs769115461 Chr.4: 117084584-117084584 on Build GRCh38
rs562701564 Chr.4: 117084489-117084489 on Build GRCh38
rs368586988 Chr.4: 117084526-117084526 on Build GRCh38
rs770547589 Chr.4: 117084712-117084712 on Build GRCh38
rs1041466311 Chr.4: 117084326-117084326 on Build GRCh38
rs868677235 Chr.4: 117084603-117084603 on Build GRCh38
rs17852851 Chr.4: 117084662-117084662 on Build GRCh38
rs751430857 Chr.4: 117084394-117084394 on Build GRCh38
rs200297518 Chr.4: 117084544-117084544 on Build GRCh38
rs758892219 Chr.4: 117084676-117084676 on Build GRCh38
rs868126129 Chr.4: 117084596-117084599 on Build GRCh38
rs759517217 Chr.4: 117084593-117084593 on Build GRCh38
rs758386040 Chr.4: 117084387-117084387 on Build GRCh38
rs1006401887 Chr.4: 117084534-117084534 on Build GRCh38
rs768932509 Chr.4: 117084303-117084303 on Build GRCh38
rs761479747 Chr.4: 117084377-117084377 on Build GRCh38
rs766298330 Chr.4: 117084390-117084390 on Build GRCh38
rs779462778 Chr.4: 117084669-117084669 on Build GRCh38
rs745724799 Chr.4: 117084354-117084355 on Build GRCh38
rs903982736 Chr.4: 117084317-117084317 on Build GRCh38
rs762544664 Chr.4: 117084589-117084589 on Build GRCh38
rs145609411 Chr.4: 117084356-117084356 on Build GRCh38
rs771080879 Chr.4: 117084295-117084296 on Build GRCh38
rs150280299 Chr.4: 117084697-117084697 on Build GRCh38
rs527665848 Chr.4: 117084568-117084568 on Build GRCh38
rs763927939 Chr.4: 117084728-117084730 on Build GRCh38
rs771632784 Chr.4: 117084463-117084463 on Build GRCh38
rs775345884 Chr.4: 117084518-117084518 on Build GRCh38
rs374685978 Chr.4: 117084412-117084412 on Build GRCh38
rs760485396 Chr.4: 117084743-117084743 on Build GRCh38
rs759373120 Chr.4: 117084717-117084717 on Build GRCh38
rs775511064 Chr.4: 117084740-117084740 on Build GRCh38
rs548978348 Chr.4: 117084464-117084464 on Build GRCh38
rs774600054 Chr.4: 117084318-117084322 on Build GRCh38
rs35809332 Chr.4: 117084302-117084302 on Build GRCh38
rs774188018 Chr.4: 117084714-117084714 on Build GRCh38
rs948036436 Chr.4: 117084742-117084742 on Build GRCh38
rs771544837 Chr.4: 117084350-117084350 on Build GRCh38
rs1019213655 Chr.4: 117084418-117084418 on Build GRCh38
rs145229083 Chr.4: 117084461-117084461 on Build GRCh38
rs140073704 Chr.4: 117084590-117084590 on Build GRCh38
rs567559679 Chr.4: 117084638-117084638 on Build GRCh38
rs760513041 Chr.4: 117084557-117084561 on Build GRCh38
rs767479036 Chr.4: 117084620-117084620 on Build GRCh38
rs755060097 Chr.4: 117084396-117084396 on Build GRCh38
rs764974473 Chr.4: 117084378-117084378 on Build GRCh38
rs201687962 Chr.4: 117084375-117084375 on Build GRCh38
rs780154992 Chr.4: 117084299-117084299 on Build GRCh38

View Full Product Details