Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 118295682-118296221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS12
Entrez Gene ID
8492
Gene Name
protease, serine 12
Gene Aliases
BSSP-3, BSSP3, MRT1
UniGene
Hs.445857
Gene Chromosome Location
Chr.4: 118280038-118353042 on Build GRCh38
Transcripts
NM_003619.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6331953 c.1860C>T p.G620G Chr.4: 118295834-118295834 on Build GRCh38
7418202 c.1880G>A p.R627Q Chr.4: 118295814-118295814 on Build GRCh38
1050340 c.1879C>T p.R627W Chr.4: 118295815-118295815 on Build GRCh38
7138891 c.1897G>T p.G633C Chr.4: 118295797-118295797 on Build GRCh38
1471744 c.1914A>T p.L638F Chr.4: 118295780-118295780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046140297 Chr.4: 118296004-118296004 on Build GRCh38
rs767904603 Chr.4: 118295763-118295763 on Build GRCh38
rs761411527 Chr.4: 118295773-118295773 on Build GRCh38
rs917125618 Chr.4: 118296136-118296136 on Build GRCh38
rs906338232 Chr.4: 118296034-118296034 on Build GRCh38
rs930129132 Chr.4: 118295915-118295915 on Build GRCh38
rs764887461 Chr.4: 118295784-118295784 on Build GRCh38
rs538483737 Chr.4: 118295882-118295882 on Build GRCh38
rs749392769 Chr.4: 118295747-118295747 on Build GRCh38
rs754920304 Chr.4: 118295815-118295815 on Build GRCh38
rs760021793 Chr.4: 118295760-118295760 on Build GRCh38
rs1030861417 Chr.4: 118296037-118296037 on Build GRCh38
rs788665 Chr.4: 118295959-118295959 on Build GRCh38
rs767503679 Chr.4: 118295913-118295913 on Build GRCh38
rs984596233 Chr.4: 118296078-118296078 on Build GRCh38
rs566067490 Chr.4: 118296057-118296057 on Build GRCh38
rs937952328 Chr.4: 118295791-118295791 on Build GRCh38
rs941694368 Chr.4: 118295958-118295958 on Build GRCh38
rs772490898 Chr.4: 118295897-118295897 on Build GRCh38
rs751500432 Chr.4: 118295814-118295814 on Build GRCh38
rs574779944 Chr.4: 118296127-118296127 on Build GRCh38
rs939843075 Chr.4: 118296115-118296115 on Build GRCh38
rs756346696 Chr.4: 118295831-118295831 on Build GRCh38
rs775761093 Chr.4: 118295767-118295767 on Build GRCh38
rs999348194 Chr.4: 118296036-118296036 on Build GRCh38
rs28503385 Chr.4: 118296063-118296063 on Build GRCh38
rs867261581 Chr.4: 118295757-118295757 on Build GRCh38
rs145983533 Chr.4: 118295842-118295842 on Build GRCh38
rs576111095 Chr.4: 118295951-118295951 on Build GRCh38
rs749590934 Chr.4: 118295851-118295851 on Build GRCh38
rs1010841888 Chr.4: 118296073-118296073 on Build GRCh38
rs771033155 Chr.4: 118295853-118295853 on Build GRCh38
rs1009774586 Chr.4: 118296023-118296023 on Build GRCh38
rs181703240 Chr.4: 118295766-118295766 on Build GRCh38
rs75215941 Chr.4: 118296100-118296100 on Build GRCh38
rs549665178 Chr.4: 118295805-118295805 on Build GRCh38
rs975442978 Chr.4: 118295734-118295734 on Build GRCh38
rs112381222 Chr.4: 118295901-118295902 on Build GRCh38
rs948656794 Chr.4: 118296184-118296184 on Build GRCh38
rs957073377 Chr.4: 118296007-118296007 on Build GRCh38
rs199606749 Chr.4: 118295806-118295806 on Build GRCh38
rs760331761 Chr.4: 118296114-118296114 on Build GRCh38
rs773377590 Chr.4: 118295740-118295740 on Build GRCh38
rs975390430 Chr.4: 118296047-118296047 on Build GRCh38
rs200497966 Chr.4: 118295786-118295786 on Build GRCh38
rs1047343541 Chr.4: 118295935-118295935 on Build GRCh38
rs1012381315 Chr.4: 118296000-118296000 on Build GRCh38
rs1012502514 Chr.4: 118295874-118295874 on Build GRCh38
rs556552045 Chr.4: 118296168-118296168 on Build GRCh38
rs752562770 Chr.4: 118295829-118295829 on Build GRCh38
rs894829575 Chr.4: 118295980-118295980 on Build GRCh38
rs976844493 Chr.4: 118295817-118295817 on Build GRCh38
rs770179339 Chr.4: 118295729-118295729 on Build GRCh38
rs778910959 Chr.4: 118295873-118295873 on Build GRCh38
rs888736562 Chr.4: 118295950-118295950 on Build GRCh38
rs148495776 Chr.4: 118295759-118295759 on Build GRCh38
rs781009683 Chr.4: 118295827-118295827 on Build GRCh38
rs952773628 Chr.4: 118296071-118296071 on Build GRCh38

View Full Product Details