Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 118298593-118299139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS12
Entrez Gene ID
8492
Gene Name
protease, serine 12
Gene Aliases
BSSP-3, BSSP3, MRT1
UniGene
Hs.445857
Gene Chromosome Location
Chr.4: 118280038-118353042 on Build GRCh38
Transcripts
NM_003619.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1050341 c.1743G>T p.K581N Chr.4: 118298827-118298827 on Build GRCh38
6893154 c.1725C>T p.S575S Chr.4: 118298845-118298845 on Build GRCh38
1426527 c.1637C>A p.P546H Chr.4: 118298933-118298933 on Build GRCh38
6166178 c.1632-2A>T p.? Chr.4: 118298940-118298940 on Build GRCh38
5788221 c.1771C>T p.H591Y Chr.4: 118298799-118298799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748815406 Chr.4: 118299023-118299026 on Build GRCh38
rs770550936 Chr.4: 118298951-118298951 on Build GRCh38
rs757463897 Chr.4: 118298787-118298787 on Build GRCh38
rs776219623 Chr.4: 118299089-118299089 on Build GRCh38
rs760652602 Chr.4: 118298976-118298976 on Build GRCh38
rs1024647487 Chr.4: 118298625-118298625 on Build GRCh38
rs750446857 Chr.4: 118298882-118298882 on Build GRCh38
rs34131974 Chr.4: 118298797-118298797 on Build GRCh38
rs748483554 Chr.4: 118298943-118298943 on Build GRCh38
rs1001369449 Chr.4: 118298971-118298971 on Build GRCh38
rs886059034 Chr.4: 118298782-118298782 on Build GRCh38
rs1035613261 Chr.4: 118298974-118298974 on Build GRCh38
rs111532161 Chr.4: 118298747-118298747 on Build GRCh38
rs1043641989 Chr.4: 118298755-118298755 on Build GRCh38
rs989247573 Chr.4: 118299032-118299032 on Build GRCh38
rs758401281 Chr.4: 118298903-118298903 on Build GRCh38
rs988856351 Chr.4: 118298744-118298744 on Build GRCh38
rs201470712 Chr.4: 118298732-118298732 on Build GRCh38
rs1026883988 Chr.4: 118298632-118298632 on Build GRCh38
rs906263224 Chr.4: 118299086-118299086 on Build GRCh38
rs777770197 Chr.4: 118298778-118298778 on Build GRCh38
rs764153997 Chr.4: 118298980-118298980 on Build GRCh38
rs1007930783 Chr.4: 118298847-118298847 on Build GRCh38
rs777027226 Chr.4: 118298824-118298824 on Build GRCh38
rs970455807 Chr.4: 118298675-118298675 on Build GRCh38
rs151322535 Chr.4: 118298860-118298860 on Build GRCh38
rs1050222821 Chr.4: 118298936-118298936 on Build GRCh38
rs201005601 Chr.4: 118298930-118298930 on Build GRCh38
rs913298615 Chr.4: 118299094-118299094 on Build GRCh38
rs757456129 Chr.4: 118298877-118298877 on Build GRCh38
rs941896254 Chr.4: 118298985-118298985 on Build GRCh38
rs780163106 Chr.4: 118298917-118298917 on Build GRCh38
rs148394828 Chr.4: 118298819-118298819 on Build GRCh38
rs370077581 Chr.4: 118298723-118298723 on Build GRCh38
rs190870811 Chr.4: 118298964-118298964 on Build GRCh38
rs878910417 Chr.4: 118298920-118298920 on Build GRCh38
rs771955102 Chr.4: 118298801-118298801 on Build GRCh38
rs142473280 Chr.4: 118298854-118298854 on Build GRCh38
rs781630455 Chr.4: 118299006-118299006 on Build GRCh38
rs776433731 Chr.4: 118298986-118298986 on Build GRCh38
rs765387681 Chr.4: 118298881-118298881 on Build GRCh38
rs763953524 Chr.4: 118298864-118298864 on Build GRCh38
rs775780097 Chr.4: 118298686-118298686 on Build GRCh38
rs561881893 Chr.4: 118298719-118298719 on Build GRCh38
rs758974333 Chr.4: 118298832-118298832 on Build GRCh38
rs146186927 Chr.4: 118298802-118298802 on Build GRCh38
rs769367735 Chr.4: 118298699-118298699 on Build GRCh38
rs767369478 Chr.4: 118298749-118298749 on Build GRCh38
rs372314430 Chr.4: 118298963-118298963 on Build GRCh38
rs772844127 Chr.4: 118298712-118298712 on Build GRCh38
rs775184293 Chr.4: 118298975-118298975 on Build GRCh38
rs974340188 Chr.4: 118298631-118298631 on Build GRCh38
rs762634563 Chr.4: 118298825-118298825 on Build GRCh38
rs140107913 Chr.4: 118298676-118298676 on Build GRCh38
rs766820784 Chr.4: 118298976-118298977 on Build GRCh38
rs969185901 Chr.4: 118299111-118299111 on Build GRCh38
rs1038018953 Chr.4: 118298993-118298993 on Build GRCh38
rs932187012 Chr.4: 118298789-118298789 on Build GRCh38
rs928936518 Chr.4: 118299060-118299060 on Build GRCh38
rs760746759 Chr.4: 118298861-118298861 on Build GRCh38
rs770519804 Chr.4: 118298826-118298826 on Build GRCh38
rs773854750 Chr.4: 118298830-118298830 on Build GRCh38
rs753721877 Chr.4: 118298866-118298866 on Build GRCh38
rs1034494273 Chr.4: 118298693-118298693 on Build GRCh38
rs1033459667 Chr.4: 118299114-118299114 on Build GRCh38
rs980002832 Chr.4: 118298683-118298683 on Build GRCh38
rs139381330 Chr.4: 118298925-118298925 on Build GRCh38
rs754093579 Chr.4: 118298783-118298783 on Build GRCh38
rs1046068014 Chr.4: 118299085-118299086 on Build GRCh38
rs761037876 Chr.4: 118298688-118298688 on Build GRCh38
rs772995173 Chr.4: 118298914-118298914 on Build GRCh38
rs779015369 Chr.4: 118298795-118298795 on Build GRCh38
rs771687782 Chr.4: 118298970-118298970 on Build GRCh38
rs900689449 Chr.4: 118298761-118298761 on Build GRCh38
rs755971322 Chr.4: 118298759-118298759 on Build GRCh38
rs781496371 Chr.4: 118298937-118298937 on Build GRCh38
rs200631717 Chr.4: 118298957-118298957 on Build GRCh38
rs938689371 Chr.4: 118298741-118298741 on Build GRCh38
rs28661939 Chr.4: 118298754-118298754 on Build GRCh38
rs897751491 Chr.4: 118299003-118299003 on Build GRCh38
rs879232387 Chr.4: 118299000-118299000 on Build GRCh38
rs747577971 Chr.4: 118298803-118298803 on Build GRCh38
rs759477288 Chr.4: 118298737-118298737 on Build GRCh38

View Full Product Details