Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121036353-121036852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDNF
Entrez Gene ID
79625
Gene Name
neuron derived neurotrophic factor
Gene Aliases
C4orf31, NORD
UniGene
Hs.709520
Gene Chromosome Location
Chr.4: 121035627-121072518 on Build GRCh38
Transcripts
NM_024574.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3917057 c.1252C>T p.L418F Chr.4: 121036719-121036719 on Build GRCh38
1731982 c.1379G>A p.R460H Chr.4: 121036592-121036592 on Build GRCh38
5622624 c.1372A>C p.K458Q Chr.4: 121036599-121036599 on Build GRCh38
6453615 c.1275G>A p.K425K Chr.4: 121036696-121036696 on Build GRCh38
1050511 c.1254C>T p.L418L Chr.4: 121036717-121036717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761790156 Chr.4: 121036514-121036514 on Build GRCh38
rs751807665 Chr.4: 121036557-121036557 on Build GRCh38
rs752982810 Chr.4: 121036781-121036783 on Build GRCh38
rs772364164 Chr.4: 121036785-121036785 on Build GRCh38
rs992511066 Chr.4: 121036722-121036722 on Build GRCh38
rs760854927 Chr.4: 121036658-121036658 on Build GRCh38
rs770609274 Chr.4: 121036741-121036741 on Build GRCh38
rs753230983 Chr.4: 121036572-121036572 on Build GRCh38
rs762583822 Chr.4: 121036607-121036607 on Build GRCh38
rs559657072 Chr.4: 121036589-121036589 on Build GRCh38
rs376739357 Chr.4: 121036636-121036636 on Build GRCh38
rs759273566 Chr.4: 121036782-121036782 on Build GRCh38
rs767608563 Chr.4: 121036617-121036617 on Build GRCh38
rs201892645 Chr.4: 121036712-121036712 on Build GRCh38
rs34766411 Chr.4: 121036404-121036404 on Build GRCh38
rs746437706 Chr.4: 121036510-121036510 on Build GRCh38
rs767760684 Chr.4: 121036569-121036569 on Build GRCh38
rs766655169 Chr.4: 121036405-121036405 on Build GRCh38
rs147508531 Chr.4: 121036410-121036410 on Build GRCh38
rs372074352 Chr.4: 121036573-121036573 on Build GRCh38
rs759848890 Chr.4: 121036644-121036644 on Build GRCh38
rs762828906 Chr.4: 121036548-121036548 on Build GRCh38
rs752598151 Chr.4: 121036700-121036700 on Build GRCh38
rs774847432 Chr.4: 121036518-121036520 on Build GRCh38
rs111583710 Chr.4: 121036730-121036730 on Build GRCh38
rs140044869 Chr.4: 121036806-121036806 on Build GRCh38
rs755094600 Chr.4: 121036435-121036435 on Build GRCh38
rs756530609 Chr.4: 121036481-121036481 on Build GRCh38
rs750747993 Chr.4: 121036823-121036823 on Build GRCh38
rs768280639 Chr.4: 121036512-121036512 on Build GRCh38
rs1008241793 Chr.4: 121036519-121036519 on Build GRCh38
rs760015105 Chr.4: 121036582-121036582 on Build GRCh38
rs772951169 Chr.4: 121036543-121036543 on Build GRCh38
rs766454071 Chr.4: 121036552-121036552 on Build GRCh38
rs1041408706 Chr.4: 121036540-121036540 on Build GRCh38
rs781480744 Chr.4: 121036477-121036477 on Build GRCh38
rs143169450 Chr.4: 121036413-121036413 on Build GRCh38
rs768030210 Chr.4: 121036586-121036586 on Build GRCh38
rs1053021247 Chr.4: 121036482-121036482 on Build GRCh38
rs774420436 Chr.4: 121036635-121036635 on Build GRCh38
rs1397645 Chr.4: 121036579-121036579 on Build GRCh38
rs774239705 Chr.4: 121036773-121036773 on Build GRCh38
rs202169296 Chr.4: 121036716-121036716 on Build GRCh38
rs528073294 Chr.4: 121036678-121036678 on Build GRCh38
rs746734949 Chr.4: 121036582-121036582 on Build GRCh38
rs750619386 Chr.4: 121036394-121036394 on Build GRCh38
rs999356853 Chr.4: 121036568-121036568 on Build GRCh38
rs755005486 Chr.4: 121036563-121036563 on Build GRCh38
rs775226205 Chr.4: 121036495-121036495 on Build GRCh38
rs769294164 Chr.4: 121036719-121036719 on Build GRCh38
rs752779002 Chr.4: 121036657-121036657 on Build GRCh38
rs778325003 Chr.4: 121036485-121036485 on Build GRCh38
rs778449790 Chr.4: 121036574-121036574 on Build GRCh38
rs148977529 Chr.4: 121036717-121036717 on Build GRCh38
rs757550265 Chr.4: 121036687-121036687 on Build GRCh38
rs745563516 Chr.4: 121036713-121036713 on Build GRCh38
rs143875438 Chr.4: 121036593-121036593 on Build GRCh38
rs773145811 Chr.4: 121036606-121036606 on Build GRCh38
rs775859713 Chr.4: 121036789-121036789 on Build GRCh38
rs771578483 Chr.4: 121036492-121036492 on Build GRCh38
rs749704430 Chr.4: 121036490-121036490 on Build GRCh38
rs1013457950 Chr.4: 121036434-121036434 on Build GRCh38
rs368469233 Chr.4: 121036479-121036479 on Build GRCh38
rs764632595 Chr.4: 121036665-121036665 on Build GRCh38
rs760832661 Chr.4: 121036802-121036802 on Build GRCh38
rs146653001 Chr.4: 121036531-121036531 on Build GRCh38
rs747685788 Chr.4: 121036592-121036592 on Build GRCh38
rs770822561 Chr.4: 121036619-121036619 on Build GRCh38
rs200281108 Chr.4: 121036576-121036576 on Build GRCh38
rs574941094 Chr.4: 121036379-121036379 on Build GRCh38
rs777682795 Chr.4: 121036721-121036721 on Build GRCh38
rs919433409 Chr.4: 121036760-121036760 on Build GRCh38
rs374404232 Chr.4: 121036411-121036411 on Build GRCh38
rs767885830 Chr.4: 121036652-121036652 on Build GRCh38
rs749304815 Chr.4: 121036737-121036737 on Build GRCh38
rs762425792 Chr.4: 121036814-121036814 on Build GRCh38

View Full Product Details