Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 123402245-123402742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRY1
Entrez Gene ID
10252
Gene Name
sprouty RTK signaling antagonist 1
Gene Aliases
hSPRY1
UniGene
Hs.744854
Gene Chromosome Location
Chr.4: 123396791-123403760 on Build GRCh38
Transcripts
NM_001258038.1 NM_001258039.1 NM_005841.2 NM_199327.2 XM_005262686.1 XM_017007635.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6269268 c.857G>T p.C286F Chr.4: 123402448-123402448 on Build GRCh38
3825124 c.956C>G p.S319* Chr.4: 123402547-123402547 on Build GRCh38
6043807 c.899C>G p.S300C Chr.4: 123402490-123402490 on Build GRCh38
1753523 c.944A>G p.Q315R Chr.4: 123402535-123402535 on Build GRCh38
4890599 c.911A>G p.Y304C Chr.4: 123402502-123402502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762804313 Chr.4: 123402534-123402534 on Build GRCh38
rs762280661 Chr.4: 123402313-123402313 on Build GRCh38
rs866445691 Chr.4: 123402325-123402325 on Build GRCh38
rs993078015 Chr.4: 123402651-123402651 on Build GRCh38
rs748590054 Chr.4: 123402374-123402374 on Build GRCh38
rs1052704088 Chr.4: 123402565-123402565 on Build GRCh38
rs775260056 Chr.4: 123402398-123402398 on Build GRCh38
rs573341223 Chr.4: 123402705-123402705 on Build GRCh38
rs778182939 Chr.4: 123402568-123402568 on Build GRCh38
rs750666163 Chr.4: 123402353-123402353 on Build GRCh38
rs775293323 Chr.4: 123402418-123402419 on Build GRCh38
rs774458098 Chr.4: 123402604-123402606 on Build GRCh38
rs780877260 Chr.4: 123402591-123402591 on Build GRCh38
rs774111531 Chr.4: 123402323-123402323 on Build GRCh38
rs746592977 Chr.4: 123402496-123402497 on Build GRCh38
rs779233192 Chr.4: 123402288-123402288 on Build GRCh38
rs773382084 Chr.4: 123402378-123402378 on Build GRCh38
rs138952375 Chr.4: 123402296-123402296 on Build GRCh38
rs370375351 Chr.4: 123402452-123402452 on Build GRCh38
rs754362074 Chr.4: 123402574-123402574 on Build GRCh38
rs548544234 Chr.4: 123402577-123402577 on Build GRCh38
rs759688943 Chr.4: 123402548-123402548 on Build GRCh38
rs899992010 Chr.4: 123402622-123402622 on Build GRCh38
rs761837505 Chr.4: 123402522-123402522 on Build GRCh38
rs757750128 Chr.4: 123402576-123402576 on Build GRCh38
rs142173292 Chr.4: 123402300-123402300 on Build GRCh38
rs756537678 Chr.4: 123402493-123402493 on Build GRCh38
rs766150666 Chr.4: 123402535-123402535 on Build GRCh38
rs760710183 Chr.4: 123402341-123402341 on Build GRCh38
rs772249078 Chr.4: 123402292-123402292 on Build GRCh38
rs961876940 Chr.4: 123402411-123402411 on Build GRCh38
rs775967448 Chr.4: 123402521-123402521 on Build GRCh38
rs1040973202 Chr.4: 123402597-123402597 on Build GRCh38
rs764924905 Chr.4: 123402352-123402352 on Build GRCh38
rs752212870 Chr.4: 123402333-123402333 on Build GRCh38
rs771800060 Chr.4: 123402383-123402383 on Build GRCh38
rs770209344 Chr.4: 123402376-123402376 on Build GRCh38
rs200878762 Chr.4: 123402415-123402415 on Build GRCh38
rs768290759 Chr.4: 123402517-123402517 on Build GRCh38
rs528974183 Chr.4: 123402551-123402551 on Build GRCh38
rs977420515 Chr.4: 123402701-123402701 on Build GRCh38
rs148117048 Chr.4: 123402649-123402649 on Build GRCh38
rs764172522 Chr.4: 123402344-123402344 on Build GRCh38
rs769345890 Chr.4: 123402601-123402601 on Build GRCh38
rs751794748 Chr.4: 123402547-123402547 on Build GRCh38
rs367610616 Chr.4: 123402495-123402495 on Build GRCh38
rs759052208 Chr.4: 123402332-123402332 on Build GRCh38
rs776384905 Chr.4: 123402284-123402284 on Build GRCh38
rs745565941 Chr.4: 123402382-123402382 on Build GRCh38
rs771387469 Chr.4: 123402505-123402505 on Build GRCh38
rs374392795 Chr.4: 123402455-123402455 on Build GRCh38
rs776763816 Chr.4: 123402414-123402414 on Build GRCh38
rs766629767 Chr.4: 123402438-123402438 on Build GRCh38
rs748693044 Chr.4: 123402466-123402466 on Build GRCh38
rs760299005 Chr.4: 123402412-123402412 on Build GRCh38
rs770979137 Chr.4: 123402530-123402531 on Build GRCh38
rs768376623 Chr.4: 123402584-123402584 on Build GRCh38
rs781715595 Chr.4: 123402373-123402373 on Build GRCh38
rs952464869 Chr.4: 123402645-123402645 on Build GRCh38
rs746642537 Chr.4: 123402507-123402507 on Build GRCh38
rs750140832 Chr.4: 123402427-123402427 on Build GRCh38
rs529529688 Chr.4: 123402675-123402675 on Build GRCh38
rs755021441 Chr.4: 123402358-123402358 on Build GRCh38
rs973274373 Chr.4: 123402487-123402487 on Build GRCh38
rs192013147 Chr.4: 123402693-123402693 on Build GRCh38
rs377271490 Chr.4: 123402562-123402562 on Build GRCh38
rs968045586 Chr.4: 123402691-123402691 on Build GRCh38
rs752669152 Chr.4: 123402564-123402564 on Build GRCh38
rs753793837 Chr.4: 123402345-123402345 on Build GRCh38
rs770049527 Chr.4: 123402317-123402317 on Build GRCh38
rs749662101 Chr.4: 123402500-123402500 on Build GRCh38
rs1043901201 Chr.4: 123402610-123402610 on Build GRCh38
rs368008252 Chr.4: 123402420-123402420 on Build GRCh38
rs566242109 Chr.4: 123402604-123402605 on Build GRCh38
rs773173631 Chr.4: 123402529-123402529 on Build GRCh38
rs777102609 Chr.4: 123402303-123402303 on Build GRCh38
rs145077253 Chr.4: 123402275-123402275 on Build GRCh38
rs952571434 Chr.4: 123402299-123402299 on Build GRCh38
rs751778456 Chr.4: 123402439-123402439 on Build GRCh38
rs374261636 Chr.4: 123402355-123402355 on Build GRCh38
rs551247908 Chr.4: 123402636-123402636 on Build GRCh38
rs300575 Chr.4: 123402628-123402628 on Build GRCh38
rs756195888 Chr.4: 123402567-123402567 on Build GRCh38
rs1030131991 Chr.4: 123402643-123402643 on Build GRCh38
rs780045221 Chr.4: 123402295-123402295 on Build GRCh38
rs755109197 Chr.4: 123402444-123402444 on Build GRCh38
rs369741147 Chr.4: 123402648-123402649 on Build GRCh38
rs746069665 Chr.4: 123402291-123402291 on Build GRCh38
rs891136732 Chr.4: 123402647-123402647 on Build GRCh38
rs139825476 Chr.4: 123402603-123402604 on Build GRCh38
rs747655877 Chr.4: 123402595-123402595 on Build GRCh38
rs187259850 Chr.4: 123402478-123402478 on Build GRCh38
rs765138425 Chr.4: 123402528-123402528 on Build GRCh38
rs775346634 Chr.4: 123402506-123402506 on Build GRCh38
rs539974780 Chr.4: 123402683-123402683 on Build GRCh38
rs936069833 Chr.4: 123402706-123402706 on Build GRCh38
rs578112056 Chr.4: 123402347-123402347 on Build GRCh38
rs1004583075 Chr.4: 123402668-123402668 on Build GRCh38
rs300574 Chr.4: 123402583-123402583 on Build GRCh38
rs571261618 Chr.4: 123402646-123402646 on Build GRCh38

View Full Product Details