Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 128270294-128270793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGRMC2
Entrez Gene ID
10424
Gene Name
progesterone receptor membrane component 2
Gene Aliases
DG6, PMBP
UniGene
Hs.507910
Gene Chromosome Location
Chr.4: 128269237-128288829 on Build GRCh38
Transcripts
NM_006320.4

SNP Detail

SNP ID Chromosome Location
rs576230932 Chr.4: 128270397-128270397 on Build GRCh38
rs374253083 Chr.4: 128270667-128270667 on Build GRCh38
rs41300347 Chr.4: 128270678-128270678 on Build GRCh38
rs775647802 Chr.4: 128270717-128270717 on Build GRCh38
rs897534361 Chr.4: 128270761-128270761 on Build GRCh38
rs1015057555 Chr.4: 128270452-128270452 on Build GRCh38
rs904357149 Chr.4: 128270669-128270669 on Build GRCh38
rs543526056 Chr.4: 128270407-128270407 on Build GRCh38
rs541172624 Chr.4: 128270675-128270675 on Build GRCh38
rs1050319235 Chr.4: 128270691-128270691 on Build GRCh38
rs1004840833 Chr.4: 128270424-128270424 on Build GRCh38
rs765437263 Chr.4: 128270548-128270548 on Build GRCh38
rs953215981 Chr.4: 128270513-128270513 on Build GRCh38
rs1010462123 Chr.4: 128270368-128270368 on Build GRCh38
rs1045817907 Chr.4: 128270335-128270335 on Build GRCh38
rs913468175 Chr.4: 128270642-128270642 on Build GRCh38
rs973272063 Chr.4: 128270493-128270493 on Build GRCh38
rs752585617 Chr.4: 128270351-128270351 on Build GRCh38
rs956366958 Chr.4: 128270589-128270589 on Build GRCh38
rs533055557 Chr.4: 128270707-128270707 on Build GRCh38
rs891640566 Chr.4: 128270367-128270367 on Build GRCh38
rs3839124 Chr.4: 128270562-128270562 on Build GRCh38
rs868597560 Chr.4: 128270453-128270453 on Build GRCh38
rs992946348 Chr.4: 128270617-128270617 on Build GRCh38
rs41300349 Chr.4: 128270378-128270378 on Build GRCh38
rs1028758768 Chr.4: 128270500-128270500 on Build GRCh38
rs567193760 Chr.4: 128270739-128270744 on Build GRCh38
rs1028704348 Chr.4: 128270419-128270419 on Build GRCh38
rs994587587 Chr.4: 128270766-128270766 on Build GRCh38
rs890139478 Chr.4: 128270718-128270718 on Build GRCh38
rs927289650 Chr.4: 128270330-128270330 on Build GRCh38
rs937371857 Chr.4: 128270394-128270394 on Build GRCh38
rs913270292 Chr.4: 128270621-128270621 on Build GRCh38
rs552181972 Chr.4: 128270715-128270715 on Build GRCh38
rs1044296 Chr.4: 128270736-128270736 on Build GRCh38
rs1008673796 Chr.4: 128270431-128270431 on Build GRCh38
rs909245391 Chr.4: 128270764-128270764 on Build GRCh38
rs1049141532 Chr.4: 128270732-128270732 on Build GRCh38
rs1036271763 Chr.4: 128270474-128270474 on Build GRCh38
rs901704655 Chr.4: 128270475-128270475 on Build GRCh38
rs927408322 Chr.4: 128270699-128270699 on Build GRCh38
rs749287447 Chr.4: 128270486-128270486 on Build GRCh38
rs397841430 Chr.4: 128270566-128270566 on Build GRCh38
rs938653057 Chr.4: 128270596-128270596 on Build GRCh38
rs1000541683 Chr.4: 128270531-128270531 on Build GRCh38
rs938803929 Chr.4: 128270333-128270336 on Build GRCh38
rs890106259 Chr.4: 128270427-128270427 on Build GRCh38
rs757310511 Chr.4: 128270502-128270502 on Build GRCh38
rs545858327 Chr.4: 128270319-128270319 on Build GRCh38
rs746118808 Chr.4: 128270448-128270448 on Build GRCh38
rs112811729 Chr.4: 128270641-128270641 on Build GRCh38
rs144077350 Chr.4: 128270497-128270497 on Build GRCh38
rs927345951 Chr.4: 128270344-128270344 on Build GRCh38
rs1049498022 Chr.4: 128270399-128270399 on Build GRCh38
rs1032464769 Chr.4: 128270580-128270580 on Build GRCh38
rs971556965 Chr.4: 128270644-128270644 on Build GRCh38

View Full Product Details