Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145081663-145082128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC10
Entrez Gene ID
10393
Gene Name
anaphase promoting complex subunit 10
Gene Aliases
APC10, DOC1
UniGene
Hs.480876
Gene Chromosome Location
Chr.4: 144967117-145098541 on Build GRCh38
Transcripts
NM_001256706.1 NM_001256707.1 NM_001256708.1 NM_001256709.1 NM_001256710.1 NM_001256711.1 NM_001256712.1 NM_001318367.1 NM_014885.4 NR_046345.1 XM_011531524.2 XM_011531525.2 XM_011531526.2 XM_011531527.2 XM_011531529.2 XM_011531530.2 XM_011531531.2 XM_011531532.2 XM_017007638.1 XM_017007639.1 XM_017007640.1 XM_017007641.1 XM_017007642.1 XM_017007643.1 XM_017007644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5785197 c.173G>A p.G58D Chr.4: 145081693-145081693 on Build GRCh38
4427714 c.136C>T p.R46* Chr.4: 145081730-145081730 on Build GRCh38
3126614 c.167C>T p.S56L Chr.4: 145081699-145081699 on Build GRCh38
1618443 c.198C>T p.I66I Chr.4: 145081668-145081668 on Build GRCh38
3660910 c.147T>C p.N49N Chr.4: 145081719-145081719 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs574388588 Chr.4: 145081872-145081872 on Build GRCh38
rs539657406 Chr.4: 145081716-145081716 on Build GRCh38
rs554591698 Chr.4: 145081838-145081838 on Build GRCh38
rs749034185 Chr.4: 145081748-145081748 on Build GRCh38
rs551215349 Chr.4: 145082098-145082098 on Build GRCh38
rs543696011 Chr.4: 145081881-145081881 on Build GRCh38
rs749451108 Chr.4: 145081824-145081824 on Build GRCh38
rs35257136 Chr.4: 145081729-145081729 on Build GRCh38
rs543358853 Chr.4: 145082049-145082049 on Build GRCh38
rs563435878 Chr.4: 145081883-145081883 on Build GRCh38
rs968321807 Chr.4: 145081909-145081909 on Build GRCh38
rs989771045 Chr.4: 145081802-145081802 on Build GRCh38
rs201073598 Chr.4: 145081800-145081800 on Build GRCh38
rs1031197300 Chr.4: 145081742-145081742 on Build GRCh38
rs34267023 Chr.4: 145081888-145081888 on Build GRCh38
rs778587349 Chr.4: 145081878-145081878 on Build GRCh38
rs771316366 Chr.4: 145082034-145082034 on Build GRCh38
rs759381610 Chr.4: 145081782-145081782 on Build GRCh38
rs953701079 Chr.4: 145081812-145081812 on Build GRCh38
rs748172940 Chr.4: 145081762-145081763 on Build GRCh38
rs201246162 Chr.4: 145081708-145081708 on Build GRCh38
rs916551953 Chr.4: 145081929-145081929 on Build GRCh38
rs373900165 Chr.4: 145081792-145081792 on Build GRCh38
rs377339394 Chr.4: 145081973-145081973 on Build GRCh38
rs779333814 Chr.4: 145081727-145081727 on Build GRCh38
rs893981690 Chr.4: 145082027-145082027 on Build GRCh38
rs1053032229 Chr.4: 145082004-145082004 on Build GRCh38
rs779983258 Chr.4: 145081734-145081734 on Build GRCh38
rs779900815 Chr.4: 145081741-145081741 on Build GRCh38
rs755696637 Chr.4: 145081696-145081696 on Build GRCh38
rs768722828 Chr.4: 145081740-145081740 on Build GRCh38
rs781488027 Chr.4: 145082094-145082094 on Build GRCh38
rs562541555 Chr.4: 145081806-145081806 on Build GRCh38
rs958269865 Chr.4: 145081763-145081763 on Build GRCh38
rs530815812 Chr.4: 145081968-145081968 on Build GRCh38
rs1038308986 Chr.4: 145082066-145082066 on Build GRCh38
rs758568951 Chr.4: 145081730-145081730 on Build GRCh38
rs561745336 Chr.4: 145081966-145081966 on Build GRCh38
rs896968777 Chr.4: 145081873-145081873 on Build GRCh38
rs752455250 Chr.4: 145081694-145081694 on Build GRCh38
rs6815519 Chr.4: 145081935-145081935 on Build GRCh38
rs754495302 Chr.4: 145081767-145081767 on Build GRCh38
rs774828431 Chr.4: 145081797-145081797 on Build GRCh38
rs965183529 Chr.4: 145081877-145081877 on Build GRCh38
rs985127095 Chr.4: 145082007-145082007 on Build GRCh38
rs910875606 Chr.4: 145082036-145082036 on Build GRCh38
rs747297622 Chr.4: 145081738-145081738 on Build GRCh38
rs545694514 Chr.4: 145081894-145081894 on Build GRCh38
rs929832704 Chr.4: 145082095-145082095 on Build GRCh38
rs1025493515 Chr.4: 145081699-145081699 on Build GRCh38
rs1031649475 Chr.4: 145081930-145081930 on Build GRCh38
rs771000852 Chr.4: 145081761-145081761 on Build GRCh38
rs559392230 Chr.4: 145081912-145081912 on Build GRCh38
rs570980930 Chr.4: 145081707-145081707 on Build GRCh38
rs999651528 Chr.4: 145081998-145081998 on Build GRCh38
rs757780229 Chr.4: 145081723-145081723 on Build GRCh38
rs912321796 Chr.4: 145081873-145081873 on Build GRCh38
rs145679062 Chr.4: 145081958-145081958 on Build GRCh38
rs1023636926 Chr.4: 145081902-145081902 on Build GRCh38

View Full Product Details