Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145136394-145136894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUD4
Entrez Gene ID
54726
Gene Name
OTU deubiquitinase 4
Gene Aliases
DUBA6, HIN1, HSHIN1
UniGene
Hs.270851
Gene Chromosome Location
Chr.4: 145133650-145180019 on Build GRCh38
Transcripts
NM_001102653.1 XM_005263079.4 XM_011532040.2 XM_011532041.2 XM_011532042.2

SNP Detail

SNP ID Chromosome Location
rs915039150 Chr.4: 145136728-145136728 on Build GRCh38
rs1042841312 Chr.4: 145136784-145136784 on Build GRCh38
rs890474668 Chr.4: 145136786-145136786 on Build GRCh38
rs28634337 Chr.4: 145136478-145136478 on Build GRCh38
rs1026770392 Chr.4: 145136581-145136581 on Build GRCh38
rs945410579 Chr.4: 145136509-145136509 on Build GRCh38
rs752879237 Chr.4: 145136473-145136473 on Build GRCh38
rs752031704 Chr.4: 145136597-145136597 on Build GRCh38
rs796950169 Chr.4: 145136477-145136477 on Build GRCh38
rs568372703 Chr.4: 145136513-145136513 on Build GRCh38
rs926512620 Chr.4: 145136638-145136638 on Build GRCh38
rs769298389 Chr.4: 145136463-145136464 on Build GRCh38
rs71250026 Chr.4: 145136485-145136486 on Build GRCh38
rs186594384 Chr.4: 145136637-145136637 on Build GRCh38
rs1823939 Chr.4: 145136454-145136454 on Build GRCh38
rs573765305 Chr.4: 145136574-145136574 on Build GRCh38
rs759248604 Chr.4: 145136646-145136646 on Build GRCh38
rs995002206 Chr.4: 145136578-145136579 on Build GRCh38
rs3063573 Chr.4: 145136486-145136487 on Build GRCh38
rs542774005 Chr.4: 145136591-145136591 on Build GRCh38
rs368967462 Chr.4: 145136694-145136695 on Build GRCh38
rs547179807 Chr.4: 145136796-145136796 on Build GRCh38
rs944689945 Chr.4: 145136793-145136793 on Build GRCh38
rs765345885 Chr.4: 145136468-145136468 on Build GRCh38
rs764367711 Chr.4: 145136470-145136487 on Build GRCh38
rs3063572 Chr.4: 145136476-145136477 on Build GRCh38
rs531174940 Chr.4: 145136610-145136610 on Build GRCh38
rs28488496 Chr.4: 145136539-145136539 on Build GRCh38
rs779128829 Chr.4: 145136780-145136781 on Build GRCh38
rs756417497 Chr.4: 145136723-145136723 on Build GRCh38
rs555245071 Chr.4: 145136551-145136551 on Build GRCh38
rs397706799 Chr.4: 145136488-145136489 on Build GRCh38
rs547376478 Chr.4: 145136534-145136534 on Build GRCh38
rs567156801 Chr.4: 145136798-145136798 on Build GRCh38
rs1027827358 Chr.4: 145136815-145136815 on Build GRCh38
rs777256700 Chr.4: 145136569-145136569 on Build GRCh38
rs937883409 Chr.4: 145136663-145136663 on Build GRCh38
rs562594148 Chr.4: 145136605-145136605 on Build GRCh38
rs182445387 Chr.4: 145136748-145136748 on Build GRCh38
rs76459724 Chr.4: 145136488-145136488 on Build GRCh38
rs867352823 Chr.4: 145136482-145136482 on Build GRCh38
rs995054454 Chr.4: 145136809-145136810 on Build GRCh38
rs7657990 Chr.4: 145136462-145136462 on Build GRCh38
rs868699565 Chr.4: 145136764-145136764 on Build GRCh38
rs893887587 Chr.4: 145136473-145136477 on Build GRCh38
rs1015279755 Chr.4: 145136593-145136593 on Build GRCh38
rs7657989 Chr.4: 145136460-145136460 on Build GRCh38
rs36227003 Chr.4: 145136672-145136672 on Build GRCh38
rs13136546 Chr.4: 145136471-145136471 on Build GRCh38
rs890412959 Chr.4: 145136519-145136519 on Build GRCh38
rs759840979 Chr.4: 145136463-145136463 on Build GRCh38
rs982491796 Chr.4: 145136592-145136592 on Build GRCh38
rs937832342 Chr.4: 145136439-145136439 on Build GRCh38
rs201466430 Chr.4: 145136485-145136485 on Build GRCh38
rs952448907 Chr.4: 145136587-145136587 on Build GRCh38
rs1002248683 Chr.4: 145136424-145136436 on Build GRCh38

View Full Product Details