Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 146255828-146256342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC10A7
Entrez Gene ID
84068
Gene Name
solute carrier family 10 member 7
Gene Aliases
C4orf13, P7
UniGene
Hs.659209
Gene Chromosome Location
Chr.4: 146253981-146522351 on Build GRCh38
Transcripts
NM_001029998.5 NM_001300842.2 NM_001317816.1 NR_133924.1 XM_011532311.2 XM_011532313.2 XM_011532314.2

SNP Detail

SNP ID Chromosome Location
rs1004874133 Chr.4: 146255986-146255986 on Build GRCh38
rs957622053 Chr.4: 146256118-146256118 on Build GRCh38
rs987774932 Chr.4: 146256169-146256169 on Build GRCh38
rs955428812 Chr.4: 146256151-146256151 on Build GRCh38
rs911851162 Chr.4: 146256175-146256175 on Build GRCh38
rs971876093 Chr.4: 146256235-146256235 on Build GRCh38
rs903349209 Chr.4: 146255914-146255914 on Build GRCh38
rs912299534 Chr.4: 146255872-146255872 on Build GRCh38
rs190892332 Chr.4: 146256297-146256297 on Build GRCh38
rs942577751 Chr.4: 146255884-146255884 on Build GRCh38
rs113968577 Chr.4: 146256288-146256288 on Build GRCh38
rs528150053 Chr.4: 146256071-146256071 on Build GRCh38
rs920323611 Chr.4: 146256252-146256252 on Build GRCh38
rs919739554 Chr.4: 146256241-146256241 on Build GRCh38
rs912315807 Chr.4: 146256198-146256198 on Build GRCh38
rs1028753104 Chr.4: 146256098-146256098 on Build GRCh38
rs11930848 Chr.4: 146256316-146256316 on Build GRCh38
rs539385960 Chr.4: 146256001-146256001 on Build GRCh38
rs963731535 Chr.4: 146256216-146256216 on Build GRCh38
rs1033059333 Chr.4: 146255975-146255975 on Build GRCh38
rs562311254 Chr.4: 146256214-146256214 on Build GRCh38
rs967476476 Chr.4: 146256170-146256170 on Build GRCh38
rs1020515368 Chr.4: 146256162-146256162 on Build GRCh38
rs975387404 Chr.4: 146256052-146256052 on Build GRCh38
rs1034480627 Chr.4: 146256116-146256116 on Build GRCh38
rs999057128 Chr.4: 146255915-146255915 on Build GRCh38
rs930410875 Chr.4: 146256285-146256285 on Build GRCh38
rs1057560 Chr.4: 146256189-146256189 on Build GRCh38
rs1039474226 Chr.4: 146255886-146255886 on Build GRCh38
rs182205936 Chr.4: 146256013-146256013 on Build GRCh38
rs570549061 Chr.4: 146255970-146255970 on Build GRCh38
rs866707106 Chr.4: 146255905-146255905 on Build GRCh38
rs572929720 Chr.4: 146256016-146256016 on Build GRCh38
rs940725056 Chr.4: 146256291-146256291 on Build GRCh38
rs1001704051 Chr.4: 146256005-146256005 on Build GRCh38
rs886464675 Chr.4: 146255973-146255973 on Build GRCh38
rs898425977 Chr.4: 146255889-146255889 on Build GRCh38
rs907282548 Chr.4: 146256004-146256004 on Build GRCh38
rs1049080590 Chr.4: 146255955-146255955 on Build GRCh38
rs931144370 Chr.4: 146256244-146256247 on Build GRCh38
rs957443830 Chr.4: 146255993-146255993 on Build GRCh38
rs1024584414 Chr.4: 146255912-146255912 on Build GRCh38

View Full Product Details