Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152627473-152627981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM154
Entrez Gene ID
201799
Gene Name
transmembrane protein 154
Gene Aliases
-
UniGene
Hs.518900
Gene Chromosome Location
Chr.4: 152626114-152680165 on Build GRCh38
Transcripts
NM_152680.2

SNP Detail

SNP ID Chromosome Location
rs956649886 Chr.4: 152627598-152627598 on Build GRCh38
rs747783565 Chr.4: 152627597-152627597 on Build GRCh38
rs185314391 Chr.4: 152627530-152627530 on Build GRCh38
rs556769904 Chr.4: 152627523-152627523 on Build GRCh38
rs762548711 Chr.4: 152627746-152627746 on Build GRCh38
rs570223523 Chr.4: 152627766-152627766 on Build GRCh38
rs929582952 Chr.4: 152627921-152627921 on Build GRCh38
rs77398858 Chr.4: 152627890-152627890 on Build GRCh38
rs551909067 Chr.4: 152627657-152627657 on Build GRCh38
rs796661271 Chr.4: 152627699-152627699 on Build GRCh38
rs1045093550 Chr.4: 152627756-152627756 on Build GRCh38
rs926353680 Chr.4: 152627778-152627778 on Build GRCh38
rs1007482112 Chr.4: 152627919-152627919 on Build GRCh38
rs964238250 Chr.4: 152627556-152627556 on Build GRCh38
rs539595127 Chr.4: 152627547-152627547 on Build GRCh38
rs939096600 Chr.4: 152627817-152627817 on Build GRCh38
rs1018276635 Chr.4: 152627552-152627552 on Build GRCh38
rs984304183 Chr.4: 152627650-152627650 on Build GRCh38
rs147649549 Chr.4: 152627641-152627641 on Build GRCh38
rs953333933 Chr.4: 152627537-152627537 on Build GRCh38
rs892286455 Chr.4: 152627854-152627854 on Build GRCh38
rs1051950226 Chr.4: 152627944-152627944 on Build GRCh38
rs556069687 Chr.4: 152627897-152627897 on Build GRCh38
rs1017658312 Chr.4: 152627551-152627551 on Build GRCh38
rs979218303 Chr.4: 152627568-152627568 on Build GRCh38
rs776454955 Chr.4: 152627596-152627596 on Build GRCh38
rs145289189 Chr.4: 152627610-152627610 on Build GRCh38
rs766760915 Chr.4: 152627792-152627792 on Build GRCh38
rs890442938 Chr.4: 152627526-152627526 on Build GRCh38
rs570907393 Chr.4: 152627844-152627844 on Build GRCh38
rs942200693 Chr.4: 152627827-152627827 on Build GRCh38
rs1030327627 Chr.4: 152627513-152627513 on Build GRCh38
rs769554058 Chr.4: 152627642-152627642 on Build GRCh38
rs888989927 Chr.4: 152627907-152627907 on Build GRCh38
rs1010664599 Chr.4: 152627858-152627858 on Build GRCh38
rs139683813 Chr.4: 152627503-152627503 on Build GRCh38

View Full Product Details