Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152679784-152680285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM154
Entrez Gene ID
201799
Gene Name
transmembrane protein 154
Gene Aliases
-
UniGene
Hs.518900
Gene Chromosome Location
Chr.4: 152626114-152680165 on Build GRCh38
Transcripts
NM_152680.2 XM_011531716.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6207901 c.41T>G p.I14S Chr.4: 152679893-152679893 on Build GRCh38
5883952 c.10C>T p.P4S Chr.4: 152679924-152679924 on Build GRCh38
3946068 c.44C>T p.A15V Chr.4: 152679890-152679890 on Build GRCh38
5653486 c.22C>G p.L8V Chr.4: 152679912-152679912 on Build GRCh38
1328655 c.12C>G p.P4P Chr.4: 152679922-152679922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200120689 Chr.4: 152679873-152679873 on Build GRCh38
rs529994820 Chr.4: 152680186-152680186 on Build GRCh38
rs895606205 Chr.4: 152679810-152679810 on Build GRCh38
rs1012548750 Chr.4: 152680000-152680000 on Build GRCh38
rs765240701 Chr.4: 152679904-152679904 on Build GRCh38
rs759627382 Chr.4: 152679851-152679851 on Build GRCh38
rs762841062 Chr.4: 152679949-152679950 on Build GRCh38
rs765280371 Chr.4: 152679915-152679927 on Build GRCh38
rs147648616 Chr.4: 152679929-152679929 on Build GRCh38
rs957510900 Chr.4: 152680102-152680102 on Build GRCh38
rs562455884 Chr.4: 152680164-152680164 on Build GRCh38
rs771281361 Chr.4: 152679934-152679934 on Build GRCh38
rs377270522 Chr.4: 152679847-152679847 on Build GRCh38
rs763952808 Chr.4: 152679860-152679860 on Build GRCh38
rs564601816 Chr.4: 152680028-152680028 on Build GRCh38
rs763052748 Chr.4: 152679844-152679844 on Build GRCh38
rs201751912 Chr.4: 152679941-152679941 on Build GRCh38
rs77090947 Chr.4: 152680203-152680203 on Build GRCh38
rs898064998 Chr.4: 152679949-152679949 on Build GRCh38
rs895450418 Chr.4: 152680175-152680175 on Build GRCh38
rs766340027 Chr.4: 152679915-152679915 on Build GRCh38
rs986156143 Chr.4: 152680177-152680177 on Build GRCh38
rs769355444 Chr.4: 152679893-152679893 on Build GRCh38
rs948485839 Chr.4: 152680178-152680178 on Build GRCh38
rs750275204 Chr.4: 152679867-152679867 on Build GRCh38
rs930815491 Chr.4: 152680230-152680230 on Build GRCh38
rs971144582 Chr.4: 152680048-152680048 on Build GRCh38
rs770040792 Chr.4: 152679836-152679836 on Build GRCh38
rs749687151 Chr.4: 152679885-152679885 on Build GRCh38
rs772323943 Chr.4: 152679942-152679942 on Build GRCh38
rs761627599 Chr.4: 152679898-152679898 on Build GRCh38
rs769549045 Chr.4: 152679953-152679953 on Build GRCh38
rs935961385 Chr.4: 152679899-152679899 on Build GRCh38
rs773673089 Chr.4: 152679837-152679837 on Build GRCh38
rs35466311 Chr.4: 152680203-152680204 on Build GRCh38
rs910406775 Chr.4: 152680012-152680012 on Build GRCh38
rs934570823 Chr.4: 152680135-152680135 on Build GRCh38
rs763042008 Chr.4: 152679963-152679963 on Build GRCh38
rs772129607 Chr.4: 152679890-152679890 on Build GRCh38
rs897954279 Chr.4: 152680227-152680227 on Build GRCh38
rs564860252 Chr.4: 152680150-152680150 on Build GRCh38
rs777277680 Chr.4: 152679824-152679824 on Build GRCh38
rs748756463 Chr.4: 152679829-152679829 on Build GRCh38
rs754726917 Chr.4: 152679920-152679920 on Build GRCh38
rs774367723 Chr.4: 152679849-152679849 on Build GRCh38
rs778359261 Chr.4: 152679930-152679930 on Build GRCh38
rs747637170 Chr.4: 152679880-152679880 on Build GRCh38
rs771085091 Chr.4: 152679886-152679886 on Build GRCh38
rs1043270508 Chr.4: 152680090-152680090 on Build GRCh38
rs780906698 Chr.4: 152679874-152679874 on Build GRCh38
rs748579822 Chr.4: 152679876-152679876 on Build GRCh38
rs1031535632 Chr.4: 152680087-152680087 on Build GRCh38
rs1044100965 Chr.4: 152680181-152680181 on Build GRCh38
rs543761087 Chr.4: 152680117-152680117 on Build GRCh38
rs924586980 Chr.4: 152680127-152680127 on Build GRCh38
rs888176224 Chr.4: 152680201-152680201 on Build GRCh38
rs760820637 Chr.4: 152679947-152679947 on Build GRCh38
rs1016167677 Chr.4: 152680226-152680226 on Build GRCh38
rs985516205 Chr.4: 152679971-152679971 on Build GRCh38
rs894109425 Chr.4: 152679986-152679986 on Build GRCh38
rs977211839 Chr.4: 152680058-152680058 on Build GRCh38
rs922590561 Chr.4: 152679943-152679943 on Build GRCh38
rs770371028 Chr.4: 152679879-152679879 on Build GRCh38
rs751244038 Chr.4: 152679918-152679918 on Build GRCh38
rs773708265 Chr.4: 152679927-152679927 on Build GRCh38
rs1036563567 Chr.4: 152679937-152679937 on Build GRCh38
rs1023984268 Chr.4: 152680018-152680018 on Build GRCh38
rs775395060 Chr.4: 152679945-152679945 on Build GRCh38
rs576449556 Chr.4: 152680105-152680105 on Build GRCh38
rs762842799 Chr.4: 152679910-152679910 on Build GRCh38
rs750138742 Chr.4: 152679949-152679949 on Build GRCh38
rs757936701 Chr.4: 152679869-152679869 on Build GRCh38
rs780816816 Chr.4: 152679924-152679924 on Build GRCh38
rs773192203 Chr.4: 152679959-152679959 on Build GRCh38
rs779861883 Chr.4: 152679872-152679872 on Build GRCh38
rs200601350 Chr.4: 152679864-152679864 on Build GRCh38
rs753774311 Chr.4: 152679862-152679862 on Build GRCh38
rs202190562 Chr.4: 152679888-152679888 on Build GRCh38
rs750124556 Chr.4: 152679906-152679906 on Build GRCh38
rs763001790 Chr.4: 152680073-152680073 on Build GRCh38
rs572876653 Chr.4: 152679932-152679932 on Build GRCh38
rs947229205 Chr.4: 152680038-152680038 on Build GRCh38
rs1036935772 Chr.4: 152680219-152680219 on Build GRCh38
rs1057193600 Chr.4: 152679804-152679804 on Build GRCh38

View Full Product Details